Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42798750-42799245 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTD1
Entrez Gene ID
124817
Gene Name
cyclin N-terminal domain containing 1
Gene Aliases
CNTD
UniGene
Hs.12272
Gene Chromosome Location
Chr.17: 42798667-42811587 on Build GRCh38
Transcripts
NM_173478.2 XM_005257043.4 XM_005257044.4 XM_005257045.4 XM_006721690.2 XM_011524310.2 XM_011524311.2 XM_017024171.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7285557 c.169+4C>T p.? Chr.17: 42799240-42799240 on Build GRCh38
7319589 c.132G>A p.S44S Chr.17: 42799199-42799199 on Build GRCh38
6586576 c.29C>A p.A10D Chr.17: 42799096-42799096 on Build GRCh38
1479605 c.32C>T p.S11F Chr.17: 42799099-42799099 on Build GRCh38
6631630 c.75T>C p.I25I Chr.17: 42799142-42799142 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748298985 Chr.17: 42799017-42799017 on Build GRCh38
rs764826119 Chr.17: 42798776-42798776 on Build GRCh38
rs777999566 Chr.17: 42799211-42799211 on Build GRCh38
rs72826975 Chr.17: 42799012-42799012 on Build GRCh38
rs919414948 Chr.17: 42798824-42798824 on Build GRCh38
rs922721478 Chr.17: 42799082-42799082 on Build GRCh38
rs963130921 Chr.17: 42798814-42798814 on Build GRCh38
rs890057069 Chr.17: 42798780-42798780 on Build GRCh38
rs916133012 Chr.17: 42798952-42798952 on Build GRCh38
rs377213591 Chr.17: 42798970-42798970 on Build GRCh38
rs61757410 Chr.17: 42798911-42798911 on Build GRCh38
rs762595375 Chr.17: 42799073-42799073 on Build GRCh38
rs560166227 Chr.17: 42798922-42798922 on Build GRCh38
rs774031094 Chr.17: 42799189-42799189 on Build GRCh38
rs748859760 Chr.17: 42798971-42798971 on Build GRCh38
rs140701179 Chr.17: 42799132-42799132 on Build GRCh38
rs745593087 Chr.17: 42798811-42798811 on Build GRCh38
rs759842089 Chr.17: 42799044-42799044 on Build GRCh38
rs542803148 Chr.17: 42798945-42798945 on Build GRCh38
rs767611711 Chr.17: 42799193-42799193 on Build GRCh38
rs1052736400 Chr.17: 42799068-42799068 on Build GRCh38
rs771359074 Chr.17: 42799039-42799039 on Build GRCh38
rs542028382 Chr.17: 42799015-42799015 on Build GRCh38
rs1049941569 Chr.17: 42799098-42799098 on Build GRCh38
rs758586396 Chr.17: 42799110-42799110 on Build GRCh38
rs771447116 Chr.17: 42799151-42799151 on Build GRCh38
rs781752330 Chr.17: 42799227-42799227 on Build GRCh38
rs61757408 Chr.17: 42798775-42798775 on Build GRCh38
rs766100218 Chr.17: 42799203-42799203 on Build GRCh38
rs12948925 Chr.17: 42799214-42799214 on Build GRCh38
rs777008722 Chr.17: 42799040-42799040 on Build GRCh38
rs551890376 Chr.17: 42798787-42798787 on Build GRCh38
rs535342634 Chr.17: 42799204-42799204 on Build GRCh38
rs757820217 Chr.17: 42798784-42798784 on Build GRCh38
rs773046639 Chr.17: 42799164-42799164 on Build GRCh38
rs997268221 Chr.17: 42799077-42799077 on Build GRCh38
rs1015917645 Chr.17: 42799128-42799128 on Build GRCh38
rs896851686 Chr.17: 42798862-42798862 on Build GRCh38
rs200907333 Chr.17: 42799107-42799107 on Build GRCh38
rs775274061 Chr.17: 42799183-42799183 on Build GRCh38
rs911415223 Chr.17: 42799127-42799127 on Build GRCh38
rs370557977 Chr.17: 42799213-42799213 on Build GRCh38
rs773826186 Chr.17: 42799030-42799030 on Build GRCh38
rs767523581 Chr.17: 42799088-42799088 on Build GRCh38
rs565774899 Chr.17: 42799138-42799138 on Build GRCh38
rs61995867 Chr.17: 42799089-42799089 on Build GRCh38
rs556301304 Chr.17: 42798816-42798816 on Build GRCh38
rs779715186 Chr.17: 42798953-42798953 on Build GRCh38
rs561280611 Chr.17: 42798959-42798959 on Build GRCh38
rs747526574 Chr.17: 42799038-42799038 on Build GRCh38
rs746267539 Chr.17: 42799160-42799160 on Build GRCh38
rs572349256 Chr.17: 42798935-42798935 on Build GRCh38
rs994312229 Chr.17: 42798905-42798905 on Build GRCh38
rs747049643 Chr.17: 42799148-42799148 on Build GRCh38
rs576984150 Chr.17: 42799090-42799090 on Build GRCh38
rs925970801 Chr.17: 42799221-42799221 on Build GRCh38
rs546514347 Chr.17: 42798823-42798823 on Build GRCh38
rs201778369 Chr.17: 42799199-42799199 on Build GRCh38
rs137860677 Chr.17: 42799191-42799191 on Build GRCh38
rs200889328 Chr.17: 42799212-42799212 on Build GRCh38
rs967954768 Chr.17: 42798994-42798994 on Build GRCh38
rs145554888 Chr.17: 42798797-42798797 on Build GRCh38
rs752271012 Chr.17: 42798779-42798779 on Build GRCh38
rs1023478580 Chr.17: 42798993-42798993 on Build GRCh38
rs545178326 Chr.17: 42798920-42798920 on Build GRCh38
rs772040170 Chr.17: 42799025-42799025 on Build GRCh38
rs762529202 Chr.17: 42799185-42799185 on Build GRCh38
rs779261716 Chr.17: 42799094-42799094 on Build GRCh38
rs764136234 Chr.17: 42798786-42798786 on Build GRCh38
rs925948229 Chr.17: 42799018-42799018 on Build GRCh38
rs12948919 Chr.17: 42799206-42799206 on Build GRCh38
rs757582857 Chr.17: 42798804-42798804 on Build GRCh38
rs979202426 Chr.17: 42799220-42799220 on Build GRCh38
rs934752269 Chr.17: 42799052-42799052 on Build GRCh38
rs142394310 Chr.17: 42799074-42799074 on Build GRCh38
rs61757409 Chr.17: 42798901-42798901 on Build GRCh38
rs972671079 Chr.17: 42798817-42798817 on Build GRCh38
rs199509114 Chr.17: 42799198-42799198 on Build GRCh38
rs563022349 Chr.17: 42798892-42798892 on Build GRCh38
rs757267570 Chr.17: 42799080-42799080 on Build GRCh38
rs12948912 Chr.17: 42799195-42799195 on Build GRCh38
rs757179702 Chr.17: 42798800-42798800 on Build GRCh38
rs764914124 Chr.17: 42799054-42799054 on Build GRCh38
rs554946865 Chr.17: 42798774-42798774 on Build GRCh38
rs774979463 Chr.17: 42799061-42799061 on Build GRCh38
rs777096742 Chr.17: 42799158-42799158 on Build GRCh38
rs145896591 Chr.17: 42799187-42799187 on Build GRCh38

View Full Product Details