Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31659787-31660288 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAG6
Entrez Gene ID
7917
Gene Name
BCL2 associated athanogene 6
Gene Aliases
BAG-6, BAT3, D6S52E, G3
UniGene
Hs.440900
Gene Chromosome Location
Chr.6: 31639028-31660900 on Build GRCh38
Transcripts
XM_017011279.1 NM_021184.3

Gene Symbol
C6orf47
Entrez Gene ID
57827
Gene Name
chromosome 6 open reading frame 47
Gene Aliases
D6S53E, G4, NG34
UniGene
Hs.247323
Gene Chromosome Location
Chr.6: 31658298-31660772 on Build GRCh38
Transcripts
XM_017011279.1 NM_021184.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6360607 c.137C>T p.A46V Chr.6: 31659811-31659811 on Build GRCh38
3253815 c.111G>A p.E37E Chr.6: 31659837-31659837 on Build GRCh38
4390674 c.107C>G p.S36W Chr.6: 31659841-31659841 on Build GRCh38
3624669 c.151G>A p.E51K Chr.6: 31659797-31659797 on Build GRCh38
6411308 c.11G>T p.R4L Chr.6: 31659937-31659937 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755751657 Chr.6: 31659889-31659889 on Build GRCh38
rs768703098 Chr.6: 31659929-31659929 on Build GRCh38
rs774283275 Chr.6: 31659873-31659873 on Build GRCh38
rs771110459 Chr.6: 31659914-31659914 on Build GRCh38
rs917905325 Chr.6: 31660041-31660041 on Build GRCh38
rs1051929852 Chr.6: 31660202-31660202 on Build GRCh38
rs539538086 Chr.6: 31660111-31660111 on Build GRCh38
rs114159073 Chr.6: 31660012-31660012 on Build GRCh38
rs776741995 Chr.6: 31659918-31659918 on Build GRCh38
rs1013527403 Chr.6: 31660237-31660237 on Build GRCh38
rs774374938 Chr.6: 31659938-31659938 on Build GRCh38
rs897826271 Chr.6: 31660127-31660127 on Build GRCh38
rs925489350 Chr.6: 31660078-31660078 on Build GRCh38
rs764168304 Chr.6: 31659869-31659869 on Build GRCh38
rs971832054 Chr.6: 31659963-31659963 on Build GRCh38
rs944759317 Chr.6: 31660057-31660057 on Build GRCh38
rs142552983 Chr.6: 31659863-31659863 on Build GRCh38
rs758697768 Chr.6: 31659903-31659903 on Build GRCh38
rs1007246410 Chr.6: 31659902-31659902 on Build GRCh38
rs990591472 Chr.6: 31659837-31659837 on Build GRCh38
rs772274930 Chr.6: 31659842-31659842 on Build GRCh38
rs1050552098 Chr.6: 31660036-31660036 on Build GRCh38
rs890567543 Chr.6: 31660050-31660050 on Build GRCh38
rs762965122 Chr.6: 31659867-31659867 on Build GRCh38
rs746012019 Chr.6: 31659921-31659921 on Build GRCh38
rs778031995 Chr.6: 31659909-31659909 on Build GRCh38
rs761844940 Chr.6: 31659983-31659983 on Build GRCh38
rs747204088 Chr.6: 31659852-31659852 on Build GRCh38
rs948024038 Chr.6: 31660060-31660060 on Build GRCh38
rs761760799 Chr.6: 31659876-31659876 on Build GRCh38
rs149664554 Chr.6: 31659890-31659890 on Build GRCh38
rs1024416706 Chr.6: 31660244-31660244 on Build GRCh38
rs1052602059 Chr.6: 31660130-31660130 on Build GRCh38
rs1016107578 Chr.6: 31659935-31659935 on Build GRCh38
rs768780232 Chr.6: 31659840-31659840 on Build GRCh38
rs892672059 Chr.6: 31660137-31660137 on Build GRCh38
rs150937586 Chr.6: 31659877-31659877 on Build GRCh38
rs1007603560 Chr.6: 31660231-31660231 on Build GRCh38
rs959085371 Chr.6: 31659827-31659827 on Build GRCh38
rs773538120 Chr.6: 31659846-31659846 on Build GRCh38
rs375377600 Chr.6: 31659899-31659899 on Build GRCh38
rs963273525 Chr.6: 31659947-31659947 on Build GRCh38
rs773366379 Chr.6: 31659829-31659829 on Build GRCh38
rs750023896 Chr.6: 31659888-31659888 on Build GRCh38
rs1026146908 Chr.6: 31659999-31659999 on Build GRCh38
rs761227440 Chr.6: 31659870-31659871 on Build GRCh38
rs75199370 Chr.6: 31660181-31660181 on Build GRCh38
rs888884794 Chr.6: 31660204-31660204 on Build GRCh38
rs970486875 Chr.6: 31660258-31660259 on Build GRCh38
rs755497677 Chr.6: 31659816-31659816 on Build GRCh38
rs992682419 Chr.6: 31660025-31660025 on Build GRCh38
rs1003246539 Chr.6: 31660261-31660261 on Build GRCh38
rs747209746 Chr.6: 31659913-31659913 on Build GRCh38
rs771004756 Chr.6: 31659857-31659857 on Build GRCh38
rs755442578 Chr.6: 31659898-31659898 on Build GRCh38
rs933070214 Chr.6: 31660179-31660179 on Build GRCh38
rs980808091 Chr.6: 31660067-31660067 on Build GRCh38
rs748548565 Chr.6: 31659841-31659841 on Build GRCh38
rs546246661 Chr.6: 31659901-31659901 on Build GRCh38

View Full Product Details