Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151051019-151051521 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASIC3
Entrez Gene ID
9311
Gene Name
acid sensing ion channel subunit 3
Gene Aliases
ACCN3, DRASIC, SLNAC1, TNaC1
UniGene
Hs.647113
Gene Chromosome Location
Chr.7: 151048292-151052756 on Build GRCh38
Transcripts
NM_004769.3 NM_020321.3 NM_020322.3 NR_046401.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4663719 c.1134G>A p.E378E Chr.7: 151051239-151051239 on Build GRCh38
5701130 c.1083_1084insG p.D362fs*55 Chr.7: 151051188-151051189 on Build GRCh38
6371685 c.1205C>A p.A402D Chr.7: 151051310-151051310 on Build GRCh38
6392425 c.1097G>T p.C366F Chr.7: 151051202-151051202 on Build GRCh38
6109596 c.1071C>A p.A357A Chr.7: 151051176-151051176 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201478022 Chr.7: 151051207-151051207 on Build GRCh38
rs748579784 Chr.7: 151051067-151051067 on Build GRCh38
rs766863604 Chr.7: 151051081-151051081 on Build GRCh38
rs940412648 Chr.7: 151051331-151051331 on Build GRCh38
rs367577120 Chr.7: 151051166-151051166 on Build GRCh38
rs956906133 Chr.7: 151051470-151051470 on Build GRCh38
rs992242612 Chr.7: 151051213-151051213 on Build GRCh38
rs776359742 Chr.7: 151051092-151051092 on Build GRCh38
rs368418085 Chr.7: 151051112-151051112 on Build GRCh38
rs756979055 Chr.7: 151051120-151051120 on Build GRCh38
rs747610290 Chr.7: 151051276-151051276 on Build GRCh38
rs776358418 Chr.7: 151051058-151051058 on Build GRCh38
rs1015933276 Chr.7: 151051144-151051144 on Build GRCh38
rs752155567 Chr.7: 151051196-151051207 on Build GRCh38
rs751732928 Chr.7: 151051252-151051252 on Build GRCh38
rs867527166 Chr.7: 151051262-151051262 on Build GRCh38
rs904418739 Chr.7: 151051359-151051359 on Build GRCh38
rs758417400 Chr.7: 151051259-151051259 on Build GRCh38
rs780339201 Chr.7: 151051269-151051269 on Build GRCh38
rs773135045 Chr.7: 151051286-151051286 on Build GRCh38
rs768140923 Chr.7: 151051217-151051217 on Build GRCh38
rs939922186 Chr.7: 151051207-151051208 on Build GRCh38
rs778829040 Chr.7: 151051132-151051132 on Build GRCh38
rs757875390 Chr.7: 151051113-151051113 on Build GRCh38
rs560303040 Chr.7: 151051110-151051110 on Build GRCh38
rs762660247 Chr.7: 151051216-151051216 on Build GRCh38
rs533277404 Chr.7: 151051073-151051073 on Build GRCh38
rs759214498 Chr.7: 151051081-151051084 on Build GRCh38
rs548026567 Chr.7: 151051288-151051288 on Build GRCh38
rs757336992 Chr.7: 151051335-151051335 on Build GRCh38
rs1034125844 Chr.7: 151051392-151051392 on Build GRCh38
rs780187832 Chr.7: 151051050-151051050 on Build GRCh38
rs755276320 Chr.7: 151051130-151051130 on Build GRCh38
rs751841571 Chr.7: 151051328-151051328 on Build GRCh38
rs571132120 Chr.7: 151051353-151051353 on Build GRCh38
rs750271120 Chr.7: 151051167-151051167 on Build GRCh38
rs761796285 Chr.7: 151051164-151051164 on Build GRCh38
rs757431074 Chr.7: 151051253-151051253 on Build GRCh38
rs909436137 Chr.7: 151051229-151051229 on Build GRCh38
rs751508282 Chr.7: 151051119-151051119 on Build GRCh38
rs769444157 Chr.7: 151051153-151051153 on Build GRCh38
rs758486487 Chr.7: 151051184-151051184 on Build GRCh38
rs747527437 Chr.7: 151051146-151051146 on Build GRCh38
rs760460254 Chr.7: 151051226-151051226 on Build GRCh38
rs527873441 Chr.7: 151051271-151051271 on Build GRCh38
rs1025022200 Chr.7: 151051405-151051405 on Build GRCh38
rs371018510 Chr.7: 151051159-151051159 on Build GRCh38
rs200502346 Chr.7: 151051044-151051044 on Build GRCh38
rs775024711 Chr.7: 151051158-151051158 on Build GRCh38
rs1010934475 Chr.7: 151051498-151051498 on Build GRCh38
rs771618030 Chr.7: 151051188-151051188 on Build GRCh38
rs1001435942 Chr.7: 151051373-151051373 on Build GRCh38
rs763966968 Chr.7: 151051117-151051117 on Build GRCh38
rs540130534 Chr.7: 151051383-151051383 on Build GRCh38
rs771498570 Chr.7: 151051277-151051277 on Build GRCh38
rs565807969 Chr.7: 151051232-151051232 on Build GRCh38
rs767557099 Chr.7: 151051348-151051348 on Build GRCh38
rs749455807 Chr.7: 151051126-151051126 on Build GRCh38
rs775138378 Chr.7: 151051208-151051208 on Build GRCh38
rs1004981898 Chr.7: 151051437-151051437 on Build GRCh38
rs764374817 Chr.7: 151051325-151051325 on Build GRCh38
rs371031278 Chr.7: 151051049-151051049 on Build GRCh38
rs762585526 Chr.7: 151051145-151051149 on Build GRCh38
rs756493982 Chr.7: 151051263-151051263 on Build GRCh38
rs771422088 Chr.7: 151051148-151051148 on Build GRCh38
rs764874352 Chr.7: 151051123-151051132 on Build GRCh38
rs763239303 Chr.7: 151051294-151051294 on Build GRCh38
rs767450510 Chr.7: 151051221-151051221 on Build GRCh38
rs779149324 Chr.7: 151051177-151051177 on Build GRCh38
rs374624215 Chr.7: 151051160-151051160 on Build GRCh38
rs752303895 Chr.7: 151051108-151051108 on Build GRCh38
rs777114538 Chr.7: 151051150-151051150 on Build GRCh38
rs772676029 Chr.7: 151051070-151051070 on Build GRCh38
rs898859828 Chr.7: 151051155-151051155 on Build GRCh38
rs752962315 Chr.7: 151051245-151051245 on Build GRCh38
rs781382564 Chr.7: 151051197-151051197 on Build GRCh38
rs921599598 Chr.7: 151051264-151051264 on Build GRCh38
rs549463375 Chr.7: 151051139-151051139 on Build GRCh38
rs750351543 Chr.7: 151051222-151051222 on Build GRCh38
rs758755360 Chr.7: 151051246-151051246 on Build GRCh38
rs867061610 Chr.7: 151051210-151051210 on Build GRCh38
rs963624902 Chr.7: 151051457-151051457 on Build GRCh38
rs1014129299 Chr.7: 151051396-151051396 on Build GRCh38
rs774545752 Chr.7: 151051061-151051061 on Build GRCh38
rs781032690 Chr.7: 151051122-151051122 on Build GRCh38
rs1016366331 Chr.7: 151051456-151051456 on Build GRCh38
rs933099289 Chr.7: 151051268-151051268 on Build GRCh38
rs905685136 Chr.7: 151051119-151051119 on Build GRCh38
rs764641723 Chr.7: 151051106-151051106 on Build GRCh38
rs755894575 Chr.7: 151051172-151051172 on Build GRCh38
rs757817730 Chr.7: 151051206-151051222 on Build GRCh38
rs759303666 Chr.7: 151051102-151051102 on Build GRCh38
rs760597883 Chr.7: 151051287-151051287 on Build GRCh38
rs746191573 Chr.7: 151051199-151051199 on Build GRCh38
rs759091268 Chr.7: 151051151-151051151 on Build GRCh38
rs1033783177 Chr.7: 151051194-151051194 on Build GRCh38
rs556245886 Chr.7: 151051453-151051453 on Build GRCh38
rs956789598 Chr.7: 151051206-151051206 on Build GRCh38
rs950958533 Chr.7: 151051423-151051423 on Build GRCh38
rs752166306 Chr.7: 151051134-151051138 on Build GRCh38
rs774022451 Chr.7: 151051272-151051272 on Build GRCh38
rs752875882 Chr.7: 151051181-151051181 on Build GRCh38
rs374468932 Chr.7: 151051041-151051041 on Build GRCh38
rs777980557 Chr.7: 151051247-151051247 on Build GRCh38
rs749528853 Chr.7: 151051051-151051051 on Build GRCh38
rs148801982 Chr.7: 151051143-151051143 on Build GRCh38
rs142443605 Chr.7: 151051278-151051278 on Build GRCh38
rs747509313 Chr.7: 151051187-151051187 on Build GRCh38
rs750476920 Chr.7: 151051218-151051218 on Build GRCh38
rs867076859 Chr.7: 151051205-151051205 on Build GRCh38
rs778069551 Chr.7: 151051186-151051186 on Build GRCh38
rs772300980 Chr.7: 151051142-151051142 on Build GRCh38
rs763510986 Chr.7: 151051192-151051193 on Build GRCh38
rs768614368 Chr.7: 151051055-151051055 on Build GRCh38
rs895143471 Chr.7: 151051393-151051393 on Build GRCh38

View Full Product Details