Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 29334045-29334538 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP4
Entrez Gene ID
1846
Gene Name
dual specificity phosphatase 4
Gene Aliases
HVH2, MKP-2, MKP2, TYP
UniGene
Hs.417962
Gene Chromosome Location
Chr.8: 29333062-29350750 on Build GRCh38
Transcripts
NM_001394.6 NM_057158.3 XM_011544428.2

SNP Detail

SNP ID Chromosome Location
rs555492915 Chr.8: 29334083-29334083 on Build GRCh38
rs543611895 Chr.8: 29334311-29334311 on Build GRCh38
rs755089997 Chr.8: 29334159-29334159 on Build GRCh38
rs28539936 Chr.8: 29334121-29334121 on Build GRCh38
rs7017547 Chr.8: 29334359-29334359 on Build GRCh38
rs933247937 Chr.8: 29334379-29334379 on Build GRCh38
rs950882695 Chr.8: 29334508-29334508 on Build GRCh38
rs971035528 Chr.8: 29334335-29334335 on Build GRCh38
rs1002163813 Chr.8: 29334450-29334450 on Build GRCh38
rs6993871 Chr.8: 29334187-29334187 on Build GRCh38
rs892523944 Chr.8: 29334122-29334122 on Build GRCh38
rs895304934 Chr.8: 29334217-29334217 on Build GRCh38
rs947244151 Chr.8: 29334437-29334437 on Build GRCh38
rs659481 Chr.8: 29334113-29334113 on Build GRCh38
rs923190615 Chr.8: 29334360-29334360 on Build GRCh38
rs540449468 Chr.8: 29334336-29334336 on Build GRCh38
rs896708079 Chr.8: 29334456-29334456 on Build GRCh38
rs943230999 Chr.8: 29334409-29334409 on Build GRCh38
rs984486408 Chr.8: 29334319-29334319 on Build GRCh38
rs1042340065 Chr.8: 29334472-29334472 on Build GRCh38
rs992314510 Chr.8: 29334291-29334291 on Build GRCh38
rs768798897 Chr.8: 29334193-29334193 on Build GRCh38
rs903943818 Chr.8: 29334442-29334442 on Build GRCh38
rs1015927838 Chr.8: 29334510-29334510 on Build GRCh38
rs891606570 Chr.8: 29334407-29334407 on Build GRCh38
rs780348843 Chr.8: 29334175-29334175 on Build GRCh38
rs528731997 Chr.8: 29334223-29334223 on Build GRCh38
rs375984116 Chr.8: 29334085-29334085 on Build GRCh38
rs577862620 Chr.8: 29334244-29334244 on Build GRCh38
rs139544751 Chr.8: 29334306-29334306 on Build GRCh38
rs775021086 Chr.8: 29334265-29334265 on Build GRCh38
rs1033894558 Chr.8: 29334203-29334203 on Build GRCh38
rs1026592321 Chr.8: 29334488-29334488 on Build GRCh38
rs901765913 Chr.8: 29334090-29334090 on Build GRCh38
rs144527402 Chr.8: 29334402-29334402 on Build GRCh38
rs551231036 Chr.8: 29334498-29334498 on Build GRCh38
rs773603573 Chr.8: 29334509-29334509 on Build GRCh38
rs1014242413 Chr.8: 29334463-29334463 on Build GRCh38
rs945312448 Chr.8: 29334460-29334460 on Build GRCh38
rs1013831784 Chr.8: 29334225-29334225 on Build GRCh38
rs950940784 Chr.8: 29334312-29334312 on Build GRCh38
rs748379754 Chr.8: 29334438-29334438 on Build GRCh38
rs964448935 Chr.8: 29334513-29334513 on Build GRCh38
rs111245274 Chr.8: 29334356-29334356 on Build GRCh38
rs868293951 Chr.8: 29334352-29334352 on Build GRCh38
rs945407180 Chr.8: 29334126-29334126 on Build GRCh38
rs943030646 Chr.8: 29334347-29334347 on Build GRCh38
rs984950201 Chr.8: 29334368-29334368 on Build GRCh38
rs951795977 Chr.8: 29334365-29334365 on Build GRCh38
rs572475749 Chr.8: 29334087-29334087 on Build GRCh38
rs528177531 Chr.8: 29334464-29334464 on Build GRCh38
rs1034112874 Chr.8: 29334452-29334452 on Build GRCh38
rs6983785 Chr.8: 29334441-29334441 on Build GRCh38
rs961249006 Chr.8: 29334228-29334228 on Build GRCh38
rs1009643568 Chr.8: 29334123-29334123 on Build GRCh38
rs997319735 Chr.8: 29334100-29334100 on Build GRCh38

View Full Product Details