Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 72206125-72206668 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERCC6L
Entrez Gene ID
54821
Gene Name
ERCC excision repair 6 like, spindle assembly checkpoint helicase
Gene Aliases
PICH, RAD26L
UniGene
Hs.47558
Gene Chromosome Location
Chr.X: 72204657-72239008 on Build GRCh38
Transcripts
NM_017669.2 NM_001170747.1

Gene Symbol
PIN4
Entrez Gene ID
5303
Gene Name
peptidylprolyl cis/trans isomerase, NIMA-interacting 4
Gene Aliases
EPVH, PAR14, PAR17
UniGene
Hs.655623
Gene Chromosome Location
Chr.X: 72181676-72263964 on Build GRCh38
Transcripts
NM_017669.2 NM_001170747.1

SNP Detail

SNP ID Chromosome Location
rs767437755 Chr.X: 72206363-72206363 on Build GRCh38
rs780115534 Chr.X: 72206433-72206433 on Build GRCh38
rs183569692 Chr.X: 72206333-72206333 on Build GRCh38
rs773634562 Chr.X: 72206481-72206481 on Build GRCh38
rs200118335 Chr.X: 72206375-72206375 on Build GRCh38
rs377308473 Chr.X: 72206169-72206169 on Build GRCh38
rs140936418 Chr.X: 72206166-72206166 on Build GRCh38
rs755042517 Chr.X: 72206439-72206439 on Build GRCh38
rs45448501 Chr.X: 72206491-72206491 on Build GRCh38
rs751204906 Chr.X: 72206345-72206345 on Build GRCh38
rs750234088 Chr.X: 72206400-72206400 on Build GRCh38
rs958041304 Chr.X: 72206644-72206644 on Build GRCh38
rs763537928 Chr.X: 72206584-72206584 on Build GRCh38
rs763850827 Chr.X: 72206329-72206329 on Build GRCh38
rs1026752035 Chr.X: 72206538-72206538 on Build GRCh38
rs767335608 Chr.X: 72206318-72206318 on Build GRCh38
rs934837493 Chr.X: 72206160-72206160 on Build GRCh38
rs761174938 Chr.X: 72206279-72206279 on Build GRCh38
rs779001189 Chr.X: 72206446-72206446 on Build GRCh38
rs760051206 Chr.X: 72206564-72206564 on Build GRCh38
rs780344606 Chr.X: 72206186-72206186 on Build GRCh38
rs749362238 Chr.X: 72206199-72206199 on Build GRCh38
rs776513413 Chr.X: 72206646-72206646 on Build GRCh38
rs200234671 Chr.X: 72206627-72206627 on Build GRCh38
rs138817796 Chr.X: 72206226-72206226 on Build GRCh38
rs149400438 Chr.X: 72206231-72206231 on Build GRCh38
rs373087197 Chr.X: 72206426-72206426 on Build GRCh38
rs138539407 Chr.X: 72206437-72206437 on Build GRCh38
rs143780574 Chr.X: 72206248-72206248 on Build GRCh38
rs761435400 Chr.X: 72206602-72206602 on Build GRCh38
rs771530996 Chr.X: 72206298-72206300 on Build GRCh38
rs768941124 Chr.X: 72206328-72206328 on Build GRCh38
rs369492714 Chr.X: 72206509-72206509 on Build GRCh38
rs750289131 Chr.X: 72206636-72206636 on Build GRCh38
rs745826258 Chr.X: 72206170-72206170 on Build GRCh38
rs773857138 Chr.X: 72206599-72206599 on Build GRCh38
rs1011305166 Chr.X: 72206201-72206201 on Build GRCh38
rs1035607206 Chr.X: 72206340-72206340 on Build GRCh38
rs368694497 Chr.X: 72206288-72206288 on Build GRCh38
rs756889450 Chr.X: 72206347-72206347 on Build GRCh38
rs768969578 Chr.X: 72206225-72206225 on Build GRCh38
rs748332306 Chr.X: 72206462-72206462 on Build GRCh38
rs748360100 Chr.X: 72206227-72206227 on Build GRCh38
rs777094514 Chr.X: 72206518-72206518 on Build GRCh38
rs777279256 Chr.X: 72206162-72206162 on Build GRCh38
rs865972418 Chr.X: 72206596-72206596 on Build GRCh38
rs150211194 Chr.X: 72206183-72206183 on Build GRCh38
rs759241784 Chr.X: 72206315-72206315 on Build GRCh38
rs375177556 Chr.X: 72206479-72206479 on Build GRCh38
rs767261847 Chr.X: 72206613-72206613 on Build GRCh38
rs755940524 Chr.X: 72206637-72206637 on Build GRCh38

View Full Product Details