Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42100927-42101495 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX58
Entrez Gene ID
79132
Gene Name
DEXH-box helicase 58
Gene Aliases
D11LGP2, D11lgp2e, LGP2, RLR-3
UniGene
Hs.55918
Gene Chromosome Location
Chr.17: 42101404-42112733 on Build GRCh38
Transcripts
NM_024119.2 XM_017025059.1 XM_017025060.1

SNP Detail

SNP ID Chromosome Location
rs906943275 Chr.17: 42101454-42101454 on Build GRCh38
rs782665759 Chr.17: 42100996-42100996 on Build GRCh38
rs1013775222 Chr.17: 42101091-42101091 on Build GRCh38
rs749259391 Chr.17: 42101029-42101029 on Build GRCh38
rs971144006 Chr.17: 42101113-42101113 on Build GRCh38
rs913089556 Chr.17: 42100993-42100993 on Build GRCh38
rs575848004 Chr.17: 42100982-42100982 on Build GRCh38
rs912615230 Chr.17: 42101230-42101230 on Build GRCh38
rs548834321 Chr.17: 42101170-42101170 on Build GRCh38
rs931546423 Chr.17: 42101301-42101301 on Build GRCh38
rs148412952 Chr.17: 42101175-42101175 on Build GRCh38
rs530345695 Chr.17: 42101116-42101116 on Build GRCh38
rs898737053 Chr.17: 42101071-42101071 on Build GRCh38
rs1046634983 Chr.17: 42101402-42101402 on Build GRCh38
rs1024021508 Chr.17: 42101102-42101102 on Build GRCh38
rs1051097602 Chr.17: 42101011-42101011 on Build GRCh38
rs986601256 Chr.17: 42101220-42101220 on Build GRCh38
rs577413116 Chr.17: 42101073-42101073 on Build GRCh38
rs1020168796 Chr.17: 42101206-42101206 on Build GRCh38
rs782346279 Chr.17: 42101347-42101347 on Build GRCh38
rs565334095 Chr.17: 42101049-42101049 on Build GRCh38
rs191479936 Chr.17: 42101325-42101325 on Build GRCh38
rs200213922 Chr.17: 42101015-42101015 on Build GRCh38
rs1039716925 Chr.17: 42101035-42101035 on Build GRCh38
rs1047316935 Chr.17: 42101321-42101321 on Build GRCh38
rs967158889 Chr.17: 42101219-42101219 on Build GRCh38
rs543464390 Chr.17: 42101025-42101025 on Build GRCh38
rs949497513 Chr.17: 42101397-42101397 on Build GRCh38
rs555468646 Chr.17: 42101221-42101221 on Build GRCh38
rs551886865 Chr.17: 42101263-42101263 on Build GRCh38
rs782650735 Chr.17: 42101169-42101169 on Build GRCh38
rs942834716 Chr.17: 42101241-42101241 on Build GRCh38
rs144438858 Chr.17: 42101085-42101085 on Build GRCh38
rs11869776 Chr.17: 42101078-42101078 on Build GRCh38
rs957496105 Chr.17: 42101174-42101174 on Build GRCh38
rs887302023 Chr.17: 42101084-42101084 on Build GRCh38
rs571944310 Chr.17: 42101404-42101404 on Build GRCh38

View Full Product Details