Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 96936826-96937376 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUTM2G
Entrez Gene ID
441457
Gene Name
NUT family member 2G
Gene Aliases
FAM22G, NUTMG
UniGene
Hs.652383
Gene Chromosome Location
Chr.9: 96929004-96942290 on Build GRCh38
Transcripts
NM_001045477.2 NM_001170741.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6997395 c.1091delC p.P364fs*56 Chr.9: 96937172-96937172 on Build GRCh38
7127803 c.1071C>T p.H357H Chr.9: 96937152-96937152 on Build GRCh38
1490247 c.1180C>T p.H394Y Chr.9: 96937261-96937261 on Build GRCh38
6901004 c.1213C>T p.R405W Chr.9: 96937294-96937294 on Build GRCh38
7273268 c.1192A>G p.T398A Chr.9: 96937273-96937273 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375305559 Chr.9: 96937016-96937018 on Build GRCh38
rs776042760 Chr.9: 96937137-96937137 on Build GRCh38
rs780266308 Chr.9: 96937323-96937323 on Build GRCh38
rs965672598 Chr.9: 96937281-96937281 on Build GRCh38
rs748294103 Chr.9: 96937014-96937014 on Build GRCh38
rs1023339999 Chr.9: 96936892-96936892 on Build GRCh38
rs756726972 Chr.9: 96937341-96937341 on Build GRCh38
rs934620412 Chr.9: 96936989-96936989 on Build GRCh38
rs753546875 Chr.9: 96937059-96937059 on Build GRCh38
rs769107127 Chr.9: 96937200-96937200 on Build GRCh38
rs773029494 Chr.9: 96937220-96937220 on Build GRCh38
rs773486956 Chr.9: 96937021-96937021 on Build GRCh38
rs369907174 Chr.9: 96937283-96937283 on Build GRCh38
rs374524334 Chr.9: 96937157-96937157 on Build GRCh38
rs765628592 Chr.9: 96937168-96937168 on Build GRCh38
rs748171656 Chr.9: 96937095-96937095 on Build GRCh38
rs758107459 Chr.9: 96937206-96937206 on Build GRCh38
rs1049006361 Chr.9: 96937230-96937230 on Build GRCh38
rs765048672 Chr.9: 96937064-96937064 on Build GRCh38
rs969988303 Chr.9: 96936908-96936908 on Build GRCh38
rs762786720 Chr.9: 96937221-96937221 on Build GRCh38
rs777264054 Chr.9: 96937326-96937327 on Build GRCh38
rs528515556 Chr.9: 96936931-96936931 on Build GRCh38
rs546969130 Chr.9: 96936934-96936934 on Build GRCh38
rs998907176 Chr.9: 96936849-96936849 on Build GRCh38
rs759265326 Chr.9: 96937310-96937310 on Build GRCh38
rs780152313 Chr.9: 96937087-96937087 on Build GRCh38
rs776189451 Chr.9: 96937206-96937206 on Build GRCh38
rs183480576 Chr.9: 96936882-96936882 on Build GRCh38
rs904528870 Chr.9: 96936960-96936960 on Build GRCh38
rs752611892 Chr.9: 96937314-96937314 on Build GRCh38
rs775878307 Chr.9: 96937190-96937190 on Build GRCh38
rs2986890 Chr.9: 96937182-96937182 on Build GRCh38
rs1027341536 Chr.9: 96936912-96936912 on Build GRCh38
rs201133792 Chr.9: 96937146-96937146 on Build GRCh38
rs913343 Chr.9: 96936847-96936847 on Build GRCh38
rs2479295 Chr.9: 96937338-96937338 on Build GRCh38
rs921454377 Chr.9: 96937127-96937127 on Build GRCh38
rs746167589 Chr.9: 96937178-96937178 on Build GRCh38
rs539208065 Chr.9: 96937025-96937025 on Build GRCh38
rs375745598 Chr.9: 96937267-96937267 on Build GRCh38
rs199979789 Chr.9: 96937136-96937136 on Build GRCh38
rs994778568 Chr.9: 96937033-96937033 on Build GRCh38
rs936713891 Chr.9: 96936913-96936924 on Build GRCh38
rs774184994 Chr.9: 96937306-96937306 on Build GRCh38
rs371380119 Chr.9: 96937224-96937224 on Build GRCh38
rs200128017 Chr.9: 96936966-96936966 on Build GRCh38
rs755326305 Chr.9: 96937137-96937139 on Build GRCh38
rs202193995 Chr.9: 96937332-96937332 on Build GRCh38
rs1044846318 Chr.9: 96937195-96937195 on Build GRCh38
rs375201513 Chr.9: 96937319-96937319 on Build GRCh38
rs562519519 Chr.9: 96937187-96937187 on Build GRCh38
rs370528929 Chr.9: 96937302-96937302 on Build GRCh38
rs747388231 Chr.9: 96937278-96937278 on Build GRCh38
rs751440589 Chr.9: 96937227-96937227 on Build GRCh38
rs758394818 Chr.9: 96937082-96937082 on Build GRCh38
rs760007563 Chr.9: 96937040-96937040 on Build GRCh38
rs774694681 Chr.9: 96937102-96937102 on Build GRCh38
rs756904941 Chr.9: 96937061-96937061 on Build GRCh38
rs748866488 Chr.9: 96937348-96937348 on Build GRCh38
rs201137010 Chr.9: 96937322-96937322 on Build GRCh38
rs374003970 Chr.9: 96937272-96937272 on Build GRCh38
rs976932695 Chr.9: 96936932-96936932 on Build GRCh38
rs571115100 Chr.9: 96936923-96936923 on Build GRCh38
rs550671322 Chr.9: 96937351-96937351 on Build GRCh38
rs531095716 Chr.9: 96936910-96936910 on Build GRCh38
rs368950642 Chr.9: 96937270-96937270 on Build GRCh38
rs572799896 Chr.9: 96937119-96937119 on Build GRCh38
rs774970338 Chr.9: 96937042-96937042 on Build GRCh38
rs763635180 Chr.9: 96937128-96937128 on Build GRCh38
rs767691060 Chr.9: 96937034-96937035 on Build GRCh38
rs770651527 Chr.9: 96937349-96937349 on Build GRCh38
rs750307487 Chr.9: 96937065-96937065 on Build GRCh38
rs762664118 Chr.9: 96937298-96937298 on Build GRCh38
rs895658180 Chr.9: 96936879-96936879 on Build GRCh38
rs747515880 Chr.9: 96937199-96937199 on Build GRCh38
rs150298455 Chr.9: 96936896-96936896 on Build GRCh38
rs781554004 Chr.9: 96937289-96937289 on Build GRCh38
rs749504282 Chr.9: 96937098-96937098 on Build GRCh38
rs761177466 Chr.9: 96937051-96937054 on Build GRCh38
rs373993403 Chr.9: 96937294-96937294 on Build GRCh38
rs767425758 Chr.9: 96937312-96937312 on Build GRCh38
rs752320981 Chr.9: 96937204-96937204 on Build GRCh38
rs1014042044 Chr.9: 96936891-96936891 on Build GRCh38
rs754178435 Chr.9: 96937122-96937123 on Build GRCh38
rs568487690 Chr.9: 96936943-96936943 on Build GRCh38
rs777419009 Chr.9: 96937342-96937342 on Build GRCh38
rs374965252 Chr.9: 96937172-96937172 on Build GRCh38
rs7866127 Chr.9: 96937203-96937203 on Build GRCh38
rs769502327 Chr.9: 96937211-96937211 on Build GRCh38
rs921535646 Chr.9: 96936937-96936937 on Build GRCh38
rs770775979 Chr.9: 96937300-96937300 on Build GRCh38
rs557147410 Chr.9: 96937000-96937000 on Build GRCh38
rs200040745 Chr.9: 96937320-96937320 on Build GRCh38
rs763590049 Chr.9: 96937052-96937052 on Build GRCh38
rs375671232 Chr.9: 96937336-96937336 on Build GRCh38
rs764853811 Chr.9: 96937143-96937143 on Build GRCh38
rs376949390 Chr.9: 96937274-96937274 on Build GRCh38
rs201094122 Chr.9: 96937295-96937295 on Build GRCh38
rs568714976 Chr.9: 96937015-96937015 on Build GRCh38
rs755328548 Chr.9: 96937330-96937330 on Build GRCh38
rs777810092 Chr.9: 96937097-96937097 on Build GRCh38
rs573525485 Chr.9: 96937152-96937152 on Build GRCh38
rs746292041 Chr.9: 96937115-96937115 on Build GRCh38
rs557673424 Chr.9: 96937096-96937096 on Build GRCh38
rs751883281 Chr.9: 96937326-96937326 on Build GRCh38
rs760598267 Chr.9: 96937353-96937353 on Build GRCh38
rs754921597 Chr.9: 96937159-96937159 on Build GRCh38
rs368543444 Chr.9: 96937350-96937350 on Build GRCh38
rs779124673 Chr.9: 96937173-96937173 on Build GRCh38
rs201931025 Chr.9: 96937169-96937169 on Build GRCh38
rs766379024 Chr.9: 96937153-96937153 on Build GRCh38
rs766808357 Chr.9: 96937107-96937151 on Build GRCh38
rs763187878 Chr.9: 96937024-96937024 on Build GRCh38
rs774629957 Chr.9: 96937026-96937026 on Build GRCh38
rs373488412 Chr.9: 96937167-96937167 on Build GRCh38
rs761836957 Chr.9: 96937315-96937315 on Build GRCh38
rs535927725 Chr.9: 96936970-96936970 on Build GRCh38
rs760142539 Chr.9: 96937123-96937123 on Build GRCh38
rs767414269 Chr.9: 96937250-96937250 on Build GRCh38
rs760195758 Chr.9: 96937046-96937046 on Build GRCh38
rs201577636 Chr.9: 96937264-96937264 on Build GRCh38
rs779071452 Chr.9: 96937268-96937268 on Build GRCh38
rs773230310 Chr.9: 96937050-96937052 on Build GRCh38
rs1043670678 Chr.9: 96936948-96936948 on Build GRCh38
rs771100266 Chr.9: 96937100-96937100 on Build GRCh38
rs759514287 Chr.9: 96937249-96937249 on Build GRCh38
rs2479294 Chr.9: 96936846-96936846 on Build GRCh38
rs753899435 Chr.9: 96937255-96937255 on Build GRCh38
rs144131056 Chr.9: 96936919-96936919 on Build GRCh38
rs548537323 Chr.9: 96936858-96936858 on Build GRCh38
rs761520760 Chr.9: 96937139-96937139 on Build GRCh38
rs775408774 Chr.9: 96937352-96937352 on Build GRCh38
rs761241199 Chr.9: 96937209-96937209 on Build GRCh38

View Full Product Details