Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 235550359-235550937 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNG4
Entrez Gene ID
2786
Gene Name
G protein subunit gamma 4
Gene Aliases
-
UniGene
Hs.159711
Gene Chromosome Location
Chr.1: 235547685-235650754 on Build GRCh38
Transcripts
NM_001098721.1 NM_001098722.1 NM_004485.3 XM_006711761.2 XM_011544167.2

SNP Detail

SNP ID Chromosome Location
rs142586600 Chr.1: 235550511-235550512 on Build GRCh38
rs968362765 Chr.1: 235550796-235550796 on Build GRCh38
rs575277433 Chr.1: 235550736-235550736 on Build GRCh38
rs947163036 Chr.1: 235550424-235550424 on Build GRCh38
rs898477776 Chr.1: 235550735-235550735 on Build GRCh38
rs779657478 Chr.1: 235550631-235550631 on Build GRCh38
rs573181958 Chr.1: 235550671-235550671 on Build GRCh38
rs968435912 Chr.1: 235550656-235550656 on Build GRCh38
rs749989086 Chr.1: 235550482-235550482 on Build GRCh38
rs995562511 Chr.1: 235550578-235550578 on Build GRCh38
rs991663382 Chr.1: 235550725-235550725 on Build GRCh38
rs977121522 Chr.1: 235550861-235550861 on Build GRCh38
rs951204406 Chr.1: 235550586-235550586 on Build GRCh38
rs1029049991 Chr.1: 235550627-235550627 on Build GRCh38
rs931342435 Chr.1: 235550741-235550741 on Build GRCh38
rs75173966 Chr.1: 235550479-235550479 on Build GRCh38
rs112699826 Chr.1: 235550776-235550776 on Build GRCh38
rs750627942 Chr.1: 235550589-235550589 on Build GRCh38
rs867470402 Chr.1: 235550465-235550465 on Build GRCh38
rs569118213 Chr.1: 235550519-235550519 on Build GRCh38
rs1055528127 Chr.1: 235550535-235550535 on Build GRCh38
rs569357257 Chr.1: 235550830-235550830 on Build GRCh38
rs35511716 Chr.1: 235550626-235550627 on Build GRCh38
rs909096834 Chr.1: 235550419-235550419 on Build GRCh38
rs889244752 Chr.1: 235550793-235550793 on Build GRCh38
rs758615235 Chr.1: 235550670-235550670 on Build GRCh38
rs1025708101 Chr.1: 235550585-235550585 on Build GRCh38
rs550764916 Chr.1: 235550795-235550795 on Build GRCh38
rs560895526 Chr.1: 235550753-235550753 on Build GRCh38
rs1029493456 Chr.1: 235550792-235550793 on Build GRCh38
rs368178273 Chr.1: 235550596-235550596 on Build GRCh38
rs144754099 Chr.1: 235550719-235550719 on Build GRCh38
rs975144064 Chr.1: 235550415-235550415 on Build GRCh38
rs1012660353 Chr.1: 235550526-235550526 on Build GRCh38
rs541702219 Chr.1: 235550813-235550817 on Build GRCh38
rs201734140 Chr.1: 235550510-235550511 on Build GRCh38
rs953341551 Chr.1: 235550639-235550639 on Build GRCh38
rs563404647 Chr.1: 235550728-235550728 on Build GRCh38
rs184337926 Chr.1: 235550680-235550680 on Build GRCh38
rs112649166 Chr.1: 235550594-235550594 on Build GRCh38
rs963995386 Chr.1: 235550614-235550614 on Build GRCh38
rs753284107 Chr.1: 235550635-235550635 on Build GRCh38
rs964040226 Chr.1: 235550396-235550396 on Build GRCh38
rs935479942 Chr.1: 235550682-235550682 on Build GRCh38
rs538746201 Chr.1: 235550398-235550398 on Build GRCh38
rs761582312 Chr.1: 235550416-235550416 on Build GRCh38
rs59100223 Chr.1: 235550742-235550742 on Build GRCh38
rs536264845 Chr.1: 235550564-235550564 on Build GRCh38
rs921789951 Chr.1: 235550386-235550386 on Build GRCh38
rs904015731 Chr.1: 235550774-235550774 on Build GRCh38
rs940362669 Chr.1: 235550472-235550472 on Build GRCh38
rs147909932 Chr.1: 235550906-235550906 on Build GRCh38
rs577414340 Chr.1: 235550709-235550709 on Build GRCh38
rs12756717 Chr.1: 235550862-235550862 on Build GRCh38
rs76474158 Chr.1: 235550513-235550513 on Build GRCh38
rs555414785 Chr.1: 235550779-235550779 on Build GRCh38
rs1016048344 Chr.1: 235550657-235550657 on Build GRCh38
rs534256349 Chr.1: 235550675-235550675 on Build GRCh38
rs1045076700 Chr.1: 235550532-235550532 on Build GRCh38
rs895324817 Chr.1: 235550540-235550540 on Build GRCh38
rs896770118 Chr.1: 235550523-235550523 on Build GRCh38
rs560160151 Chr.1: 235550807-235550810 on Build GRCh38
rs189232114 Chr.1: 235550766-235550766 on Build GRCh38
rs1006708421 Chr.1: 235550804-235550804 on Build GRCh38
rs779984183 Chr.1: 235550705-235550705 on Build GRCh38
rs948219508 Chr.1: 235550548-235550548 on Build GRCh38
rs192754651 Chr.1: 235550630-235550630 on Build GRCh38
rs997492866 Chr.1: 235550786-235550786 on Build GRCh38
rs182223490 Chr.1: 235550780-235550780 on Build GRCh38
rs866302543 Chr.1: 235550775-235550775 on Build GRCh38
rs60214059 Chr.1: 235550512-235550513 on Build GRCh38
rs767070874 Chr.1: 235550427-235550427 on Build GRCh38

View Full Product Details