Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 42079007-42079600 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM109B
Entrez Gene ID
150368
Gene Name
family with sequence similarity 109 member B
Gene Aliases
IPIP27B, Ses2
UniGene
Hs.368312
Gene Chromosome Location
Chr.22: 42074244-42079438 on Build GRCh38
Transcripts
NM_001002034.2 XM_005261373.3

SNP Detail

SNP ID Chromosome Location
rs779877007 Chr.22: 42079191-42079191 on Build GRCh38
rs529384944 Chr.22: 42079550-42079550 on Build GRCh38
rs746918768 Chr.22: 42079216-42079216 on Build GRCh38
rs187166093 Chr.22: 42079525-42079525 on Build GRCh38
rs927657771 Chr.22: 42079553-42079553 on Build GRCh38
rs997594800 Chr.22: 42079407-42079407 on Build GRCh38
rs966044679 Chr.22: 42079240-42079240 on Build GRCh38
rs888410809 Chr.22: 42079181-42079181 on Build GRCh38
rs8135801 Chr.22: 42079564-42079564 on Build GRCh38
rs901423887 Chr.22: 42079432-42079432 on Build GRCh38
rs1048322519 Chr.22: 42079578-42079578 on Build GRCh38
rs571875684 Chr.22: 42079472-42079472 on Build GRCh38
rs1026932832 Chr.22: 42079170-42079170 on Build GRCh38
rs1017524872 Chr.22: 42079369-42079369 on Build GRCh38
rs1054963046 Chr.22: 42079313-42079313 on Build GRCh38
rs894870777 Chr.22: 42079104-42079104 on Build GRCh38
rs977734408 Chr.22: 42079522-42079522 on Build GRCh38
rs117020658 Chr.22: 42079356-42079356 on Build GRCh38
rs142757792 Chr.22: 42079426-42079426 on Build GRCh38
rs1055403039 Chr.22: 42079155-42079155 on Build GRCh38
rs148509374 Chr.22: 42079105-42079105 on Build GRCh38
rs993803207 Chr.22: 42079137-42079137 on Build GRCh38
rs964725558 Chr.22: 42079174-42079174 on Build GRCh38
rs528685649 Chr.22: 42079433-42079433 on Build GRCh38
rs377218463 Chr.22: 42079275-42079275 on Build GRCh38
rs562469112 Chr.22: 42079277-42079277 on Build GRCh38
rs77848693 Chr.22: 42079098-42079098 on Build GRCh38
rs60412323 Chr.22: 42079453-42079453 on Build GRCh38
rs927851422 Chr.22: 42079302-42079302 on Build GRCh38
rs536278165 Chr.22: 42079562-42079562 on Build GRCh38
rs1032281260 Chr.22: 42079481-42079481 on Build GRCh38
rs914938823 Chr.22: 42079129-42079129 on Build GRCh38
rs768611234 Chr.22: 42079266-42079266 on Build GRCh38
rs966342779 Chr.22: 42079287-42079287 on Build GRCh38
rs35768281 Chr.22: 42079285-42079286 on Build GRCh38
rs994057473 Chr.22: 42079299-42079299 on Build GRCh38
rs916501606 Chr.22: 42079566-42079566 on Build GRCh38
rs759567762 Chr.22: 42079498-42079498 on Build GRCh38
rs11551670 Chr.22: 42079256-42079256 on Build GRCh38
rs977765561 Chr.22: 42079297-42079297 on Build GRCh38
rs930499836 Chr.22: 42079352-42079352 on Build GRCh38
rs1047986492 Chr.22: 42079355-42079356 on Build GRCh38
rs1004158160 Chr.22: 42079400-42079400 on Build GRCh38
rs916432420 Chr.22: 42079314-42079314 on Build GRCh38
rs939165731 Chr.22: 42079154-42079154 on Build GRCh38
rs576679178 Chr.22: 42079152-42079152 on Build GRCh38
rs541884296 Chr.22: 42079371-42079371 on Build GRCh38
rs1013455463 Chr.22: 42079168-42079168 on Build GRCh38
rs1026962929 Chr.22: 42079150-42079152 on Build GRCh38
rs953813546 Chr.22: 42079268-42079268 on Build GRCh38
rs894540044 Chr.22: 42079167-42079167 on Build GRCh38
rs374484357 Chr.22: 42079504-42079504 on Build GRCh38
rs966945009 Chr.22: 42079219-42079219 on Build GRCh38
rs757995449 Chr.22: 42079430-42079430 on Build GRCh38
rs1006179803 Chr.22: 42079365-42079365 on Build GRCh38
rs976930768 Chr.22: 42079247-42079247 on Build GRCh38
rs939288068 Chr.22: 42079305-42079305 on Build GRCh38
rs80060478 Chr.22: 42079153-42079153 on Build GRCh38
rs532815309 Chr.22: 42079491-42079491 on Build GRCh38
rs925012604 Chr.22: 42079084-42079084 on Build GRCh38
rs1037018508 Chr.22: 42079405-42079405 on Build GRCh38
rs1028582442 Chr.22: 42079463-42079463 on Build GRCh38
rs960646661 Chr.22: 42079555-42079555 on Build GRCh38
rs975895359 Chr.22: 42079187-42079192 on Build GRCh38
rs1054910492 Chr.22: 42079099-42079099 on Build GRCh38
rs1028949682 Chr.22: 42079252-42079252 on Build GRCh38
rs1011097525 Chr.22: 42079487-42079487 on Build GRCh38
rs544474048 Chr.22: 42079385-42079385 on Build GRCh38
rs945036812 Chr.22: 42079141-42079141 on Build GRCh38
rs753641022 Chr.22: 42079108-42079108 on Build GRCh38
rs916339066 Chr.22: 42079286-42079286 on Build GRCh38
rs749580864 Chr.22: 42079372-42079372 on Build GRCh38
rs1015771681 Chr.22: 42079237-42079237 on Build GRCh38
rs916285319 Chr.22: 42079549-42079549 on Build GRCh38
rs763166255 Chr.22: 42079163-42079164 on Build GRCh38
rs35404966 Chr.22: 42079471-42079472 on Build GRCh38
rs989195728 Chr.22: 42079270-42079270 on Build GRCh38
rs978058616 Chr.22: 42079241-42079241 on Build GRCh38
rs540072668 Chr.22: 42079122-42079122 on Build GRCh38
rs923162806 Chr.22: 42079207-42079207 on Build GRCh38
rs1022024384 Chr.22: 42079499-42079499 on Build GRCh38
rs888037302 Chr.22: 42079581-42079581 on Build GRCh38
rs978300003 Chr.22: 42079454-42079454 on Build GRCh38
rs558421268 Chr.22: 42079151-42079151 on Build GRCh38

View Full Product Details