Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96536365-96536835 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARID5A
Entrez Gene ID
10865
Gene Name
AT-rich interaction domain 5A
Gene Aliases
MRF-1, MRF1, RP11-363D14
UniGene
Hs.920
Gene Chromosome Location
Chr.2: 96536719-96552638 on Build GRCh38
Transcripts
NM_001319085.1 NM_001319087.1 NM_001319093.1 NM_001319094.1 NM_001319096.1 NM_212481.2 XM_017003184.1

SNP Detail

SNP ID Chromosome Location
rs1001863213 Chr.2: 96536678-96536678 on Build GRCh38
rs949851100 Chr.2: 96536752-96536752 on Build GRCh38
rs561129356 Chr.2: 96536792-96536792 on Build GRCh38
rs1007856401 Chr.2: 96536625-96536625 on Build GRCh38
rs945528473 Chr.2: 96536807-96536807 on Build GRCh38
rs552601264 Chr.2: 96536511-96536511 on Build GRCh38
rs914027358 Chr.2: 96536806-96536806 on Build GRCh38
rs966197726 Chr.2: 96536605-96536605 on Build GRCh38
rs936726440 Chr.2: 96536619-96536619 on Build GRCh38
rs976257500 Chr.2: 96536608-96536608 on Build GRCh38
rs945582423 Chr.2: 96536658-96536658 on Build GRCh38
rs974704065 Chr.2: 96536778-96536778 on Build GRCh38
rs966881368 Chr.2: 96536721-96536721 on Build GRCh38
rs898835434 Chr.2: 96536688-96536688 on Build GRCh38
rs1033415454 Chr.2: 96536406-96536406 on Build GRCh38
rs1026498727 Chr.2: 96536808-96536808 on Build GRCh38
rs936691782 Chr.2: 96536783-96536783 on Build GRCh38
rs892598381 Chr.2: 96536633-96536633 on Build GRCh38
rs906816816 Chr.2: 96536676-96536676 on Build GRCh38
rs918037676 Chr.2: 96536748-96536748 on Build GRCh38
rs71386015 Chr.2: 96536723-96536723 on Build GRCh38
rs987327883 Chr.2: 96536729-96536729 on Build GRCh38
rs973195960 Chr.2: 96536422-96536422 on Build GRCh38
rs944035993 Chr.2: 96536549-96536549 on Build GRCh38
rs1007676583 Chr.2: 96536786-96536786 on Build GRCh38
rs374892963 Chr.2: 96536743-96536743 on Build GRCh38
rs987380151 Chr.2: 96536512-96536512 on Build GRCh38
rs921286121 Chr.2: 96536614-96536614 on Build GRCh38
rs911853636 Chr.2: 96536513-96536513 on Build GRCh38
rs558154332 Chr.2: 96536700-96536700 on Build GRCh38
rs964958739 Chr.2: 96536772-96536772 on Build GRCh38
rs989514741 Chr.2: 96536801-96536801 on Build GRCh38
rs975850160 Chr.2: 96536578-96536578 on Build GRCh38
rs55768571 Chr.2: 96536753-96536753 on Build GRCh38
rs950531424 Chr.2: 96536725-96536725 on Build GRCh38
rs1026529901 Chr.2: 96536652-96536652 on Build GRCh38
rs1018862108 Chr.2: 96536734-96536734 on Build GRCh38
rs934608036 Chr.2: 96536776-96536776 on Build GRCh38
rs577923454 Chr.2: 96536781-96536781 on Build GRCh38
rs1017388308 Chr.2: 96536586-96536586 on Build GRCh38
rs1033741475 Chr.2: 96536679-96536679 on Build GRCh38
rs2708978 Chr.2: 96536506-96536506 on Build GRCh38
rs1046576050 Chr.2: 96536812-96536812 on Build GRCh38
rs1046628621 Chr.2: 96536659-96536659 on Build GRCh38
rs568599510 Chr.2: 96536403-96536403 on Build GRCh38
rs979175164 Chr.2: 96536724-96536724 on Build GRCh38
rs13018837 Chr.2: 96536573-96536573 on Build GRCh38
rs538216209 Chr.2: 96536626-96536626 on Build GRCh38
rs903115613 Chr.2: 96536560-96536560 on Build GRCh38
rs924497276 Chr.2: 96536762-96536762 on Build GRCh38
rs998885509 Chr.2: 96536572-96536572 on Build GRCh38
rs538643640 Chr.2: 96536414-96536414 on Build GRCh38
rs572615786 Chr.2: 96536551-96536551 on Build GRCh38
rs13013773 Chr.2: 96536393-96536393 on Build GRCh38
rs892783884 Chr.2: 96536780-96536780 on Build GRCh38
rs1028195858 Chr.2: 96536606-96536606 on Build GRCh38
rs890827327 Chr.2: 96536618-96536618 on Build GRCh38
rs13020129 Chr.2: 96536530-96536530 on Build GRCh38
rs1026103609 Chr.2: 96536711-96536711 on Build GRCh38
rs548745968 Chr.2: 96536397-96536397 on Build GRCh38
rs1055168620 Chr.2: 96536443-96536443 on Build GRCh38
rs894295744 Chr.2: 96536474-96536475 on Build GRCh38
rs892847970 Chr.2: 96536581-96536581 on Build GRCh38
rs13020073 Chr.2: 96536461-96536461 on Build GRCh38
rs1007297140 Chr.2: 96536585-96536585 on Build GRCh38
rs1020580637 Chr.2: 96536630-96536630 on Build GRCh38
rs951015346 Chr.2: 96536656-96536656 on Build GRCh38
rs959663292 Chr.2: 96536732-96536732 on Build GRCh38

View Full Product Details