Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 71288748-71289300 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMTR2
Entrez Gene ID
55783
Gene Name
cap methyltransferase 2
Gene Aliases
AFT, FTSJD1, HMTr2, MTr2
UniGene
Hs.72782
Gene Chromosome Location
Chr.16: 71282300-71290178 on Build GRCh38
Transcripts
NM_001099642.1 NM_001324374.1 NM_001324377.1 NM_001324378.1 NM_001324379.1 NM_018348.5 XM_011523228.1 XM_011523229.1 XM_017023444.1

SNP Detail

SNP ID Chromosome Location
rs34981087 Chr.16: 71288875-71288875 on Build GRCh38
rs771947494 Chr.16: 71288885-71288885 on Build GRCh38
rs954973849 Chr.16: 71288929-71288929 on Build GRCh38
rs900581571 Chr.16: 71289102-71289102 on Build GRCh38
rs922902115 Chr.16: 71288947-71288947 on Build GRCh38
rs1016535058 Chr.16: 71289095-71289095 on Build GRCh38
rs549026128 Chr.16: 71288871-71288871 on Build GRCh38
rs923304236 Chr.16: 71288886-71288886 on Build GRCh38
rs1016209359 Chr.16: 71289086-71289086 on Build GRCh38
rs564264951 Chr.16: 71288872-71288872 on Build GRCh38
rs1047656436 Chr.16: 71289046-71289046 on Build GRCh38
rs761950152 Chr.16: 71289138-71289138 on Build GRCh38
rs538044421 Chr.16: 71289130-71289130 on Build GRCh38
rs1006077260 Chr.16: 71288859-71288859 on Build GRCh38
rs553751184 Chr.16: 71289021-71289021 on Build GRCh38
rs770545946 Chr.16: 71288782-71288782 on Build GRCh38
rs556470777 Chr.16: 71289259-71289259 on Build GRCh38
rs369759143 Chr.16: 71288955-71288955 on Build GRCh38
rs765415339 Chr.16: 71289178-71289178 on Build GRCh38
rs546815149 Chr.16: 71289169-71289169 on Build GRCh38
rs906552879 Chr.16: 71289223-71289223 on Build GRCh38
rs958600740 Chr.16: 71289047-71289047 on Build GRCh38
rs909115189 Chr.16: 71288891-71288891 on Build GRCh38
rs936876264 Chr.16: 71289126-71289126 on Build GRCh38
rs940378531 Chr.16: 71288905-71288905 on Build GRCh38
rs928423226 Chr.16: 71289194-71289194 on Build GRCh38
rs373982828 Chr.16: 71289176-71289176 on Build GRCh38
rs1002825304 Chr.16: 71288776-71288776 on Build GRCh38
rs1047457939 Chr.16: 71289260-71289260 on Build GRCh38
rs916770675 Chr.16: 71289147-71289147 on Build GRCh38
rs989663899 Chr.16: 71289146-71289146 on Build GRCh38
rs548577002 Chr.16: 71289064-71289064 on Build GRCh38
rs892500909 Chr.16: 71288775-71288775 on Build GRCh38
rs866735293 Chr.16: 71289151-71289151 on Build GRCh38
rs373253484 Chr.16: 71289005-71289005 on Build GRCh38
rs970774384 Chr.16: 71288828-71288828 on Build GRCh38
rs1002153025 Chr.16: 71289227-71289227 on Build GRCh38
rs922934593 Chr.16: 71289105-71289105 on Build GRCh38
rs1046594725 Chr.16: 71289215-71289215 on Build GRCh38
rs566870649 Chr.16: 71289077-71289077 on Build GRCh38
rs760623893 Chr.16: 71289042-71289042 on Build GRCh38
rs886336726 Chr.16: 71289263-71289263 on Build GRCh38
rs892691147 Chr.16: 71288988-71288988 on Build GRCh38
rs145637162 Chr.16: 71288825-71288825 on Build GRCh38
rs182827014 Chr.16: 71288954-71288954 on Build GRCh38
rs1030421831 Chr.16: 71289117-71289117 on Build GRCh38
rs374962553 Chr.16: 71288916-71288916 on Build GRCh38
rs376685489 Chr.16: 71289111-71289111 on Build GRCh38
rs948254785 Chr.16: 71289171-71289171 on Build GRCh38
rs971710314 Chr.16: 71288906-71288906 on Build GRCh38
rs1006044412 Chr.16: 71289073-71289073 on Build GRCh38
rs906828762 Chr.16: 71288989-71288989 on Build GRCh38
rs759302044 Chr.16: 71288860-71288860 on Build GRCh38
rs989638396 Chr.16: 71288967-71288967 on Build GRCh38
rs531403844 Chr.16: 71288881-71288881 on Build GRCh38
rs552500935 Chr.16: 71289120-71289120 on Build GRCh38
rs1009554014 Chr.16: 71288996-71288996 on Build GRCh38
rs762984424 Chr.16: 71289224-71289224 on Build GRCh38
rs1009519545 Chr.16: 71288803-71288803 on Build GRCh38
rs764282996 Chr.16: 71289059-71289059 on Build GRCh38
rs950492819 Chr.16: 71288853-71288853 on Build GRCh38
rs945652615 Chr.16: 71289192-71289192 on Build GRCh38
rs938331352 Chr.16: 71288990-71288990 on Build GRCh38
rs561948235 Chr.16: 71288980-71288980 on Build GRCh38
rs775604099 Chr.16: 71288952-71288952 on Build GRCh38
rs978080157 Chr.16: 71289122-71289122 on Build GRCh38
rs2303257 Chr.16: 71289124-71289124 on Build GRCh38
rs914045356 Chr.16: 71289179-71289179 on Build GRCh38
rs954324983 Chr.16: 71289110-71289110 on Build GRCh38
rs936816826 Chr.16: 71288956-71288956 on Build GRCh38
rs1044062073 Chr.16: 71288985-71288985 on Build GRCh38
rs948517269 Chr.16: 71288981-71288981 on Build GRCh38
rs906523397 Chr.16: 71289044-71289044 on Build GRCh38

View Full Product Details