Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 73207854-73208412 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COG1
Entrez Gene ID
9382
Gene Name
component of oligomeric golgi complex 1
Gene Aliases
CDG2G, LDLB
UniGene
Hs.103555
Gene Chromosome Location
Chr.17: 73193034-73208507 on Build GRCh38
Transcripts
NM_018714.2 NM_001098832.1 NM_001289410.1 NM_001289411.1 NM_001289412.1 NM_032837.2

Gene Symbol
FAM104A
Entrez Gene ID
84923
Gene Name
family with sequence similarity 104 member A
Gene Aliases
-
UniGene
Hs.741276
Gene Chromosome Location
Chr.17: 73207353-73232394 on Build GRCh38
Transcripts
NM_018714.2 NM_001098832.1 NM_001289410.1 NM_001289411.1 NM_001289412.1 NM_032837.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4972773 c.2861_2874del14 p.F954fs*1 Chr.17: 73208369-73208382 on Build GRCh38
5853368 c.2810_2811insC p.A940fs*7 Chr.17: 73208318-73208319 on Build GRCh38
122257 c.2816C>T p.P939L Chr.17: 73208324-73208324 on Build GRCh38
3820434 c.2837A>G p.D946G Chr.17: 73208345-73208345 on Build GRCh38
3691764 c.2841G>A p.P947P Chr.17: 73208349-73208349 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs747195916 Chr.17: 73208302-73208302 on Build GRCh38
rs145071676 Chr.17: 73208351-73208351 on Build GRCh38
rs939990905 Chr.17: 73208245-73208245 on Build GRCh38
rs759050295 Chr.17: 73208330-73208330 on Build GRCh38
rs967516662 Chr.17: 73207963-73207963 on Build GRCh38
rs373538467 Chr.17: 73208379-73208379 on Build GRCh38
rs975743817 Chr.17: 73208070-73208070 on Build GRCh38
rs940147997 Chr.17: 73208139-73208139 on Build GRCh38
rs189587565 Chr.17: 73208248-73208248 on Build GRCh38
rs768934064 Chr.17: 73208362-73208362 on Build GRCh38
rs116132000 Chr.17: 73208211-73208211 on Build GRCh38
rs755921036 Chr.17: 73208348-73208348 on Build GRCh38
rs897763480 Chr.17: 73208128-73208128 on Build GRCh38
rs771628866 Chr.17: 73208255-73208255 on Build GRCh38
rs574563160 Chr.17: 73208244-73208244 on Build GRCh38
rs776834154 Chr.17: 73208318-73208319 on Build GRCh38
rs549811635 Chr.17: 73208102-73208102 on Build GRCh38
rs572268033 Chr.17: 73208319-73208319 on Build GRCh38
rs907476725 Chr.17: 73208194-73208194 on Build GRCh38
rs181641412 Chr.17: 73208249-73208249 on Build GRCh38
rs772019650 Chr.17: 73208265-73208265 on Build GRCh38
rs560960051 Chr.17: 73207947-73207947 on Build GRCh38
rs547662558 Chr.17: 73208155-73208155 on Build GRCh38
rs376824047 Chr.17: 73208352-73208352 on Build GRCh38
rs768371015 Chr.17: 73208233-73208233 on Build GRCh38
rs374616145 Chr.17: 73207935-73207935 on Build GRCh38
rs898669897 Chr.17: 73208166-73208166 on Build GRCh38
rs976076424 Chr.17: 73208242-73208242 on Build GRCh38
rs763355798 Chr.17: 73208386-73208386 on Build GRCh38
rs1022010610 Chr.17: 73208217-73208217 on Build GRCh38
rs779085570 Chr.17: 73208068-73208068 on Build GRCh38
rs910514238 Chr.17: 73208091-73208092 on Build GRCh38
rs139440017 Chr.17: 73208344-73208344 on Build GRCh38
rs149643307 Chr.17: 73208134-73208134 on Build GRCh38
rs147965233 Chr.17: 73208339-73208339 on Build GRCh38
rs752683727 Chr.17: 73208256-73208256 on Build GRCh38
rs78674872 Chr.17: 73207946-73207946 on Build GRCh38
rs755258012 Chr.17: 73208060-73208060 on Build GRCh38
rs766614159 Chr.17: 73208370-73208370 on Build GRCh38
rs1009321107 Chr.17: 73208216-73208216 on Build GRCh38
rs936133127 Chr.17: 73208031-73208031 on Build GRCh38
rs761967468 Chr.17: 73208389-73208390 on Build GRCh38
rs554412385 Chr.17: 73208176-73208176 on Build GRCh38
rs1001923742 Chr.17: 73208145-73208145 on Build GRCh38
rs975248419 Chr.17: 73208224-73208224 on Build GRCh38
rs141573449 Chr.17: 73208301-73208301 on Build GRCh38
rs747128241 Chr.17: 73208360-73208360 on Build GRCh38
rs763565694 Chr.17: 73208324-73208324 on Build GRCh38
rs956469994 Chr.17: 73207919-73207919 on Build GRCh38
rs560309960 Chr.17: 73207908-73207908 on Build GRCh38
rs773693157 Chr.17: 73208380-73208380 on Build GRCh38
rs148796681 Chr.17: 73208247-73208247 on Build GRCh38
rs1003111466 Chr.17: 73208205-73208205 on Build GRCh38
rs748475815 Chr.17: 73208374-73208374 on Build GRCh38
rs777590708 Chr.17: 73208349-73208349 on Build GRCh38
rs879096929 Chr.17: 73207935-73207935 on Build GRCh38
rs764085125 Chr.17: 73208294-73208298 on Build GRCh38
rs986678621 Chr.17: 73208235-73208235 on Build GRCh38
rs903624939 Chr.17: 73208350-73208350 on Build GRCh38
rs779987364 Chr.17: 73208356-73208356 on Build GRCh38
rs1034668168 Chr.17: 73207917-73207917 on Build GRCh38
rs751136946 Chr.17: 73208329-73208329 on Build GRCh38
rs757194205 Chr.17: 73208132-73208132 on Build GRCh38
rs896249142 Chr.17: 73208320-73208320 on Build GRCh38
rs779937596 Chr.17: 73208210-73208210 on Build GRCh38
rs370117464 Chr.17: 73208065-73208065 on Build GRCh38
rs76058650 Chr.17: 73208144-73208144 on Build GRCh38
rs112178973 Chr.17: 73208167-73208167 on Build GRCh38
rs761285886 Chr.17: 73208307-73208307 on Build GRCh38
rs374570467 Chr.17: 73208001-73208001 on Build GRCh38
rs79914967 Chr.17: 73207914-73207914 on Build GRCh38
rs934010560 Chr.17: 73208101-73208101 on Build GRCh38
rs777081216 Chr.17: 73208315-73208315 on Build GRCh38
rs184781213 Chr.17: 73208201-73208201 on Build GRCh38
rs977610394 Chr.17: 73207993-73207993 on Build GRCh38
rs752836568 Chr.17: 73208097-73208097 on Build GRCh38
rs144896007 Chr.17: 73208326-73208326 on Build GRCh38
rs748845630 Chr.17: 73208223-73208223 on Build GRCh38
rs144366335 Chr.17: 73208234-73208234 on Build GRCh38
rs907361057 Chr.17: 73208286-73208286 on Build GRCh38
rs767159115 Chr.17: 73208334-73208334 on Build GRCh38

View Full Product Details