Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26382837-26383404 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTN2A2
Entrez Gene ID
10385
Gene Name
butyrophilin subfamily 2 member A2
Gene Aliases
BT2.2, BTF2, BTN2.2
UniGene
Hs.373938
Gene Chromosome Location
Chr.6: 26382891-26394874 on Build GRCh38
Transcripts
NM_001197237.1 NM_001197238.1 NM_001197239.1 NM_001197240.1 NM_006995.4 NM_181531.2 XM_005248797.3 XM_005248798.4 XM_006714953.3 XM_006714955.3 XM_011514228.2 XM_011514229.2 XM_011514231.2 XM_017010166.1 XM_017010167.1 XM_017010168.1 XM_017010169.1 XM_011514268.2

Gene Symbol
BTN3A2
Entrez Gene ID
11118
Gene Name
butyrophilin subfamily 3 member A2
Gene Aliases
BT3.2, BTF4, BTN3.2, CD277
UniGene
Hs.376046
Gene Chromosome Location
Chr.6: 26365159-26451697 on Build GRCh38
Transcripts
NM_001197237.1 NM_001197238.1 NM_001197239.1 NM_001197240.1 NM_006995.4 NM_181531.2 XM_005248797.3 XM_005248798.4 XM_006714953.3 XM_006714955.3 XM_011514228.2 XM_011514229.2 XM_011514231.2 XM_017010166.1 XM_017010167.1 XM_017010168.1 XM_017010169.1 XM_011514268.2

SNP Detail

SNP ID Chromosome Location
rs760818181 Chr.6: 26382878-26382878 on Build GRCh38
rs1023081117 Chr.6: 26382898-26382898 on Build GRCh38
rs749849041 Chr.6: 26382867-26382878 on Build GRCh38
rs745510066 Chr.6: 26382948-26382948 on Build GRCh38
rs9461249 Chr.6: 26383158-26383158 on Build GRCh38
rs944554955 Chr.6: 26383049-26383049 on Build GRCh38
rs1004587496 Chr.6: 26383123-26383123 on Build GRCh38
rs72842959 Chr.6: 26382955-26382955 on Build GRCh38
rs548899983 Chr.6: 26383279-26383279 on Build GRCh38
rs934256007 Chr.6: 26383259-26383259 on Build GRCh38
rs1027387389 Chr.6: 26382888-26382888 on Build GRCh38
rs397840731 Chr.6: 26383298-26383299 on Build GRCh38
rs115878961 Chr.6: 26382961-26382961 on Build GRCh38
rs9467740 Chr.6: 26383022-26383022 on Build GRCh38
rs568432182 Chr.6: 26383337-26383337 on Build GRCh38
rs887447503 Chr.6: 26383122-26383122 on Build GRCh38
rs1049901968 Chr.6: 26382972-26382972 on Build GRCh38
rs929655755 Chr.6: 26383367-26383367 on Build GRCh38
rs565125465 Chr.6: 26382939-26382939 on Build GRCh38
rs766776295 Chr.6: 26383357-26383357 on Build GRCh38
rs958717245 Chr.6: 26382960-26382960 on Build GRCh38
rs529980281 Chr.6: 26383000-26383000 on Build GRCh38
rs1038084960 Chr.6: 26382997-26382997 on Build GRCh38
rs116499448 Chr.6: 26383059-26383059 on Build GRCh38
rs996125841 Chr.6: 26383113-26383113 on Build GRCh38
rs9467741 Chr.6: 26383231-26383231 on Build GRCh38
rs572892735 Chr.6: 26383057-26383057 on Build GRCh38
rs756739441 Chr.6: 26382880-26382880 on Build GRCh38
rs780211507 Chr.6: 26383208-26383208 on Build GRCh38
rs979292460 Chr.6: 26383131-26383131 on Build GRCh38
rs745903100 Chr.6: 26383167-26383167 on Build GRCh38
rs761774137 Chr.6: 26383126-26383126 on Build GRCh38
rs1034965046 Chr.6: 26383013-26383013 on Build GRCh38
rs912679820 Chr.6: 26383368-26383368 on Build GRCh38
rs1049176476 Chr.6: 26383118-26383118 on Build GRCh38
rs1023488358 Chr.6: 26383105-26383105 on Build GRCh38
rs374126658 Chr.6: 26382998-26382998 on Build GRCh38
rs778446975 Chr.6: 26382916-26382916 on Build GRCh38
rs993842476 Chr.6: 26383011-26383011 on Build GRCh38
rs567798593 Chr.6: 26382947-26382947 on Build GRCh38
rs923765765 Chr.6: 26383044-26383044 on Build GRCh38
rs764601437 Chr.6: 26382974-26382974 on Build GRCh38
rs1003021632 Chr.6: 26383156-26383156 on Build GRCh38
rs967875371 Chr.6: 26383112-26383112 on Build GRCh38
rs1040730906 Chr.6: 26382874-26382874 on Build GRCh38
rs879906625 Chr.6: 26382987-26382988 on Build GRCh38
rs1004597355 Chr.6: 26382905-26382905 on Build GRCh38
rs960564553 Chr.6: 26382867-26382867 on Build GRCh38
rs1015007498 Chr.6: 26382932-26382932 on Build GRCh38
rs374326304 Chr.6: 26382893-26382893 on Build GRCh38
rs971527467 Chr.6: 26383147-26383147 on Build GRCh38
rs934673613 Chr.6: 26382945-26382945 on Build GRCh38
rs546642191 Chr.6: 26383006-26383006 on Build GRCh38
rs970150335 Chr.6: 26382904-26382904 on Build GRCh38
rs561008862 Chr.6: 26382994-26382994 on Build GRCh38
rs1014660931 Chr.6: 26383146-26383146 on Build GRCh38
rs914594232 Chr.6: 26382978-26382978 on Build GRCh38
rs989500933 Chr.6: 26382946-26382946 on Build GRCh38
rs372327483 Chr.6: 26383350-26383350 on Build GRCh38
rs911392948 Chr.6: 26383333-26383333 on Build GRCh38
rs551977089 Chr.6: 26383129-26383129 on Build GRCh38
rs988752003 Chr.6: 26383204-26383204 on Build GRCh38
rs545850453 Chr.6: 26382907-26382907 on Build GRCh38
rs946078610 Chr.6: 26382993-26382993 on Build GRCh38
rs3832422 Chr.6: 26383296-26383297 on Build GRCh38
rs756210842 Chr.6: 26383207-26383207 on Build GRCh38
rs977494350 Chr.6: 26383358-26383358 on Build GRCh38
rs575165872 Chr.6: 26382884-26382884 on Build GRCh38
rs375893732 Chr.6: 26383295-26383295 on Build GRCh38

View Full Product Details