Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 127589338-127589920 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARF5
Entrez Gene ID
381
Gene Name
ADP ribosylation factor 5
Gene Aliases
-
UniGene
Hs.653202
Gene Chromosome Location
Chr.7: 127588352-127591705 on Build GRCh38
Transcripts
NM_001662.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
484769 c.257A>G p.Q86R Chr.7: 127589593-127589593 on Build GRCh38
7096747 c.249G>A p.Q83Q Chr.7: 127589585-127589585 on Build GRCh38
5997268 c.202G>A p.V68M Chr.7: 127589538-127589538 on Build GRCh38
5729973 c.208G>A p.G70S Chr.7: 127589544-127589544 on Build GRCh38
3632950 c.204G>T p.V68V Chr.7: 127589540-127589540 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1039475305 Chr.7: 127589517-127589517 on Build GRCh38
rs370775243 Chr.7: 127589623-127589623 on Build GRCh38
rs900970205 Chr.7: 127589383-127589383 on Build GRCh38
rs968825966 Chr.7: 127589736-127589736 on Build GRCh38
rs987756715 Chr.7: 127589845-127589845 on Build GRCh38
rs1005245803 Chr.7: 127589866-127589866 on Build GRCh38
rs779055906 Chr.7: 127589629-127589629 on Build GRCh38
rs774867595 Chr.7: 127589462-127589463 on Build GRCh38
rs764230627 Chr.7: 127589667-127589667 on Build GRCh38
rs771978864 Chr.7: 127589879-127589879 on Build GRCh38
rs991596740 Chr.7: 127589407-127589407 on Build GRCh38
rs772209207 Chr.7: 127589638-127589638 on Build GRCh38
rs990617455 Chr.7: 127589397-127589430 on Build GRCh38
rs200093249 Chr.7: 127589479-127589479 on Build GRCh38
rs370698583 Chr.7: 127589641-127589641 on Build GRCh38
rs545388471 Chr.7: 127589673-127589673 on Build GRCh38
rs200732482 Chr.7: 127589438-127589438 on Build GRCh38
rs779899044 Chr.7: 127589627-127589627 on Build GRCh38
rs773751209 Chr.7: 127589463-127589463 on Build GRCh38
rs138066010 Chr.7: 127589555-127589555 on Build GRCh38
rs996555749 Chr.7: 127589428-127589428 on Build GRCh38
rs755911930 Chr.7: 127589614-127589614 on Build GRCh38
rs1004133152 Chr.7: 127589648-127589648 on Build GRCh38
rs549079833 Chr.7: 127589440-127589440 on Build GRCh38
rs774948946 Chr.7: 127589482-127589482 on Build GRCh38
rs190575120 Chr.7: 127589459-127589459 on Build GRCh38
rs762671314 Chr.7: 127589484-127589484 on Build GRCh38
rs771134679 Chr.7: 127589653-127589653 on Build GRCh38
rs777075800 Chr.7: 127589656-127589656 on Build GRCh38
rs943675844 Chr.7: 127589489-127589489 on Build GRCh38
rs369533412 Chr.7: 127589601-127589601 on Build GRCh38
rs749965373 Chr.7: 127589400-127589400 on Build GRCh38
rs761391070 Chr.7: 127589665-127589665 on Build GRCh38
rs568951415 Chr.7: 127589470-127589470 on Build GRCh38
rs981999965 Chr.7: 127589558-127589558 on Build GRCh38
rs78893975 Chr.7: 127589794-127589794 on Build GRCh38
rs182359215 Chr.7: 127589707-127589707 on Build GRCh38
rs779226533 Chr.7: 127589710-127589710 on Build GRCh38
rs562012325 Chr.7: 127589408-127589408 on Build GRCh38
rs191752924 Chr.7: 127589841-127589841 on Build GRCh38
rs186921350 Chr.7: 127589831-127589831 on Build GRCh38
rs117589386 Chr.7: 127589644-127589644 on Build GRCh38
rs535792660 Chr.7: 127589811-127589811 on Build GRCh38
rs748197231 Chr.7: 127589635-127589635 on Build GRCh38
rs774211289 Chr.7: 127589664-127589664 on Build GRCh38
rs112290402 Chr.7: 127589628-127589628 on Build GRCh38
rs553236614 Chr.7: 127589791-127589791 on Build GRCh38
rs756865852 Chr.7: 127589546-127589546 on Build GRCh38
rs979404198 Chr.7: 127589677-127589677 on Build GRCh38
rs1013308705 Chr.7: 127589681-127589681 on Build GRCh38
rs763463998 Chr.7: 127589519-127589519 on Build GRCh38
rs780471979 Chr.7: 127589420-127589420 on Build GRCh38
rs888096250 Chr.7: 127589839-127589839 on Build GRCh38
rs566872844 Chr.7: 127589698-127589698 on Build GRCh38
rs140755629 Chr.7: 127589448-127589448 on Build GRCh38
rs751090975 Chr.7: 127589537-127589537 on Build GRCh38
rs759866266 Chr.7: 127589659-127589659 on Build GRCh38
rs924924089 Chr.7: 127589697-127589697 on Build GRCh38
rs371055003 Chr.7: 127589443-127589443 on Build GRCh38
rs932376589 Chr.7: 127589800-127589800 on Build GRCh38
rs34355308 Chr.7: 127589596-127589597 on Build GRCh38
rs767072120 Chr.7: 127589376-127589376 on Build GRCh38
rs754222019 Chr.7: 127589518-127589518 on Build GRCh38
rs934875160 Chr.7: 127589362-127589362 on Build GRCh38
rs912240810 Chr.7: 127589488-127589488 on Build GRCh38
rs767556326 Chr.7: 127589552-127589552 on Build GRCh38
rs747528741 Chr.7: 127589651-127589651 on Build GRCh38

View Full Product Details