Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 23370976-23371575 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF436-AS1
Entrez Gene ID
148898
Gene Name
ZNF436 antisense RNA 1
Gene Aliases
C1orf213
UniGene
Hs.61884
Gene Chromosome Location
Chr.1: 23368971-23371839 on Build GRCh38
Transcripts
NR_033690.1 NR_033691.1

SNP Detail

SNP ID Chromosome Location
rs949415003 Chr.1: 23371081-23371081 on Build GRCh38
rs958620573 Chr.1: 23371274-23371274 on Build GRCh38
rs771086870 Chr.1: 23371184-23371184 on Build GRCh38
rs767820922 Chr.1: 23371205-23371205 on Build GRCh38
rs751503234 Chr.1: 23371445-23371445 on Build GRCh38
rs748625350 Chr.1: 23371441-23371442 on Build GRCh38
rs756691877 Chr.1: 23371454-23371455 on Build GRCh38
rs757833348 Chr.1: 23371145-23371145 on Build GRCh38
rs764870750 Chr.1: 23371224-23371224 on Build GRCh38
rs143047446 Chr.1: 23371406-23371406 on Build GRCh38
rs927221594 Chr.1: 23371233-23371233 on Build GRCh38
rs755271394 Chr.1: 23371217-23371218 on Build GRCh38
rs775028899 Chr.1: 23371187-23371187 on Build GRCh38
rs1011164951 Chr.1: 23371275-23371275 on Build GRCh38
rs772879107 Chr.1: 23371513-23371513 on Build GRCh38
rs374669495 Chr.1: 23371520-23371520 on Build GRCh38
rs928590398 Chr.1: 23371040-23371040 on Build GRCh38
rs772627172 Chr.1: 23371502-23371502 on Build GRCh38
rs1052444390 Chr.1: 23371434-23371434 on Build GRCh38
rs774090111 Chr.1: 23371016-23371016 on Build GRCh38
rs979867595 Chr.1: 23371029-23371029 on Build GRCh38
rs952698270 Chr.1: 23371426-23371426 on Build GRCh38
rs535765420 Chr.1: 23371149-23371149 on Build GRCh38
rs570968705 Chr.1: 23371018-23371018 on Build GRCh38
rs538038841 Chr.1: 23371080-23371080 on Build GRCh38
rs748379703 Chr.1: 23371169-23371169 on Build GRCh38
rs764390527 Chr.1: 23371508-23371508 on Build GRCh38
rs890657900 Chr.1: 23371435-23371435 on Build GRCh38
rs545822759 Chr.1: 23371277-23371277 on Build GRCh38
rs766503415 Chr.1: 23371103-23371105 on Build GRCh38
rs747980087 Chr.1: 23371465-23371465 on Build GRCh38
rs560499467 Chr.1: 23371506-23371506 on Build GRCh38
rs755916068 Chr.1: 23371481-23371481 on Build GRCh38
rs960005334 Chr.1: 23371344-23371344 on Build GRCh38
rs370366451 Chr.1: 23371494-23371494 on Build GRCh38
rs376850613 Chr.1: 23371142-23371142 on Build GRCh38
rs371080704 Chr.1: 23371163-23371163 on Build GRCh38
rs778452230 Chr.1: 23371140-23371140 on Build GRCh38
rs905598515 Chr.1: 23371309-23371309 on Build GRCh38
rs766998259 Chr.1: 23371099-23371099 on Build GRCh38
rs576115190 Chr.1: 23371416-23371416 on Build GRCh38
rs775909545 Chr.1: 23371209-23371209 on Build GRCh38
rs373890599 Chr.1: 23371128-23371128 on Build GRCh38
rs757790413 Chr.1: 23371290-23371290 on Build GRCh38
rs746419450 Chr.1: 23371500-23371500 on Build GRCh38
rs760223154 Chr.1: 23371307-23371308 on Build GRCh38
rs751868838 Chr.1: 23371226-23371226 on Build GRCh38
rs780919518 Chr.1: 23371456-23371456 on Build GRCh38
rs973057513 Chr.1: 23371406-23371406 on Build GRCh38
rs374427515 Chr.1: 23371262-23371262 on Build GRCh38
rs761435602 Chr.1: 23371214-23371214 on Build GRCh38
rs377732804 Chr.1: 23371522-23371522 on Build GRCh38
rs984082902 Chr.1: 23371503-23371503 on Build GRCh38
rs960197562 Chr.1: 23371045-23371045 on Build GRCh38
rs116911595 Chr.1: 23371505-23371505 on Build GRCh38
rs767882277 Chr.1: 23371120-23371120 on Build GRCh38
rs911274569 Chr.1: 23371133-23371133 on Build GRCh38
rs891455387 Chr.1: 23371220-23371220 on Build GRCh38
rs370751796 Chr.1: 23371076-23371076 on Build GRCh38
rs765847480 Chr.1: 23371243-23371243 on Build GRCh38
rs34362736 Chr.1: 23371138-23371138 on Build GRCh38
rs749394683 Chr.1: 23371182-23371182 on Build GRCh38
rs754771741 Chr.1: 23371157-23371157 on Build GRCh38
rs552590253 Chr.1: 23371531-23371531 on Build GRCh38
rs935563944 Chr.1: 23371216-23371216 on Build GRCh38
rs900757052 Chr.1: 23371348-23371348 on Build GRCh38
rs638722 Chr.1: 23371124-23371124 on Build GRCh38
rs767505456 Chr.1: 23371539-23371539 on Build GRCh38
rs1052829229 Chr.1: 23371218-23371218 on Build GRCh38
rs765860799 Chr.1: 23371327-23371327 on Build GRCh38
rs183734892 Chr.1: 23371150-23371150 on Build GRCh38
rs969874468 Chr.1: 23371026-23371026 on Build GRCh38
rs544381109 Chr.1: 23371423-23371423 on Build GRCh38
rs371237287 Chr.1: 23371101-23371101 on Build GRCh38
rs752530185 Chr.1: 23371544-23371544 on Build GRCh38
rs1011279506 Chr.1: 23371007-23371007 on Build GRCh38
rs774005000 Chr.1: 23371405-23371406 on Build GRCh38
rs954464122 Chr.1: 23371181-23371181 on Build GRCh38
rs371215069 Chr.1: 23371529-23371529 on Build GRCh38
rs988558893 Chr.1: 23371204-23371204 on Build GRCh38
rs759841819 Chr.1: 23371115-23371115 on Build GRCh38
rs377562079 Chr.1: 23371151-23371151 on Build GRCh38
rs373931462 Chr.1: 23371177-23371177 on Build GRCh38
rs757923307 Chr.1: 23371231-23371231 on Build GRCh38
rs557563911 Chr.1: 23371310-23371310 on Build GRCh38
rs781706821 Chr.1: 23371218-23371218 on Build GRCh38
rs188450257 Chr.1: 23371482-23371482 on Build GRCh38
rs760055332 Chr.1: 23371198-23371198 on Build GRCh38
rs751844747 Chr.1: 23371205-23371207 on Build GRCh38
rs1037677291 Chr.1: 23371347-23371347 on Build GRCh38
rs754934848 Chr.1: 23371447-23371447 on Build GRCh38
rs767878537 Chr.1: 23371343-23371343 on Build GRCh38
rs369711350 Chr.1: 23371164-23371164 on Build GRCh38
rs907947466 Chr.1: 23371332-23371332 on Build GRCh38
rs568347741 Chr.1: 23371147-23371147 on Build GRCh38
rs972786735 Chr.1: 23371061-23371061 on Build GRCh38
rs114152668 Chr.1: 23371492-23371492 on Build GRCh38
rs763091609 Chr.1: 23371068-23371068 on Build GRCh38
rs996759395 Chr.1: 23371172-23371172 on Build GRCh38
rs955813517 Chr.1: 23371001-23371001 on Build GRCh38
rs375429453 Chr.1: 23371110-23371110 on Build GRCh38

View Full Product Details