Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 10906397-10906868 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIITA
Entrez Gene ID
4261
Gene Name
class II, major histocompatibility complex, transactivator
Gene Aliases
C2TA, CIITAIV, MHC2TA, NLRA
UniGene
Hs.592051
Gene Chromosome Location
Chr.16: 10866208-10941562 on Build GRCh38
Transcripts
NM_000246.3 NM_001286402.1 NM_001286403.1 NR_104444.1 XM_006720880.3 XM_011522484.2 XM_011522485.2 XM_011522486.2 XM_011522487.2 XM_011522488.2 XM_011522489.2 XM_011522490.2 XM_011522491.2 XM_011522492.2 XM_011522493.2 XM_011522494.2 XM_011522495.2 XM_011522496.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6303549 c.1332G>A p.Q444Q Chr.16: 10906824-10906824 on Build GRCh38
4578653 c.1225C>T p.R409W Chr.16: 10906717-10906717 on Build GRCh38
6838963 c.1351G>A p.V451I Chr.16: 10906843-10906843 on Build GRCh38
3505742 c.1154C>T p.P385L Chr.16: 10906646-10906646 on Build GRCh38
3505741 c.1141G>A p.E381K Chr.16: 10906633-10906633 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756879407 Chr.16: 10906796-10906796 on Build GRCh38
rs754076286 Chr.16: 10906467-10906467 on Build GRCh38
rs773334233 Chr.16: 10906747-10906747 on Build GRCh38
rs374576307 Chr.16: 10906558-10906558 on Build GRCh38
rs747411244 Chr.16: 10906735-10906735 on Build GRCh38
rs991589072 Chr.16: 10906762-10906762 on Build GRCh38
rs757559019 Chr.16: 10906535-10906535 on Build GRCh38
rs965811609 Chr.16: 10906437-10906437 on Build GRCh38
rs761770874 Chr.16: 10906617-10906617 on Build GRCh38
rs182369772 Chr.16: 10906660-10906660 on Build GRCh38
rs778649467 Chr.16: 10906646-10906646 on Build GRCh38
rs752871181 Chr.16: 10906476-10906476 on Build GRCh38
rs199476069 Chr.16: 10906722-10906722 on Build GRCh38
rs773675585 Chr.16: 10906845-10906845 on Build GRCh38
rs371950591 Chr.16: 10906462-10906462 on Build GRCh38
rs770306852 Chr.16: 10906643-10906643 on Build GRCh38
rs140683241 Chr.16: 10906536-10906536 on Build GRCh38
rs543881189 Chr.16: 10906446-10906446 on Build GRCh38
rs145558777 Chr.16: 10906669-10906669 on Build GRCh38
rs144735718 Chr.16: 10906539-10906539 on Build GRCh38
rs760539934 Chr.16: 10906786-10906786 on Build GRCh38
rs756276934 Chr.16: 10906525-10906525 on Build GRCh38
rs147670132 Chr.16: 10906502-10906502 on Build GRCh38
rs765200122 Chr.16: 10906805-10906805 on Build GRCh38
rs746768787 Chr.16: 10906491-10906491 on Build GRCh38
rs1009895562 Chr.16: 10906489-10906489 on Build GRCh38
rs113334711 Chr.16: 10906776-10906776 on Build GRCh38
rs762597561 Chr.16: 10906512-10906512 on Build GRCh38
rs758132900 Chr.16: 10906483-10906483 on Build GRCh38
rs750658139 Chr.16: 10906674-10906674 on Build GRCh38
rs145508828 Chr.16: 10906714-10906714 on Build GRCh38
rs766572360 Chr.16: 10906682-10906682 on Build GRCh38
rs752480407 Chr.16: 10906635-10906635 on Build GRCh38
rs190232414 Chr.16: 10906468-10906468 on Build GRCh38
rs776145930 Chr.16: 10906565-10906565 on Build GRCh38
rs772815490 Chr.16: 10906465-10906465 on Build GRCh38
rs377634337 Chr.16: 10906504-10906504 on Build GRCh38
rs984494067 Chr.16: 10906688-10906688 on Build GRCh38
rs759796074 Chr.16: 10906521-10906521 on Build GRCh38
rs930885758 Chr.16: 10906647-10906647 on Build GRCh38
rs568273747 Chr.16: 10906534-10906534 on Build GRCh38
rs886051638 Chr.16: 10906707-10906707 on Build GRCh38
rs777212471 Chr.16: 10906742-10906742 on Build GRCh38
rs779878144 Chr.16: 10906820-10906820 on Build GRCh38
rs763945523 Chr.16: 10906640-10906640 on Build GRCh38
rs975820291 Chr.16: 10906444-10906444 on Build GRCh38
rs763246371 Chr.16: 10906467-10906467 on Build GRCh38
rs766625949 Chr.16: 10906758-10906758 on Build GRCh38
rs772860663 Chr.16: 10906511-10906511 on Build GRCh38
rs764203096 Chr.16: 10906530-10906530 on Build GRCh38
rs756098713 Chr.16: 10906637-10906637 on Build GRCh38
rs765940448 Chr.16: 10906520-10906520 on Build GRCh38
rs142317834 Chr.16: 10906745-10906745 on Build GRCh38
rs754732155 Chr.16: 10906541-10906541 on Build GRCh38
rs909482150 Chr.16: 10906472-10906472 on Build GRCh38
rs953029793 Chr.16: 10906452-10906452 on Build GRCh38
rs755571997 Chr.16: 10906471-10906471 on Build GRCh38
rs756230223 Chr.16: 10906481-10906481 on Build GRCh38
rs539096456 Chr.16: 10906589-10906589 on Build GRCh38
rs768632103 Chr.16: 10906492-10906492 on Build GRCh38
rs372566990 Chr.16: 10906836-10906836 on Build GRCh38
rs753214558 Chr.16: 10906524-10906524 on Build GRCh38
rs749994940 Chr.16: 10906806-10906806 on Build GRCh38
rs548126822 Chr.16: 10906503-10906503 on Build GRCh38
rs757431582 Chr.16: 10906612-10906620 on Build GRCh38
rs886051639 Chr.16: 10906788-10906788 on Build GRCh38
rs148904933 Chr.16: 10906706-10906706 on Build GRCh38
rs762995873 Chr.16: 10906749-10906749 on Build GRCh38
rs199476068 Chr.16: 10906445-10906445 on Build GRCh38
rs768123074 Chr.16: 10906474-10906474 on Build GRCh38
rs748540952 Chr.16: 10906743-10906743 on Build GRCh38
rs144597413 Chr.16: 10906837-10906837 on Build GRCh38
rs763224509 Chr.16: 10906680-10906680 on Build GRCh38
rs928795511 Chr.16: 10906485-10906485 on Build GRCh38
rs759977402 Chr.16: 10906563-10906563 on Build GRCh38
rs777405510 Chr.16: 10906552-10906552 on Build GRCh38
rs779968345 Chr.16: 10906490-10906490 on Build GRCh38
rs776966289 Chr.16: 10906661-10906661 on Build GRCh38
rs572327540 Chr.16: 10906733-10906733 on Build GRCh38
rs769294144 Chr.16: 10906551-10906551 on Build GRCh38
rs746605931 Chr.16: 10906828-10906828 on Build GRCh38
rs745358587 Chr.16: 10906718-10906718 on Build GRCh38
rs137852602 Chr.16: 10906633-10906633 on Build GRCh38
rs372456158 Chr.16: 10906659-10906659 on Build GRCh38
rs769100548 Chr.16: 10906569-10906569 on Build GRCh38
rs754000556 Chr.16: 10906611-10906611 on Build GRCh38
rs137895901 Chr.16: 10906717-10906717 on Build GRCh38
rs761293424 Chr.16: 10906455-10906455 on Build GRCh38
rs570428236 Chr.16: 10906564-10906564 on Build GRCh38
rs201966261 Chr.16: 10906723-10906723 on Build GRCh38
rs371128355 Chr.16: 10906631-10906631 on Build GRCh38
rs755378764 Chr.16: 10906835-10906835 on Build GRCh38
rs763739028 Chr.16: 10906795-10906795 on Build GRCh38
rs368178094 Chr.16: 10906482-10906482 on Build GRCh38
rs527539420 Chr.16: 10906559-10906559 on Build GRCh38
rs767281522 Chr.16: 10906461-10906462 on Build GRCh38
rs201662987 Chr.16: 10906814-10906814 on Build GRCh38
rs771846946 Chr.16: 10906649-10906649 on Build GRCh38
rs556203901 Chr.16: 10906599-10906599 on Build GRCh38
rs761978819 Chr.16: 10906668-10906668 on Build GRCh38
rs750567359 Chr.16: 10906630-10906630 on Build GRCh38
rs147912483 Chr.16: 10906545-10906545 on Build GRCh38
rs757201187 Chr.16: 10906798-10906798 on Build GRCh38
rs748867171 Chr.16: 10906641-10906641 on Build GRCh38
rs953080654 Chr.16: 10906675-10906675 on Build GRCh38
rs535545291 Chr.16: 10906715-10906715 on Build GRCh38
rs765530997 Chr.16: 10906622-10906622 on Build GRCh38
rs752576726 Chr.16: 10906546-10906546 on Build GRCh38
rs76445850 Chr.16: 10906469-10906469 on Build GRCh38
rs752535345 Chr.16: 10906700-10906700 on Build GRCh38
rs768178240 Chr.16: 10906523-10906523 on Build GRCh38
rs140486686 Chr.16: 10906560-10906560 on Build GRCh38
rs958559248 Chr.16: 10906787-10906787 on Build GRCh38

View Full Product Details