Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67930086-67930674 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTRL
Entrez Gene ID
1506
Gene Name
chymotrypsin like
Gene Aliases
CTRL1
UniGene
Hs.654546
Gene Chromosome Location
Chr.16: 67929570-67931875 on Build GRCh38
Transcripts
NM_001907.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4579178 c.474C>T p.T158T Chr.16: 67930433-67930433 on Build GRCh38
972622 c.461C>T p.T154M Chr.16: 67930446-67930446 on Build GRCh38
6637741 c.643G>A p.G215R Chr.16: 67930086-67930086 on Build GRCh38
6637740 c.319-1G>T p.? Chr.16: 67930589-67930589 on Build GRCh38
2995899 c.319-2A>T p.? Chr.16: 67930590-67930590 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757480812 Chr.16: 67930408-67930408 on Build GRCh38
rs373052940 Chr.16: 67930547-67930547 on Build GRCh38
rs748560668 Chr.16: 67930168-67930168 on Build GRCh38
rs757525801 Chr.16: 67930494-67930494 on Build GRCh38
rs527379915 Chr.16: 67930169-67930169 on Build GRCh38
rs774498995 Chr.16: 67930309-67930337 on Build GRCh38
rs558031928 Chr.16: 67930432-67930432 on Build GRCh38
rs763914968 Chr.16: 67930137-67930137 on Build GRCh38
rs768948553 Chr.16: 67930116-67930116 on Build GRCh38
rs953168454 Chr.16: 67930368-67930368 on Build GRCh38
rs778195736 Chr.16: 67930382-67930382 on Build GRCh38
rs774698487 Chr.16: 67930118-67930118 on Build GRCh38
rs755722971 Chr.16: 67930223-67930223 on Build GRCh38
rs1030205196 Chr.16: 67930114-67930114 on Build GRCh38
rs1028414545 Chr.16: 67930339-67930339 on Build GRCh38
rs779062367 Chr.16: 67930604-67930604 on Build GRCh38
rs560251808 Chr.16: 67930493-67930493 on Build GRCh38
rs752630898 Chr.16: 67930133-67930133 on Build GRCh38
rs772433173 Chr.16: 67930280-67930280 on Build GRCh38
rs745712154 Chr.16: 67930609-67930609 on Build GRCh38
rs746126935 Chr.16: 67930412-67930412 on Build GRCh38
rs758986472 Chr.16: 67930544-67930544 on Build GRCh38
rs564361150 Chr.16: 67930191-67930191 on Build GRCh38
rs760359841 Chr.16: 67930318-67930318 on Build GRCh38
rs769628028 Chr.16: 67930276-67930276 on Build GRCh38
rs369956891 Chr.16: 67930306-67930306 on Build GRCh38
rs751718042 Chr.16: 67930172-67930172 on Build GRCh38
rs894659488 Chr.16: 67930330-67930330 on Build GRCh38
rs754443280 Chr.16: 67930332-67930332 on Build GRCh38
rs148841919 Chr.16: 67930469-67930469 on Build GRCh38
rs770921682 Chr.16: 67930606-67930606 on Build GRCh38
rs755452700 Chr.16: 67930243-67930243 on Build GRCh38
rs1031687227 Chr.16: 67930455-67930455 on Build GRCh38
rs74336993 Chr.16: 67930335-67930335 on Build GRCh38
rs564872445 Chr.16: 67930608-67930608 on Build GRCh38
rs767930831 Chr.16: 67930579-67930579 on Build GRCh38
rs753312924 Chr.16: 67930327-67930327 on Build GRCh38
rs764660418 Chr.16: 67930468-67930468 on Build GRCh38
rs748683548 Chr.16: 67930272-67930272 on Build GRCh38
rs780263014 Chr.16: 67930502-67930502 on Build GRCh38
rs772942369 Chr.16: 67930314-67930314 on Build GRCh38
rs749373891 Chr.16: 67930516-67930516 on Build GRCh38
rs780951372 Chr.16: 67930607-67930607 on Build GRCh38
rs767272614 Chr.16: 67930311-67930311 on Build GRCh38
rs201773939 Chr.16: 67930472-67930472 on Build GRCh38
rs762981045 Chr.16: 67930195-67930195 on Build GRCh38
rs200120972 Chr.16: 67930422-67930422 on Build GRCh38
rs771126434 Chr.16: 67930287-67930287 on Build GRCh38
rs776839938 Chr.16: 67930289-67930289 on Build GRCh38
rs958005816 Chr.16: 67930439-67930439 on Build GRCh38
rs762412511 Chr.16: 67930555-67930555 on Build GRCh38
rs372151792 Chr.16: 67930155-67930155 on Build GRCh38
rs201911333 Chr.16: 67930211-67930211 on Build GRCh38
rs374941235 Chr.16: 67930255-67930255 on Build GRCh38
rs535923319 Chr.16: 67930278-67930278 on Build GRCh38
rs754328333 Chr.16: 67930589-67930590 on Build GRCh38
rs776415766 Chr.16: 67930546-67930546 on Build GRCh38
rs922294011 Chr.16: 67930121-67930121 on Build GRCh38
rs772089153 Chr.16: 67930461-67930462 on Build GRCh38
rs768686532 Chr.16: 67930301-67930301 on Build GRCh38
rs764186734 Chr.16: 67930206-67930206 on Build GRCh38
rs766191852 Chr.16: 67930448-67930448 on Build GRCh38
rs1004607429 Chr.16: 67930291-67930291 on Build GRCh38
rs141530174 Chr.16: 67930128-67930128 on Build GRCh38
rs747900569 Chr.16: 67930127-67930127 on Build GRCh38
rs199681957 Chr.16: 67930598-67930598 on Build GRCh38
rs756379071 Chr.16: 67930589-67930589 on Build GRCh38
rs763692820 Chr.16: 67930566-67930566 on Build GRCh38
rs767995453 Chr.16: 67930626-67930626 on Build GRCh38
rs989280634 Chr.16: 67930256-67930256 on Build GRCh38
rs779233462 Chr.16: 67930167-67930167 on Build GRCh38
rs150887585 Chr.16: 67930159-67930159 on Build GRCh38
rs766066544 Chr.16: 67930321-67930321 on Build GRCh38
rs746032624 Chr.16: 67930500-67930500 on Build GRCh38
rs781710033 Chr.16: 67930499-67930499 on Build GRCh38
rs771956662 Chr.16: 67930433-67930433 on Build GRCh38
rs769494672 Chr.16: 67930549-67930549 on Build GRCh38
rs750046458 Chr.16: 67930222-67930222 on Build GRCh38
rs11552953 Chr.16: 67930458-67930458 on Build GRCh38
rs754118996 Chr.16: 67930595-67930595 on Build GRCh38
rs773443961 Chr.16: 67930175-67930175 on Build GRCh38
rs201248168 Chr.16: 67930158-67930158 on Build GRCh38
rs753144410 Chr.16: 67930338-67930338 on Build GRCh38
rs781607887 Chr.16: 67930411-67930411 on Build GRCh38
rs1037057911 Chr.16: 67930292-67930292 on Build GRCh38
rs762427384 Chr.16: 67930575-67930575 on Build GRCh38
rs1037699159 Chr.16: 67930548-67930548 on Build GRCh38
rs756467231 Chr.16: 67930501-67930501 on Build GRCh38
rs774967341 Chr.16: 67930621-67930621 on Build GRCh38
rs555968867 Chr.16: 67930132-67930132 on Build GRCh38
rs147636333 Chr.16: 67930446-67930446 on Build GRCh38
rs137951739 Chr.16: 67930572-67930572 on Build GRCh38
rs145332055 Chr.16: 67930539-67930539 on Build GRCh38
rs778011948 Chr.16: 67930281-67930281 on Build GRCh38
rs747392320 Chr.16: 67930405-67930405 on Build GRCh38
rs761682713 Chr.16: 67930310-67930310 on Build GRCh38
rs759785324 Chr.16: 67930355-67930356 on Build GRCh38
rs139670985 Chr.16: 67930456-67930456 on Build GRCh38
rs758834077 Chr.16: 67930347-67930347 on Build GRCh38
rs200353482 Chr.16: 67930581-67930581 on Build GRCh38
rs749873776 Chr.16: 67930156-67930156 on Build GRCh38
rs938182473 Chr.16: 67930266-67930266 on Build GRCh38
rs775044536 Chr.16: 67930554-67930554 on Build GRCh38
rs200944618 Chr.16: 67930525-67930525 on Build GRCh38
rs369491967 Chr.16: 67930538-67930538 on Build GRCh38
rs762691602 Chr.16: 67930624-67930624 on Build GRCh38
rs769216342 Chr.16: 67930162-67930162 on Build GRCh38
rs768184264 Chr.16: 67930638-67930638 on Build GRCh38
rs1031802012 Chr.16: 67930238-67930238 on Build GRCh38
rs982378123 Chr.16: 67930521-67930521 on Build GRCh38
rs751468951 Chr.16: 67930140-67930140 on Build GRCh38
rs750783578 Chr.16: 67930640-67930640 on Build GRCh38
rs758425028 Chr.16: 67930136-67930136 on Build GRCh38
rs775747112 Chr.16: 67930489-67930515 on Build GRCh38
rs756959771 Chr.16: 67930144-67930144 on Build GRCh38
rs1134760 Chr.16: 67930300-67930300 on Build GRCh38
rs769406670 Chr.16: 67930610-67930610 on Build GRCh38
rs769949436 Chr.16: 67930423-67930423 on Build GRCh38
rs760783713 Chr.16: 67930527-67930527 on Build GRCh38
rs761742626 Chr.16: 67930214-67930214 on Build GRCh38
rs753402362 Chr.16: 67930235-67930235 on Build GRCh38
rs773034873 Chr.16: 67930445-67930445 on Build GRCh38
rs531656200 Chr.16: 67930192-67930192 on Build GRCh38
rs202013679 Chr.16: 67930283-67930283 on Build GRCh38
rs201073485 Chr.16: 67930216-67930216 on Build GRCh38
rs764707859 Chr.16: 67930588-67930589 on Build GRCh38
rs575916915 Chr.16: 67930134-67930134 on Build GRCh38
rs368276407 Chr.16: 67930130-67930130 on Build GRCh38
rs775728878 Chr.16: 67930431-67930431 on Build GRCh38
rs762392658 Chr.16: 67930635-67930635 on Build GRCh38
rs780176504 Chr.16: 67930592-67930592 on Build GRCh38
rs746854480 Chr.16: 67930605-67930605 on Build GRCh38
rs779607998 Chr.16: 67930230-67930230 on Build GRCh38
rs878997572 Chr.16: 67930201-67930201 on Build GRCh38
rs1022132991 Chr.16: 67930268-67930268 on Build GRCh38
rs200154546 Chr.16: 67930120-67930120 on Build GRCh38
rs781051139 Chr.16: 67930126-67930127 on Build GRCh38

View Full Product Details