Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44520550-44520909 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEACAM20
Entrez Gene ID
125931
Gene Name
carcinoembryonic antigen related cell adhesion molecule 20
Gene Aliases
UNQ9366
UniGene
Hs.689632
Gene Chromosome Location
Chr.19: 44503838-44530100 on Build GRCh38
Transcripts
NM_001102597.2 NM_001102598.2 NM_001102599.2 NM_001102600.2 XM_011526429.2 XM_011526430.2 XM_011526431.2 XM_011526432.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6201378 c.949A>G p.N317D Chr.19: 44520555-44520555 on Build GRCh38
3535624 c.871G>A p.G291S Chr.19: 44520633-44520633 on Build GRCh38
1394479 c.910G>A p.A304T Chr.19: 44520594-44520594 on Build GRCh38
4977085 c.818A>G p.D273G Chr.19: 44520686-44520686 on Build GRCh38
5580601 c.858G>C p.Q286H Chr.19: 44520646-44520646 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs71171253 Chr.19: 44520869-44520870 on Build GRCh38
rs753817043 Chr.19: 44520844-44520845 on Build GRCh38
rs1005180499 Chr.19: 44520878-44520878 on Build GRCh38
rs538155382 Chr.19: 44520670-44520670 on Build GRCh38
rs372792332 Chr.19: 44520853-44520853 on Build GRCh38
rs371754410 Chr.19: 44520755-44520755 on Build GRCh38
rs935190926 Chr.19: 44520756-44520756 on Build GRCh38
rs781574800 Chr.19: 44520578-44520578 on Build GRCh38
rs755331826 Chr.19: 44520569-44520569 on Build GRCh38
rs770524597 Chr.19: 44520669-44520669 on Build GRCh38
rs199992941 Chr.19: 44520581-44520581 on Build GRCh38
rs576421937 Chr.19: 44520876-44520876 on Build GRCh38
rs370056603 Chr.19: 44520838-44520841 on Build GRCh38
rs764898114 Chr.19: 44520648-44520648 on Build GRCh38
rs753070558 Chr.19: 44520785-44520785 on Build GRCh38
rs57244256 Chr.19: 44520850-44520851 on Build GRCh38
rs993648773 Chr.19: 44520863-44520863 on Build GRCh38
rs371139859 Chr.19: 44520647-44520647 on Build GRCh38
rs571734560 Chr.19: 44520685-44520685 on Build GRCh38
rs567439171 Chr.19: 44520735-44520735 on Build GRCh38
rs374531494 Chr.19: 44520841-44520844 on Build GRCh38
rs777813540 Chr.19: 44520601-44520601 on Build GRCh38
rs889167620 Chr.19: 44520825-44520825 on Build GRCh38
rs776655285 Chr.19: 44520655-44520656 on Build GRCh38
rs771109952 Chr.19: 44520619-44520619 on Build GRCh38
rs112567919 Chr.19: 44520801-44520801 on Build GRCh38
rs779118825 Chr.19: 44520721-44520721 on Build GRCh38
rs774516586 Chr.19: 44520626-44520626 on Build GRCh38
rs1022889342 Chr.19: 44520680-44520680 on Build GRCh38
rs969549236 Chr.19: 44520676-44520676 on Build GRCh38
rs534153165 Chr.19: 44520694-44520694 on Build GRCh38
rs200315176 Chr.19: 44520628-44520628 on Build GRCh38
rs756587203 Chr.19: 44520681-44520681 on Build GRCh38
rs377659033 Chr.19: 44520634-44520634 on Build GRCh38
rs569489595 Chr.19: 44520643-44520643 on Build GRCh38
rs759815289 Chr.19: 44520690-44520691 on Build GRCh38
rs12975863 Chr.19: 44520841-44520841 on Build GRCh38
rs773938341 Chr.19: 44520762-44520762 on Build GRCh38
rs555482841 Chr.19: 44520839-44520840 on Build GRCh38
rs201619802 Chr.19: 44520730-44520730 on Build GRCh38
rs946670612 Chr.19: 44520809-44520809 on Build GRCh38
rs749557067 Chr.19: 44520616-44520616 on Build GRCh38
rs367917751 Chr.19: 44520741-44520741 on Build GRCh38
rs915191214 Chr.19: 44520803-44520803 on Build GRCh38
rs772826789 Chr.19: 44520658-44520658 on Build GRCh38
rs746801300 Chr.19: 44520631-44520631 on Build GRCh38
rs115198078 Chr.19: 44520672-44520672 on Build GRCh38
rs112984873 Chr.19: 44520850-44520850 on Build GRCh38
rs762737789 Chr.19: 44520664-44520664 on Build GRCh38
rs377383877 Chr.19: 44520688-44520688 on Build GRCh38
rs1050874739 Chr.19: 44520817-44520817 on Build GRCh38
rs922514112 Chr.19: 44520843-44520843 on Build GRCh38
rs762576327 Chr.19: 44520746-44520746 on Build GRCh38
rs796650954 Chr.19: 44520868-44520869 on Build GRCh38
rs769826753 Chr.19: 44520739-44520739 on Build GRCh38
rs75302460 Chr.19: 44520845-44520845 on Build GRCh38
rs767816836 Chr.19: 44520781-44520781 on Build GRCh38
rs757277740 Chr.19: 44520717-44520717 on Build GRCh38
rs952362514 Chr.19: 44520639-44520639 on Build GRCh38
rs756212642 Chr.19: 44520589-44520589 on Build GRCh38
rs765479154 Chr.19: 44520762-44520762 on Build GRCh38
rs760276464 Chr.19: 44520625-44520625 on Build GRCh38
rs962234106 Chr.19: 44520570-44520570 on Build GRCh38
rs370523022 Chr.19: 44520729-44520729 on Build GRCh38
rs879163361 Chr.19: 44520851-44520852 on Build GRCh38
rs374169311 Chr.19: 44520763-44520763 on Build GRCh38
rs111641915 Chr.19: 44520851-44520851 on Build GRCh38
rs556278877 Chr.19: 44520869-44520869 on Build GRCh38
rs766165296 Chr.19: 44520665-44520665 on Build GRCh38

View Full Product Details