Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 76004591-76005047 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXCL9
Entrez Gene ID
4283
Gene Name
C-X-C motif chemokine ligand 9
Gene Aliases
CMK, Humig, MIG, SCYB9, crg-10
UniGene
Hs.77367
Gene Chromosome Location
Chr.4: 76001342-76007523 on Build GRCh38
Transcripts
NM_002416.2 NR_125906.1

Gene Symbol
LOC101928809
Entrez Gene ID
101928809
Gene Name
uncharacterized LOC101928809
Gene Aliases
-
UniGene
Hs.637575
Gene Chromosome Location
Chr.4: 75980790-76005942 on Build GRCh38
Transcripts
NM_002416.2 NR_125906.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5897486 c.267G>A p.W89* Chr.4: 76004818-76004818 on Build GRCh38
5866750 c.276+2T>C p.? Chr.4: 76004807-76004807 on Build GRCh38
1431048 c.276+1G>T p.? Chr.4: 76004808-76004808 on Build GRCh38
4125990 c.276G>T p.Q92H Chr.4: 76004809-76004809 on Build GRCh38
7079890 c.209G>C p.G70A Chr.4: 76004876-76004876 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs182254922 Chr.4: 76004956-76004956 on Build GRCh38
rs773702858 Chr.4: 76004890-76004890 on Build GRCh38
rs958014692 Chr.4: 76004642-76004642 on Build GRCh38
rs926478781 Chr.4: 76004978-76004978 on Build GRCh38
rs762574216 Chr.4: 76004867-76004867 on Build GRCh38
rs767385020 Chr.4: 76004770-76004770 on Build GRCh38
rs765669638 Chr.4: 76004776-76004777 on Build GRCh38
rs940094317 Chr.4: 76004754-76004754 on Build GRCh38
rs1052555198 Chr.4: 76004896-76004896 on Build GRCh38
rs757119596 Chr.4: 76004758-76004758 on Build GRCh38
rs747595704 Chr.4: 76004808-76004808 on Build GRCh38
rs765471587 Chr.4: 76004944-76004944 on Build GRCh38
rs952990055 Chr.4: 76004622-76004622 on Build GRCh38
rs900046839 Chr.4: 76004717-76004717 on Build GRCh38
rs746164300 Chr.4: 76004846-76004846 on Build GRCh38
rs939127474 Chr.4: 76004640-76004640 on Build GRCh38
rs1049977282 Chr.4: 76004843-76004843 on Build GRCh38
rs376222900 Chr.4: 76004908-76004908 on Build GRCh38
rs189320006 Chr.4: 76004934-76004934 on Build GRCh38
rs1052122213 Chr.4: 76004648-76004648 on Build GRCh38
rs1007245548 Chr.4: 76004634-76004634 on Build GRCh38
rs765416292 Chr.4: 76004735-76004735 on Build GRCh38
rs372997222 Chr.4: 76004906-76004906 on Build GRCh38
rs997037318 Chr.4: 76004743-76004743 on Build GRCh38
rs773160990 Chr.4: 76004915-76004915 on Build GRCh38
rs750569935 Chr.4: 76004919-76004920 on Build GRCh38
rs578028142 Chr.4: 76004750-76004750 on Build GRCh38
rs1020708418 Chr.4: 76004926-76004926 on Build GRCh38
rs778270553 Chr.4: 76004791-76004791 on Build GRCh38
rs1032030911 Chr.4: 76004636-76004636 on Build GRCh38
rs762326022 Chr.4: 76004774-76004777 on Build GRCh38
rs761922086 Chr.4: 76004709-76004709 on Build GRCh38
rs558080224 Chr.4: 76004716-76004716 on Build GRCh38
rs901303891 Chr.4: 76004816-76004816 on Build GRCh38
rs760392187 Chr.4: 76004939-76004939 on Build GRCh38
rs41324144 Chr.4: 76004752-76004752 on Build GRCh38
rs980637541 Chr.4: 76004970-76004970 on Build GRCh38
rs759380326 Chr.4: 76004759-76004764 on Build GRCh38
rs987943497 Chr.4: 76004664-76004664 on Build GRCh38
rs890060522 Chr.4: 76004876-76004876 on Build GRCh38
rs926557220 Chr.4: 76004700-76004700 on Build GRCh38
rs755284441 Chr.4: 76004783-76004783 on Build GRCh38
rs970539025 Chr.4: 76004683-76004683 on Build GRCh38
rs532901786 Chr.4: 76004779-76004779 on Build GRCh38
rs779047088 Chr.4: 76004786-76004786 on Build GRCh38
rs998363388 Chr.4: 76004820-76004820 on Build GRCh38
rs769576198 Chr.4: 76004849-76004849 on Build GRCh38
rs758459547 Chr.4: 76004929-76004931 on Build GRCh38
rs529162253 Chr.4: 76004672-76004672 on Build GRCh38
rs776956921 Chr.4: 76004703-76004703 on Build GRCh38
rs369241957 Chr.4: 76004790-76004790 on Build GRCh38
rs569336277 Chr.4: 76004873-76004873 on Build GRCh38
rs1009690233 Chr.4: 76004919-76004919 on Build GRCh38
rs113881582 Chr.4: 76004692-76004692 on Build GRCh38
rs781665039 Chr.4: 76004845-76004845 on Build GRCh38
rs551057545 Chr.4: 76004842-76004842 on Build GRCh38
rs777008142 Chr.4: 76004763-76004763 on Build GRCh38

View Full Product Details