Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 157385720-157386317 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJB6
Entrez Gene ID
10049
Gene Name
DnaJ heat shock protein family (Hsp40) member B6
Gene Aliases
DJ4, DnaJ, HHDJ1, HSJ-2, HSJ2, LGMD1D, LGMD1E, MRJ, MSJ-1
UniGene
Hs.490745
Gene Chromosome Location
Chr.7: 157337004-157417439 on Build GRCh38
Transcripts
NM_005494.2 NM_058246.3 XM_005249515.2 XM_005249516.2 XM_006715823.1 XM_011515704.1 XM_017011633.1

SNP Detail

SNP ID Chromosome Location
rs971468326 Chr.7: 157385996-157385996 on Build GRCh38
rs768057270 Chr.7: 157385862-157385863 on Build GRCh38
rs1049267804 Chr.7: 157386099-157386099 on Build GRCh38
rs983671122 Chr.7: 157385813-157385813 on Build GRCh38
rs935725587 Chr.7: 157385887-157385887 on Build GRCh38
rs574721919 Chr.7: 157386206-157386206 on Build GRCh38
rs994133871 Chr.7: 157385902-157385902 on Build GRCh38
rs909056405 Chr.7: 157386131-157386131 on Build GRCh38
rs1024430002 Chr.7: 157385945-157385945 on Build GRCh38
rs931857799 Chr.7: 157385780-157385780 on Build GRCh38
rs923046243 Chr.7: 157386082-157386082 on Build GRCh38
rs186070488 Chr.7: 157386062-157386062 on Build GRCh38
rs994208782 Chr.7: 157386271-157386271 on Build GRCh38
rs901594725 Chr.7: 157385753-157385753 on Build GRCh38
rs995029815 Chr.7: 157385758-157385758 on Build GRCh38
rs535563639 Chr.7: 157385777-157385777 on Build GRCh38
rs562034247 Chr.7: 157385862-157385864 on Build GRCh38
rs1050665138 Chr.7: 157385799-157385799 on Build GRCh38
rs528833412 Chr.7: 157385873-157385873 on Build GRCh38
rs556767512 Chr.7: 157385784-157385784 on Build GRCh38
rs1051000074 Chr.7: 157386012-157386012 on Build GRCh38
rs980162437 Chr.7: 157385875-157385875 on Build GRCh38
rs762660517 Chr.7: 157386242-157386243 on Build GRCh38
rs200518051 Chr.7: 157386276-157386276 on Build GRCh38
rs989953951 Chr.7: 157385919-157385919 on Build GRCh38
rs900604506 Chr.7: 157386070-157386070 on Build GRCh38
rs141622946 Chr.7: 157385820-157385820 on Build GRCh38
rs537641438 Chr.7: 157386026-157386026 on Build GRCh38
rs1001226193 Chr.7: 157386201-157386201 on Build GRCh38
rs949416632 Chr.7: 157386281-157386281 on Build GRCh38
rs1024589081 Chr.7: 157386186-157386186 on Build GRCh38
rs1045749732 Chr.7: 157386287-157386287 on Build GRCh38
rs887726158 Chr.7: 157385804-157385804 on Build GRCh38
rs967052635 Chr.7: 157386013-157386013 on Build GRCh38
rs1039429864 Chr.7: 157386070-157386070 on Build GRCh38
rs989818057 Chr.7: 157386276-157386276 on Build GRCh38
rs913982470 Chr.7: 157386260-157386260 on Build GRCh38
rs375100576 Chr.7: 157386204-157386204 on Build GRCh38
rs1036695826 Chr.7: 157386173-157386173 on Build GRCh38
rs1036256492 Chr.7: 157385818-157385818 on Build GRCh38
rs971127137 Chr.7: 157385841-157385841 on Build GRCh38
rs1040110302 Chr.7: 157385751-157385751 on Build GRCh38
rs912907843 Chr.7: 157385747-157385747 on Build GRCh38
rs865956055 Chr.7: 157386195-157386195 on Build GRCh38
rs879148022 Chr.7: 157386261-157386261 on Build GRCh38
rs779635898 Chr.7: 157386245-157386246 on Build GRCh38
rs35732045 Chr.7: 157386260-157386261 on Build GRCh38
rs1050152529 Chr.7: 157386092-157386092 on Build GRCh38
rs913452424 Chr.7: 157386007-157386007 on Build GRCh38
rs1006562626 Chr.7: 157385812-157385812 on Build GRCh38
rs2366514 Chr.7: 157386244-157386244 on Build GRCh38
rs945678730 Chr.7: 157385749-157385749 on Build GRCh38
rs990811401 Chr.7: 157386006-157386006 on Build GRCh38
rs577573373 Chr.7: 157386086-157386086 on Build GRCh38
rs886567445 Chr.7: 157386104-157386104 on Build GRCh38
rs1034400683 Chr.7: 157386203-157386203 on Build GRCh38
rs1027802839 Chr.7: 157385793-157385793 on Build GRCh38
rs941834124 Chr.7: 157386153-157386153 on Build GRCh38
rs927087396 Chr.7: 157385880-157385880 on Build GRCh38
rs972038686 Chr.7: 157386198-157386198 on Build GRCh38
rs931717272 Chr.7: 157386090-157386090 on Build GRCh38
rs35152520 Chr.7: 157386243-157386244 on Build GRCh38
rs550595099 Chr.7: 157386172-157386175 on Build GRCh38
rs199729919 Chr.7: 157385841-157385842 on Build GRCh38
rs372347010 Chr.7: 157385808-157385809 on Build GRCh38
rs1034944898 Chr.7: 157386000-157386000 on Build GRCh38
rs762364468 Chr.7: 157385769-157385770 on Build GRCh38
rs541453050 Chr.7: 157386093-157386093 on Build GRCh38
rs957539231 Chr.7: 157386005-157386005 on Build GRCh38
rs1001733285 Chr.7: 157385999-157385999 on Build GRCh38

View Full Product Details