Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 110017353-110017847 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHCYL1
Entrez Gene ID
10768
Gene Name
adenosylhomocysteinase like 1
Gene Aliases
DCAL, IRBIT, PPP1R78, PRO0233, XPVKONA
UniGene
Hs.743973
Gene Chromosome Location
Chr.1: 109984741-110023742 on Build GRCh38
Transcripts
NM_001242673.1 NM_001242674.1 NM_001242675.1 NM_001242676.1 NM_006621.5 XM_011540535.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4140744 c.967G>T p.G323C Chr.1: 110017498-110017498 on Build GRCh38
1728867 c.967G>A p.G323S Chr.1: 110017498-110017498 on Build GRCh38
4912718 c.987T>A p.A329A Chr.1: 110017518-110017518 on Build GRCh38
3417845 c.1021G>T p.E341* Chr.1: 110017552-110017552 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1013134859 Chr.1: 110017804-110017804 on Build GRCh38
rs1012858597 Chr.1: 110017424-110017427 on Build GRCh38
rs749201627 Chr.1: 110017637-110017637 on Build GRCh38
rs879659369 Chr.1: 110017808-110017808 on Build GRCh38
rs561078720 Chr.1: 110017380-110017380 on Build GRCh38
rs909443352 Chr.1: 110017734-110017735 on Build GRCh38
rs748793308 Chr.1: 110017474-110017475 on Build GRCh38
rs985267044 Chr.1: 110017425-110017425 on Build GRCh38
rs372518177 Chr.1: 110017536-110017536 on Build GRCh38
rs1021482571 Chr.1: 110017448-110017448 on Build GRCh38
rs755031896 Chr.1: 110017593-110017593 on Build GRCh38
rs528274229 Chr.1: 110017599-110017599 on Build GRCh38
rs35533865 Chr.1: 110017731-110017731 on Build GRCh38
rs909666315 Chr.1: 110017431-110017431 on Build GRCh38
rs546769633 Chr.1: 110017417-110017417 on Build GRCh38
rs904407139 Chr.1: 110017820-110017820 on Build GRCh38
rs754386294 Chr.1: 110017591-110017591 on Build GRCh38
rs376729973 Chr.1: 110017604-110017604 on Build GRCh38
rs976898621 Chr.1: 110017588-110017588 on Build GRCh38
rs759077032 Chr.1: 110017523-110017523 on Build GRCh38
rs530259539 Chr.1: 110017386-110017386 on Build GRCh38
rs770772346 Chr.1: 110017793-110017793 on Build GRCh38
rs755611999 Chr.1: 110017469-110017469 on Build GRCh38
rs1043492996 Chr.1: 110017781-110017781 on Build GRCh38
rs567150035 Chr.1: 110017810-110017810 on Build GRCh38
rs776731240 Chr.1: 110017489-110017489 on Build GRCh38
rs551574907 Chr.1: 110017700-110017700 on Build GRCh38
rs777613724 Chr.1: 110017551-110017551 on Build GRCh38
rs919252519 Chr.1: 110017776-110017776 on Build GRCh38
rs550554642 Chr.1: 110017742-110017742 on Build GRCh38
rs533624092 Chr.1: 110017544-110017544 on Build GRCh38
rs200784996 Chr.1: 110017538-110017538 on Build GRCh38
rs562980952 Chr.1: 110017442-110017442 on Build GRCh38
rs369768930 Chr.1: 110017606-110017606 on Build GRCh38
rs571582737 Chr.1: 110017707-110017707 on Build GRCh38
rs770878550 Chr.1: 110017635-110017635 on Build GRCh38
rs894337009 Chr.1: 110017782-110017782 on Build GRCh38
rs770372358 Chr.1: 110017612-110017612 on Build GRCh38
rs866819070 Chr.1: 110017748-110017748 on Build GRCh38
rs748363317 Chr.1: 110017454-110017454 on Build GRCh38
rs368961402 Chr.1: 110017481-110017481 on Build GRCh38
rs775306764 Chr.1: 110017378-110017379 on Build GRCh38
rs536378386 Chr.1: 110017822-110017822 on Build GRCh38
rs938820274 Chr.1: 110017777-110017777 on Build GRCh38
rs778067153 Chr.1: 110017619-110017619 on Build GRCh38
rs748394745 Chr.1: 110017605-110017605 on Build GRCh38
rs756495353 Chr.1: 110017586-110017586 on Build GRCh38
rs942013638 Chr.1: 110017749-110017749 on Build GRCh38
rs1036330468 Chr.1: 110017764-110017764 on Build GRCh38
rs537305098 Chr.1: 110017728-110017728 on Build GRCh38
rs763919814 Chr.1: 110017800-110017800 on Build GRCh38
rs780501526 Chr.1: 110017587-110017587 on Build GRCh38
rs768758664 Chr.1: 110017450-110017450 on Build GRCh38
rs977032116 Chr.1: 110017491-110017491 on Build GRCh38
rs746750687 Chr.1: 110017622-110017622 on Build GRCh38
rs916347430 Chr.1: 110017708-110017708 on Build GRCh38
rs771883489 Chr.1: 110017468-110017468 on Build GRCh38
rs200320120 Chr.1: 110017485-110017485 on Build GRCh38
rs1029049950 Chr.1: 110017415-110017415 on Build GRCh38
rs774605189 Chr.1: 110017453-110017453 on Build GRCh38
rs750697489 Chr.1: 110017557-110017557 on Build GRCh38
rs550747242 Chr.1: 110017430-110017430 on Build GRCh38

View Full Product Details