Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 32734472-32735040 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAXBP1
Entrez Gene ID
94104
Gene Name
PAX3 and PAX7 binding protein 1
Gene Aliases
BM020, C21orf66, FSAP105, GCFC, GCFC1
UniGene
Hs.644004
Gene Chromosome Location
Chr.21: 32733899-32771858 on Build GRCh38
Transcripts
NM_016631.3 NR_027873.1 XM_006724066.1 NR_038879.1

Gene Symbol
PAXBP1-AS1
Entrez Gene ID
100506215
Gene Name
PAXBP1 antisense RNA 1
Gene Aliases
GCFC1-AS1
UniGene
Hs.657123
Gene Chromosome Location
Chr.21: 32728114-32743122 on Build GRCh38
Transcripts
NM_016631.3 NR_027873.1 XM_006724066.1 NR_038879.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1030239 c.2677C>T p.R893* Chr.21: 32735027-32735027 on Build GRCh38
7099118 c.2689C>T p.H897Y Chr.21: 32735015-32735015 on Build GRCh38
1030238 c.2689C>A p.H897N Chr.21: 32735015-32735015 on Build GRCh38
6489374 c.2694T>A p.A898A Chr.21: 32735010-32735010 on Build GRCh38
7155207 c.2721G>A p.V907V Chr.21: 32734983-32734983 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770144337 Chr.21: 32734987-32734987 on Build GRCh38
rs1009818719 Chr.21: 32734782-32734782 on Build GRCh38
rs147785424 Chr.21: 32734630-32734630 on Build GRCh38
rs746260675 Chr.21: 32734948-32734948 on Build GRCh38
rs570729984 Chr.21: 32734691-32734691 on Build GRCh38
rs148415052 Chr.21: 32734750-32734750 on Build GRCh38
rs917535419 Chr.21: 32734932-32734932 on Build GRCh38
rs569270227 Chr.21: 32734635-32734635 on Build GRCh38
rs983119759 Chr.21: 32734540-32734540 on Build GRCh38
rs755847726 Chr.21: 32734761-32734761 on Build GRCh38
rs973537176 Chr.21: 32734628-32734628 on Build GRCh38
rs755797875 Chr.21: 32734933-32734933 on Build GRCh38
rs983744092 Chr.21: 32734757-32734757 on Build GRCh38
rs895693855 Chr.21: 32734675-32734675 on Build GRCh38
rs745822834 Chr.21: 32734543-32734543 on Build GRCh38
rs773248636 Chr.21: 32734992-32734992 on Build GRCh38
rs574672047 Chr.21: 32734691-32734691 on Build GRCh38
rs976925937 Chr.21: 32734563-32734563 on Build GRCh38
rs766080046 Chr.21: 32735008-32735008 on Build GRCh38
rs778865658 Chr.21: 32734730-32734730 on Build GRCh38
rs768905578 Chr.21: 32734968-32734968 on Build GRCh38
rs954635628 Chr.21: 32734756-32734756 on Build GRCh38
rs1044208246 Chr.21: 32734693-32734693 on Build GRCh38
rs200465600 Chr.21: 32734923-32734923 on Build GRCh38
rs76923710 Chr.21: 32734588-32734588 on Build GRCh38
rs143512881 Chr.21: 32734961-32734961 on Build GRCh38
rs373333916 Chr.21: 32734915-32734915 on Build GRCh38
rs568578363 Chr.21: 32734891-32734891 on Build GRCh38
rs1018583636 Chr.21: 32734876-32734876 on Build GRCh38
rs547478950 Chr.21: 32734982-32734982 on Build GRCh38
rs965631295 Chr.21: 32734902-32734902 on Build GRCh38
rs748197981 Chr.21: 32734986-32734986 on Build GRCh38
rs977439581 Chr.21: 32734940-32734940 on Build GRCh38
rs559426518 Chr.21: 32734542-32734542 on Build GRCh38
rs752157441 Chr.21: 32734924-32734924 on Build GRCh38
rs116339270 Chr.21: 32734962-32734962 on Build GRCh38
rs950027523 Chr.21: 32734686-32734686 on Build GRCh38
rs1030196524 Chr.21: 32734500-32734500 on Build GRCh38
rs531114895 Chr.21: 32734636-32734636 on Build GRCh38
rs762586001 Chr.21: 32734913-32734913 on Build GRCh38
rs781317673 Chr.21: 32734526-32734526 on Build GRCh38
rs1018278647 Chr.21: 32734739-32734739 on Build GRCh38
rs995961334 Chr.21: 32734747-32734751 on Build GRCh38
rs937533424 Chr.21: 32734892-32734892 on Build GRCh38
rs773544499 Chr.21: 32734862-32734862 on Build GRCh38
rs369016103 Chr.21: 32734906-32734906 on Build GRCh38
rs762861716 Chr.21: 32734996-32734996 on Build GRCh38
rs937458370 Chr.21: 32734631-32734631 on Build GRCh38
rs935990060 Chr.21: 32734994-32734994 on Build GRCh38
rs531999511 Chr.21: 32734741-32734741 on Build GRCh38
rs776872837 Chr.21: 32734976-32734976 on Build GRCh38
rs372725852 Chr.21: 32734945-32734945 on Build GRCh38
rs750885622 Chr.21: 32734927-32734928 on Build GRCh38
rs940035887 Chr.21: 32734661-32734661 on Build GRCh38
rs200811797 Chr.21: 32735001-32735001 on Build GRCh38
rs577903328 Chr.21: 32734660-32734660 on Build GRCh38
rs376031602 Chr.21: 32734967-32734967 on Build GRCh38
rs751899153 Chr.21: 32734751-32734752 on Build GRCh38
rs891556500 Chr.21: 32734771-32734771 on Build GRCh38
rs552427134 Chr.21: 32734666-32734666 on Build GRCh38
rs756581949 Chr.21: 32734987-32734988 on Build GRCh38
rs540852092 Chr.21: 32734508-32734508 on Build GRCh38

View Full Product Details