Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 14689819-14690391 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf20
Entrez Gene ID
84077
Gene Name
chromosome 3 open reading frame 20
Gene Aliases
-
UniGene
Hs.661452
Gene Chromosome Location
Chr.3: 14673532-14773036 on Build GRCh38
Transcripts
NM_001184957.1 NM_001184958.1 NM_032137.4 XM_006713341.2 XM_006713342.2 XM_006713343.2 XM_011534153.2 XM_011534154.2 XM_011534157.2 XM_011534158.2 XM_017007310.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1039636 c.633C>T p.L211L Chr.3: 14690004-14690004 on Build GRCh38
4114290 c.726A>C p.K242N Chr.3: 14690097-14690097 on Build GRCh38
7129390 c.652G>A p.G218R Chr.3: 14690023-14690023 on Build GRCh38
4387998 c.646G>A p.A216T Chr.3: 14690017-14690017 on Build GRCh38
3915092 c.667T>G p.Y223D Chr.3: 14690038-14690038 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs545224682 Chr.3: 14690044-14690044 on Build GRCh38
rs752220067 Chr.3: 14689994-14689994 on Build GRCh38
rs934157746 Chr.3: 14690218-14690218 on Build GRCh38
rs878865650 Chr.3: 14690170-14690170 on Build GRCh38
rs115653711 Chr.3: 14690079-14690079 on Build GRCh38
rs772253785 Chr.3: 14690007-14690007 on Build GRCh38
rs989240422 Chr.3: 14690355-14690355 on Build GRCh38
rs190393325 Chr.3: 14690303-14690303 on Build GRCh38
rs766995153 Chr.3: 14689993-14689993 on Build GRCh38
rs368628973 Chr.3: 14690132-14690132 on Build GRCh38
rs371029457 Chr.3: 14690083-14690083 on Build GRCh38
rs200768036 Chr.3: 14690004-14690004 on Build GRCh38
rs760799279 Chr.3: 14690167-14690174 on Build GRCh38
rs775623218 Chr.3: 14689960-14689960 on Build GRCh38
rs1044874045 Chr.3: 14689839-14689839 on Build GRCh38
rs769445239 Chr.3: 14689917-14689917 on Build GRCh38
rs374531507 Chr.3: 14690147-14690147 on Build GRCh38
rs745987667 Chr.3: 14689976-14689979 on Build GRCh38
rs971721067 Chr.3: 14689985-14689985 on Build GRCh38
rs759621916 Chr.3: 14690370-14690370 on Build GRCh38
rs199721807 Chr.3: 14689979-14689979 on Build GRCh38
rs187372155 Chr.3: 14690016-14690016 on Build GRCh38
rs1006353511 Chr.3: 14690189-14690189 on Build GRCh38
rs368271638 Chr.3: 14689938-14689938 on Build GRCh38
rs777709119 Chr.3: 14690005-14690005 on Build GRCh38
rs1432601 Chr.3: 14690301-14690301 on Build GRCh38
rs745312944 Chr.3: 14689910-14689910 on Build GRCh38
rs544547536 Chr.3: 14689878-14689878 on Build GRCh38
rs751178198 Chr.3: 14689981-14689981 on Build GRCh38
rs761155697 Chr.3: 14689961-14689961 on Build GRCh38
rs915154462 Chr.3: 14690297-14690297 on Build GRCh38
rs118084922 Chr.3: 14690166-14690166 on Build GRCh38
rs374809461 Chr.3: 14690110-14690110 on Build GRCh38
rs921391767 Chr.3: 14690182-14690182 on Build GRCh38
rs775910494 Chr.3: 14690060-14690061 on Build GRCh38
rs746131672 Chr.3: 14690017-14690017 on Build GRCh38
rs893703600 Chr.3: 14690081-14690081 on Build GRCh38
rs201159660 Chr.3: 14690133-14690133 on Build GRCh38
rs547182966 Chr.3: 14689932-14689932 on Build GRCh38
rs983478278 Chr.3: 14689908-14689908 on Build GRCh38
rs772330997 Chr.3: 14690020-14690020 on Build GRCh38
rs767174595 Chr.3: 14690114-14690114 on Build GRCh38
rs935012893 Chr.3: 14690266-14690266 on Build GRCh38
rs962969799 Chr.3: 14690176-14690176 on Build GRCh38
rs757046631 Chr.3: 14690012-14690012 on Build GRCh38
rs757171157 Chr.3: 14690151-14690151 on Build GRCh38
rs9830589 Chr.3: 14690116-14690116 on Build GRCh38
rs961790857 Chr.3: 14690236-14690236 on Build GRCh38
rs146624816 Chr.3: 14690021-14690021 on Build GRCh38
rs377496570 Chr.3: 14690065-14690065 on Build GRCh38
rs1054458792 Chr.3: 14689875-14689875 on Build GRCh38
rs975992816 Chr.3: 14690181-14690181 on Build GRCh38
rs934987597 Chr.3: 14690069-14690069 on Build GRCh38
rs868866593 Chr.3: 14690018-14690018 on Build GRCh38
rs575916421 Chr.3: 14689861-14689861 on Build GRCh38
rs372889451 Chr.3: 14690033-14690033 on Build GRCh38
rs530849610 Chr.3: 14690089-14690089 on Build GRCh38
rs149190031 Chr.3: 14690034-14690034 on Build GRCh38
rs17852774 Chr.3: 14690060-14690060 on Build GRCh38
rs376158012 Chr.3: 14690011-14690011 on Build GRCh38
rs953513012 Chr.3: 14689933-14689933 on Build GRCh38
rs957953188 Chr.3: 14690331-14690331 on Build GRCh38
rs747065741 Chr.3: 14690028-14690028 on Build GRCh38
rs1016447659 Chr.3: 14690216-14690216 on Build GRCh38
rs753783364 Chr.3: 14690137-14690137 on Build GRCh38
rs1023440385 Chr.3: 14690123-14690123 on Build GRCh38
rs921697853 Chr.3: 14689987-14689987 on Build GRCh38
rs758599217 Chr.3: 14690165-14690165 on Build GRCh38
rs200040671 Chr.3: 14690059-14690059 on Build GRCh38
rs566232895 Chr.3: 14690244-14690244 on Build GRCh38
rs982124030 Chr.3: 14689880-14689880 on Build GRCh38
rs373183860 Chr.3: 14689959-14689959 on Build GRCh38
rs764669177 Chr.3: 14689966-14689966 on Build GRCh38
rs780778966 Chr.3: 14689904-14689904 on Build GRCh38
rs75938220 Chr.3: 14690062-14690062 on Build GRCh38
rs750205548 Chr.3: 14690159-14690159 on Build GRCh38
rs559124794 Chr.3: 14690300-14690300 on Build GRCh38
rs374982515 Chr.3: 14690167-14690167 on Build GRCh38
rs777187494 Chr.3: 14689970-14689970 on Build GRCh38
rs762294394 Chr.3: 14689973-14689973 on Build GRCh38
rs987215334 Chr.3: 14690241-14690241 on Build GRCh38
rs765773169 Chr.3: 14689879-14689879 on Build GRCh38
rs371682903 Chr.3: 14690158-14690158 on Build GRCh38
rs1006427569 Chr.3: 14689975-14689976 on Build GRCh38
rs546149890 Chr.3: 14690240-14690240 on Build GRCh38
rs367731559 Chr.3: 14690157-14690157 on Build GRCh38
rs200754472 Chr.3: 14690172-14690172 on Build GRCh38
rs144480539 Chr.3: 14689976-14689977 on Build GRCh38
rs375588541 Chr.3: 14690097-14690097 on Build GRCh38
rs905055651 Chr.3: 14689844-14689846 on Build GRCh38
rs538318699 Chr.3: 14690283-14690283 on Build GRCh38
rs540775293 Chr.3: 14689996-14689996 on Build GRCh38
rs554984644 Chr.3: 14689903-14689903 on Build GRCh38
rs754601075 Chr.3: 14689983-14689983 on Build GRCh38
rs148859823 Chr.3: 14690048-14690048 on Build GRCh38

View Full Product Details