Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 65669354-65669920 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND4A
Entrez Gene ID
10260
Gene Name
DENN domain containing 4A
Gene Aliases
IRLB, MYCPBP
UniGene
Hs.654567
Gene Chromosome Location
Chr.15: 65659123-65792293 on Build GRCh38
Transcripts
NM_001144823.2 NM_001320835.1 NM_005848.4 XM_005254120.4 XM_005254121.3 XM_005254122.4 XM_006720371.3 XM_011521157.2 XM_017021862.1 XM_017021863.1 XM_017021864.1 XM_017021865.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5447156 c.4581G>T p.Q1527H Chr.15: 65669853-65669853 on Build GRCh38
5772515 c.4556C>A p.S1519Y Chr.15: 65669878-65669878 on Build GRCh38
3711726 c.4774G>C p.D1592H Chr.15: 65669789-65669789 on Build GRCh38
3719011 c.4621G>A p.A1541T Chr.15: 65669813-65669813 on Build GRCh38
7364110 c.4750G>T p.A1584S Chr.15: 65669813-65669813 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760850251 Chr.15: 65669882-65669882 on Build GRCh38
rs947481792 Chr.15: 65669568-65669568 on Build GRCh38
rs756244551 Chr.15: 65669741-65669741 on Build GRCh38
rs139132750 Chr.15: 65669467-65669467 on Build GRCh38
rs978579481 Chr.15: 65669605-65669605 on Build GRCh38
rs1021303599 Chr.15: 65669664-65669664 on Build GRCh38
rs540461440 Chr.15: 65669663-65669663 on Build GRCh38
rs1002741294 Chr.15: 65669625-65669625 on Build GRCh38
rs895513369 Chr.15: 65669440-65669440 on Build GRCh38
rs750307625 Chr.15: 65669801-65669801 on Build GRCh38
rs935856987 Chr.15: 65669447-65669469 on Build GRCh38
rs538312136 Chr.15: 65669479-65669479 on Build GRCh38
rs1036566409 Chr.15: 65669612-65669612 on Build GRCh38
rs907043482 Chr.15: 65669622-65669622 on Build GRCh38
rs201852758 Chr.15: 65669867-65669867 on Build GRCh38
rs1057426781 Chr.15: 65669676-65669676 on Build GRCh38
rs1012654019 Chr.15: 65669445-65669445 on Build GRCh38
rs558633756 Chr.15: 65669465-65669465 on Build GRCh38
rs754212947 Chr.15: 65669697-65669697 on Build GRCh38
rs764215682 Chr.15: 65669890-65669890 on Build GRCh38
rs753798130 Chr.15: 65669796-65669796 on Build GRCh38
rs8036784 Chr.15: 65669644-65669644 on Build GRCh38
rs199856490 Chr.15: 65669810-65669810 on Build GRCh38
rs554493363 Chr.15: 65669715-65669715 on Build GRCh38
rs768244115 Chr.15: 65669840-65669840 on Build GRCh38
rs77436088 Chr.15: 65669795-65669795 on Build GRCh38
rs763902936 Chr.15: 65669771-65669771 on Build GRCh38
rs746454114 Chr.15: 65669817-65669817 on Build GRCh38
rs1019597065 Chr.15: 65669505-65669505 on Build GRCh38
rs774633216 Chr.15: 65669856-65669856 on Build GRCh38
rs376354896 Chr.15: 65669461-65669461 on Build GRCh38
rs555077406 Chr.15: 65669589-65669589 on Build GRCh38
rs756202942 Chr.15: 65669576-65669576 on Build GRCh38
rs912747383 Chr.15: 65669431-65669431 on Build GRCh38
rs746572726 Chr.15: 65669859-65669859 on Build GRCh38
rs780718297 Chr.15: 65669846-65669846 on Build GRCh38
rs372616644 Chr.15: 65669633-65669634 on Build GRCh38
rs71396469 Chr.15: 65669466-65669466 on Build GRCh38
rs921380233 Chr.15: 65669545-65669545 on Build GRCh38
rs780239167 Chr.15: 65669430-65669432 on Build GRCh38
rs887163617 Chr.15: 65669541-65669541 on Build GRCh38
rs1034264658 Chr.15: 65669631-65669631 on Build GRCh38
rs983453632 Chr.15: 65669499-65669499 on Build GRCh38
rs977199610 Chr.15: 65669680-65669680 on Build GRCh38
rs905354069 Chr.15: 65669450-65669450 on Build GRCh38
rs752404659 Chr.15: 65669406-65669406 on Build GRCh38
rs544878548 Chr.15: 65669492-65669492 on Build GRCh38
rs948402472 Chr.15: 65669732-65669732 on Build GRCh38
rs749771490 Chr.15: 65669848-65669848 on Build GRCh38
rs8036725 Chr.15: 65669852-65669852 on Build GRCh38
rs975578999 Chr.15: 65669756-65669756 on Build GRCh38
rs533300984 Chr.15: 65669860-65669860 on Build GRCh38
rs546910119 Chr.15: 65669389-65669389 on Build GRCh38
rs1048412293 Chr.15: 65669532-65669532 on Build GRCh38
rs965371381 Chr.15: 65669509-65669509 on Build GRCh38
rs965508296 Chr.15: 65669755-65669755 on Build GRCh38
rs756687860 Chr.15: 65669507-65669507 on Build GRCh38
rs940139317 Chr.15: 65669602-65669602 on Build GRCh38
rs566645182 Chr.15: 65669408-65669408 on Build GRCh38
rs967507389 Chr.15: 65669665-65669665 on Build GRCh38
rs778834273 Chr.15: 65669798-65669798 on Build GRCh38
rs369864397 Chr.15: 65669813-65669813 on Build GRCh38

View Full Product Details