Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 120739599-120740143 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACADS
Entrez Gene ID
35
Gene Name
acyl-CoA dehydrogenase, C-2 to C-3 short chain
Gene Aliases
ACAD3, SCAD
UniGene
Hs.507076
Gene Chromosome Location
Chr.12: 120725738-120740008 on Build GRCh38
Transcripts
NM_000017.3 NM_001302554.1

SNP Detail

SNP ID Chromosome Location
rs17848083 Chr.12: 120740063-120740063 on Build GRCh38
rs537512881 Chr.12: 120739714-120739714 on Build GRCh38
rs934165933 Chr.12: 120739857-120739857 on Build GRCh38
rs971566729 Chr.12: 120739914-120739914 on Build GRCh38
rs1019054605 Chr.12: 120739822-120739822 on Build GRCh38
rs1050365579 Chr.12: 120740038-120740038 on Build GRCh38
rs116733542 Chr.12: 120739758-120739758 on Build GRCh38
rs981839041 Chr.12: 120739666-120739666 on Build GRCh38
rs369720260 Chr.12: 120739721-120739721 on Build GRCh38
rs981556913 Chr.12: 120739932-120739932 on Build GRCh38
rs1051175241 Chr.12: 120739858-120739858 on Build GRCh38
rs17848081 Chr.12: 120740021-120740021 on Build GRCh38
rs761842948 Chr.12: 120739818-120739818 on Build GRCh38
rs1023144602 Chr.12: 120739867-120739867 on Build GRCh38
rs550652942 Chr.12: 120739753-120739753 on Build GRCh38
rs898798115 Chr.12: 120739751-120739751 on Build GRCh38
rs979788409 Chr.12: 120740043-120740043 on Build GRCh38
rs539430171 Chr.12: 120739764-120739764 on Build GRCh38
rs993837906 Chr.12: 120739969-120739969 on Build GRCh38
rs17848082 Chr.12: 120740025-120740025 on Build GRCh38
rs940390916 Chr.12: 120739698-120739698 on Build GRCh38
rs9204 Chr.12: 120739975-120739975 on Build GRCh38
rs890051751 Chr.12: 120739870-120739870 on Build GRCh38
rs113752987 Chr.12: 120740008-120740008 on Build GRCh38
rs769916996 Chr.12: 120740095-120740095 on Build GRCh38
rs1025383241 Chr.12: 120739732-120739732 on Build GRCh38
rs1023443903 Chr.12: 120740039-120740039 on Build GRCh38
rs980138444 Chr.12: 120739772-120739772 on Build GRCh38
rs748219959 Chr.12: 120740053-120740053 on Build GRCh38
rs11065238 Chr.12: 120739835-120739835 on Build GRCh38
rs886049032 Chr.12: 120739622-120739622 on Build GRCh38
rs999346811 Chr.12: 120740117-120740117 on Build GRCh38
rs564150245 Chr.12: 120740069-120740069 on Build GRCh38
rs628909 Chr.12: 120739697-120739697 on Build GRCh38
rs780335677 Chr.12: 120739831-120739831 on Build GRCh38
rs192425836 Chr.12: 120739829-120739829 on Build GRCh38
rs137990231 Chr.12: 120739648-120739648 on Build GRCh38
rs902980587 Chr.12: 120740113-120740113 on Build GRCh38
rs927773299 Chr.12: 120739945-120739945 on Build GRCh38
rs998301901 Chr.12: 120739642-120739642 on Build GRCh38
rs916079637 Chr.12: 120739637-120739637 on Build GRCh38
rs548735871 Chr.12: 120739653-120739653 on Build GRCh38
rs957326664 Chr.12: 120739842-120739842 on Build GRCh38
rs930266628 Chr.12: 120740035-120740035 on Build GRCh38
rs991716792 Chr.12: 120739860-120739860 on Build GRCh38
rs760566413 Chr.12: 120740040-120740040 on Build GRCh38
rs889999763 Chr.12: 120739689-120739689 on Build GRCh38
rs184552056 Chr.12: 120740026-120740026 on Build GRCh38
rs373553465 Chr.12: 120739765-120739765 on Build GRCh38
rs639667 Chr.12: 120739948-120739948 on Build GRCh38
rs1008725542 Chr.12: 120739786-120739786 on Build GRCh38

View Full Product Details