Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44756719-44757318 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL3
Entrez Gene ID
602
Gene Name
B-cell CLL/lymphoma 3
Gene Aliases
BCL4, D19S37
UniGene
Hs.31210
Gene Chromosome Location
Chr.19: 44742621-44760044 on Build GRCh38
Transcripts
NM_005178.4 XM_011527197.2 XM_011527198.2 XM_011527200.2 XM_017027109.1 XM_017027110.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4928003 c.524C>T p.P175L Chr.19: 44757021-44757021 on Build GRCh38
1325199 c.613G>A p.G205S Chr.19: 44757110-44757110 on Build GRCh38
3535742 c.646C>T p.R216C Chr.19: 44757143-44757143 on Build GRCh38
1394495 c.647G>A p.R216H Chr.19: 44757144-44757144 on Build GRCh38
3389121 c.520-4C>T p.? Chr.19: 44757013-44757013 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs933739227 Chr.19: 44756777-44756777 on Build GRCh38
rs935747685 Chr.19: 44756832-44756832 on Build GRCh38
rs1038255791 Chr.19: 44756821-44756821 on Build GRCh38
rs758480693 Chr.19: 44756974-44756974 on Build GRCh38
rs531798695 Chr.19: 44756985-44756985 on Build GRCh38
rs775921841 Chr.19: 44757122-44757122 on Build GRCh38
rs757076905 Chr.19: 44757178-44757178 on Build GRCh38
rs766582877 Chr.19: 44757145-44757145 on Build GRCh38
rs779825081 Chr.19: 44757188-44757188 on Build GRCh38
rs778502369 Chr.19: 44757182-44757182 on Build GRCh38
rs754616800 Chr.19: 44757290-44757290 on Build GRCh38
rs763828094 Chr.19: 44757162-44757162 on Build GRCh38
rs200056289 Chr.19: 44757282-44757282 on Build GRCh38
rs767312366 Chr.19: 44756977-44756977 on Build GRCh38
rs758001387 Chr.19: 44757284-44757284 on Build GRCh38
rs763413198 Chr.19: 44757148-44757148 on Build GRCh38
rs770186550 Chr.19: 44757214-44757214 on Build GRCh38
rs549192479 Chr.19: 44757259-44757259 on Build GRCh38
rs1055233231 Chr.19: 44756861-44756861 on Build GRCh38
rs781662900 Chr.19: 44757291-44757291 on Build GRCh38
rs771802418 Chr.19: 44757289-44757289 on Build GRCh38
rs377622191 Chr.19: 44757000-44757000 on Build GRCh38
rs778248294 Chr.19: 44757004-44757004 on Build GRCh38
rs920991250 Chr.19: 44756746-44756746 on Build GRCh38
rs113972085 Chr.19: 44757118-44757118 on Build GRCh38
rs747854207 Chr.19: 44757285-44757285 on Build GRCh38
rs554053234 Chr.19: 44756894-44756894 on Build GRCh38
rs765943465 Chr.19: 44757287-44757287 on Build GRCh38
rs746607839 Chr.19: 44757193-44757193 on Build GRCh38
rs751258622 Chr.19: 44757289-44757289 on Build GRCh38
rs937923434 Chr.19: 44757136-44757136 on Build GRCh38
rs902695375 Chr.19: 44757219-44757219 on Build GRCh38
rs546909663 Chr.19: 44757177-44757177 on Build GRCh38
rs939576968 Chr.19: 44757260-44757260 on Build GRCh38
rs748699383 Chr.19: 44757198-44757198 on Build GRCh38
rs778958434 Chr.19: 44756817-44756817 on Build GRCh38
rs756816090 Chr.19: 44757001-44757001 on Build GRCh38
rs568970988 Chr.19: 44757279-44757279 on Build GRCh38
rs866079348 Chr.19: 44757222-44757222 on Build GRCh38
rs949538010 Chr.19: 44757179-44757179 on Build GRCh38
rs769332537 Chr.19: 44757091-44757091 on Build GRCh38
rs369271131 Chr.19: 44757234-44757234 on Build GRCh38
rs759715721 Chr.19: 44757250-44757250 on Build GRCh38
rs35980686 Chr.19: 44757093-44757093 on Build GRCh38
rs138512809 Chr.19: 44757137-44757137 on Build GRCh38
rs770116824 Chr.19: 44757036-44757036 on Build GRCh38
rs376224051 Chr.19: 44757217-44757217 on Build GRCh38
rs752068025 Chr.19: 44757237-44757237 on Build GRCh38
rs939960430 Chr.19: 44756798-44756798 on Build GRCh38
rs758195840 Chr.19: 44757184-44757184 on Build GRCh38
rs528515769 Chr.19: 44756834-44756834 on Build GRCh38
rs987068395 Chr.19: 44756779-44756779 on Build GRCh38
rs747679905 Chr.19: 44756870-44756870 on Build GRCh38
rs375139748 Chr.19: 44756777-44756778 on Build GRCh38
rs371678460 Chr.19: 44756990-44756990 on Build GRCh38
rs957362834 Chr.19: 44757150-44757150 on Build GRCh38
rs529075140 Chr.19: 44757253-44757253 on Build GRCh38
rs565238653 Chr.19: 44757029-44757029 on Build GRCh38
rs937980921 Chr.19: 44756859-44756859 on Build GRCh38
rs781343322 Chr.19: 44757196-44757196 on Build GRCh38
rs771795873 Chr.19: 44757023-44757023 on Build GRCh38
rs769169222 Chr.19: 44757194-44757194 on Build GRCh38
rs546578138 Chr.19: 44756918-44756918 on Build GRCh38
rs773105917 Chr.19: 44757133-44757133 on Build GRCh38
rs908368326 Chr.19: 44756786-44756786 on Build GRCh38
rs760441291 Chr.19: 44757155-44757155 on Build GRCh38
rs148862121 Chr.19: 44756865-44756865 on Build GRCh38
rs774578129 Chr.19: 44757245-44757245 on Build GRCh38
rs950679779 Chr.19: 44757051-44757051 on Build GRCh38
rs557386805 Chr.19: 44756774-44756775 on Build GRCh38
rs911728864 Chr.19: 44756740-44756740 on Build GRCh38
rs1032530248 Chr.19: 44756956-44756956 on Build GRCh38
rs921717544 Chr.19: 44756818-44756818 on Build GRCh38
rs1045420444 Chr.19: 44757224-44757224 on Build GRCh38
rs749312414 Chr.19: 44757121-44757121 on Build GRCh38
rs11880948 Chr.19: 44757297-44757297 on Build GRCh38
rs149106894 Chr.19: 44757061-44757061 on Build GRCh38
rs2927456 Chr.19: 44756957-44756957 on Build GRCh38
rs745541576 Chr.19: 44757006-44757006 on Build GRCh38
rs868852688 Chr.19: 44757209-44757209 on Build GRCh38
rs201738573 Chr.19: 44756988-44756988 on Build GRCh38
rs146977654 Chr.19: 44756902-44756902 on Build GRCh38
rs1042411700 Chr.19: 44757211-44757211 on Build GRCh38
rs749993258 Chr.19: 44757283-44757283 on Build GRCh38
rs138060189 Chr.19: 44757028-44757028 on Build GRCh38
rs749890130 Chr.19: 44757183-44757183 on Build GRCh38

View Full Product Details