Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 123777596-123778142 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM91A1
Entrez Gene ID
157769
Gene Name
family with sequence similarity 91 member A1
Gene Aliases
-
UniGene
Hs.459174
Gene Chromosome Location
Chr.8: 123768439-123815452 on Build GRCh38
Transcripts
NM_001317917.1 NM_001317918.1 NM_144963.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6837313 c.374G>A p.R125K Chr.8: 123778031-123778031 on Build GRCh38
7367556 c.368-1G>T p.? Chr.8: 123778024-123778024 on Build GRCh38
6660334 c.368-10delT p.? Chr.8: 123778015-123778015 on Build GRCh38
4587759 c.375G>A p.R125R Chr.8: 123778032-123778032 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs920261156 Chr.8: 123777887-123777887 on Build GRCh38
rs375000683 Chr.8: 123777988-123777988 on Build GRCh38
rs755008041 Chr.8: 123777802-123777803 on Build GRCh38
rs567960935 Chr.8: 123777657-123777658 on Build GRCh38
rs571847773 Chr.8: 123777982-123777982 on Build GRCh38
rs542214273 Chr.8: 123777637-123777637 on Build GRCh38
rs962877754 Chr.8: 123777885-123777885 on Build GRCh38
rs1029658073 Chr.8: 123777852-123777852 on Build GRCh38
rs913544719 Chr.8: 123777941-123777941 on Build GRCh38
rs780164628 Chr.8: 123777996-123777996 on Build GRCh38
rs951623106 Chr.8: 123777864-123777864 on Build GRCh38
rs922825353 Chr.8: 123777959-123777959 on Build GRCh38
rs530706171 Chr.8: 123777790-123777790 on Build GRCh38
rs981344429 Chr.8: 123777886-123777886 on Build GRCh38
rs760075063 Chr.8: 123778078-123778078 on Build GRCh38
rs191234196 Chr.8: 123777958-123777958 on Build GRCh38
rs546971660 Chr.8: 123777866-123777866 on Build GRCh38
rs778115342 Chr.8: 123778109-123778109 on Build GRCh38
rs778921905 Chr.8: 123778019-123778019 on Build GRCh38
rs535385172 Chr.8: 123777786-123777786 on Build GRCh38
rs901161500 Chr.8: 123777648-123777648 on Build GRCh38
rs777838969 Chr.8: 123778049-123778049 on Build GRCh38
rs372630069 Chr.8: 123778032-123778032 on Build GRCh38
rs187332372 Chr.8: 123777808-123777808 on Build GRCh38
rs374275968 Chr.8: 123777927-123777927 on Build GRCh38
rs759750294 Chr.8: 123777961-123777961 on Build GRCh38
rs777098598 Chr.8: 123778067-123778067 on Build GRCh38
rs926357088 Chr.8: 123777661-123777661 on Build GRCh38
rs910054596 Chr.8: 123777870-123777870 on Build GRCh38
rs1056037802 Chr.8: 123777756-123777756 on Build GRCh38
rs544058968 Chr.8: 123777801-123777801 on Build GRCh38
rs998222381 Chr.8: 123777844-123777844 on Build GRCh38
rs904568249 Chr.8: 123777718-123777718 on Build GRCh38
rs747087431 Chr.8: 123778066-123778066 on Build GRCh38
rs932890743 Chr.8: 123777969-123777969 on Build GRCh38
rs939085246 Chr.8: 123777931-123777931 on Build GRCh38
rs939069302 Chr.8: 123777671-123777671 on Build GRCh38
rs200076157 Chr.8: 123778029-123778029 on Build GRCh38
rs78955281 Chr.8: 123777942-123777942 on Build GRCh38
rs754792334 Chr.8: 123778017-123778017 on Build GRCh38
rs1009408244 Chr.8: 123777779-123777779 on Build GRCh38
rs903759719 Chr.8: 123778003-123778003 on Build GRCh38
rs770138743 Chr.8: 123778103-123778103 on Build GRCh38
rs940199546 Chr.8: 123777950-123777950 on Build GRCh38
rs758795842 Chr.8: 123778014-123778015 on Build GRCh38
rs1044781673 Chr.8: 123777657-123777657 on Build GRCh38
rs773199556 Chr.8: 123777807-123777807 on Build GRCh38
rs533951215 Chr.8: 123777986-123777986 on Build GRCh38
rs964474561 Chr.8: 123777829-123777829 on Build GRCh38
rs903644490 Chr.8: 123777800-123777800 on Build GRCh38
rs1031496311 Chr.8: 123777806-123777806 on Build GRCh38
rs1028345745 Chr.8: 123777680-123777680 on Build GRCh38
rs753877792 Chr.8: 123777997-123777997 on Build GRCh38
rs10505441 Chr.8: 123777934-123777934 on Build GRCh38
rs892411771 Chr.8: 123777757-123777757 on Build GRCh38

View Full Product Details