Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15481977-15482540 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELA2B
Entrez Gene ID
51032
Gene Name
chymotrypsin like elastase family member 2B
Gene Aliases
ELA2B
UniGene
Hs.631871
Gene Chromosome Location
Chr.1: 15476101-15491400 on Build GRCh38
Transcripts
NM_015849.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3475926 c.280G>A p.E94K Chr.1: 15482317-15482317 on Build GRCh38
3475924 c.230C>T p.S77F Chr.1: 15482267-15482267 on Build GRCh38
4671400 c.355G>A p.V119I Chr.1: 15482335-15482335 on Build GRCh38
5900854 c.228-4C>T p.? Chr.1: 15482261-15482261 on Build GRCh38
4023629 c.323A>G p.N108S Chr.1: 15482303-15482303 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs553091551 Chr.1: 15482452-15482452 on Build GRCh38
rs756008525 Chr.1: 15482356-15482356 on Build GRCh38
rs570413115 Chr.1: 15482046-15482046 on Build GRCh38
rs920404854 Chr.1: 15482137-15482137 on Build GRCh38
rs568648004 Chr.1: 15482311-15482311 on Build GRCh38
rs377744220 Chr.1: 15482426-15482426 on Build GRCh38
rs781188736 Chr.1: 15482346-15482346 on Build GRCh38
rs747919248 Chr.1: 15482442-15482442 on Build GRCh38
rs753294464 Chr.1: 15482322-15482322 on Build GRCh38
rs746990321 Chr.1: 15482444-15482446 on Build GRCh38
rs758763592 Chr.1: 15482415-15482415 on Build GRCh38
rs199804944 Chr.1: 15482276-15482276 on Build GRCh38
rs770956773 Chr.1: 15482233-15482233 on Build GRCh38
rs780169361 Chr.1: 15482422-15482422 on Build GRCh38
rs566599564 Chr.1: 15482478-15482478 on Build GRCh38
rs774035469 Chr.1: 15482313-15482313 on Build GRCh38
rs3820071 Chr.1: 15482272-15482272 on Build GRCh38
rs543444102 Chr.1: 15482163-15482163 on Build GRCh38
rs539544426 Chr.1: 15482069-15482069 on Build GRCh38
rs573092791 Chr.1: 15482096-15482096 on Build GRCh38
rs767131277 Chr.1: 15482316-15482316 on Build GRCh38
rs555578936 Chr.1: 15482516-15482516 on Build GRCh38
rs559018886 Chr.1: 15482317-15482317 on Build GRCh38
rs752806310 Chr.1: 15482371-15482371 on Build GRCh38
rs772806551 Chr.1: 15482379-15482379 on Build GRCh38
rs1040368917 Chr.1: 15482177-15482177 on Build GRCh38
rs776050729 Chr.1: 15482392-15482392 on Build GRCh38
rs775219814 Chr.1: 15482293-15482293 on Build GRCh38
rs555353488 Chr.1: 15482140-15482140 on Build GRCh38
rs751797790 Chr.1: 15482335-15482335 on Build GRCh38
rs371863649 Chr.1: 15482302-15482302 on Build GRCh38
rs770614462 Chr.1: 15482385-15482385 on Build GRCh38
rs758925686 Chr.1: 15482281-15482281 on Build GRCh38
rs762333807 Chr.1: 15482323-15482323 on Build GRCh38
rs147273139 Chr.1: 15482502-15482502 on Build GRCh38
rs990655736 Chr.1: 15482124-15482124 on Build GRCh38
rs777949883 Chr.1: 15482403-15482404 on Build GRCh38
rs772345241 Chr.1: 15482219-15482219 on Build GRCh38
rs776526114 Chr.1: 15482234-15482234 on Build GRCh38
rs138336343 Chr.1: 15482250-15482250 on Build GRCh38
rs761431469 Chr.1: 15482314-15482314 on Build GRCh38
rs767760364 Chr.1: 15482410-15482410 on Build GRCh38
rs113521521 Chr.1: 15482204-15482204 on Build GRCh38
rs4661635 Chr.1: 15482207-15482207 on Build GRCh38
rs761844523 Chr.1: 15482130-15482130 on Build GRCh38
rs779184022 Chr.1: 15482347-15482348 on Build GRCh38
rs754383761 Chr.1: 15482273-15482273 on Build GRCh38
rs775532639 Chr.1: 15482463-15482463 on Build GRCh38
rs531244107 Chr.1: 15482353-15482353 on Build GRCh38
rs762538602 Chr.1: 15482256-15482256 on Build GRCh38
rs765874325 Chr.1: 15482320-15482320 on Build GRCh38
rs745736468 Chr.1: 15482292-15482292 on Build GRCh38
rs921931244 Chr.1: 15482507-15482507 on Build GRCh38
rs981782314 Chr.1: 15482063-15482063 on Build GRCh38
rs753809532 Chr.1: 15482424-15482424 on Build GRCh38
rs756168301 Chr.1: 15482416-15482416 on Build GRCh38
rs747243084 Chr.1: 15482284-15482284 on Build GRCh38
rs370955886 Chr.1: 15482485-15482485 on Build GRCh38
rs749236031 Chr.1: 15482360-15482360 on Build GRCh38
rs760229577 Chr.1: 15482380-15482380 on Build GRCh38
rs764423042 Chr.1: 15482334-15482334 on Build GRCh38
rs544100922 Chr.1: 15482367-15482367 on Build GRCh38
rs368752949 Chr.1: 15482262-15482262 on Build GRCh38
rs35206351 Chr.1: 15482012-15482013 on Build GRCh38
rs758579635 Chr.1: 15482401-15482402 on Build GRCh38
rs150116668 Chr.1: 15482277-15482277 on Build GRCh38
rs553179175 Chr.1: 15482095-15482095 on Build GRCh38
rs765570455 Chr.1: 15482361-15482361 on Build GRCh38
rs200537312 Chr.1: 15482310-15482310 on Build GRCh38
rs907987154 Chr.1: 15482474-15482474 on Build GRCh38
rs778713017 Chr.1: 15482439-15482439 on Build GRCh38
rs959310383 Chr.1: 15482119-15482122 on Build GRCh38
rs944746917 Chr.1: 15482175-15482175 on Build GRCh38
rs766015394 Chr.1: 15482100-15482100 on Build GRCh38
rs575542421 Chr.1: 15482159-15482159 on Build GRCh38
rs751880541 Chr.1: 15482404-15482404 on Build GRCh38
rs3766160 Chr.1: 15482377-15482377 on Build GRCh38
rs773698946 Chr.1: 15482270-15482270 on Build GRCh38
rs762043886 Chr.1: 15482409-15482409 on Build GRCh38
rs781312258 Chr.1: 15482290-15482290 on Build GRCh38
rs35220329 Chr.1: 15482471-15482472 on Build GRCh38
rs747142175 Chr.1: 15482231-15482231 on Build GRCh38
rs774081200 Chr.1: 15482368-15482368 on Build GRCh38
rs779939071 Chr.1: 15482357-15482357 on Build GRCh38
rs761359259 Chr.1: 15482271-15482271 on Build GRCh38
rs748378137 Chr.1: 15482218-15482218 on Build GRCh38
rs773906672 Chr.1: 15482196-15482196 on Build GRCh38
rs143911244 Chr.1: 15482448-15482448 on Build GRCh38
rs200845243 Chr.1: 15482011-15482012 on Build GRCh38
rs764448810 Chr.1: 15482398-15482398 on Build GRCh38
rs757343753 Chr.1: 15482285-15482285 on Build GRCh38
rs760064005 Chr.1: 15482319-15482319 on Build GRCh38
rs532898958 Chr.1: 15482393-15482393 on Build GRCh38
rs966807591 Chr.1: 15482056-15482056 on Build GRCh38
rs542080873 Chr.1: 15482115-15482115 on Build GRCh38
rs370469816 Chr.1: 15482220-15482220 on Build GRCh38
rs199633397 Chr.1: 15482428-15482428 on Build GRCh38
rs747771040 Chr.1: 15482376-15482376 on Build GRCh38
rs951885007 Chr.1: 15482413-15482413 on Build GRCh38
rs753132870 Chr.1: 15482282-15482282 on Build GRCh38
rs202068600 Chr.1: 15482015-15482016 on Build GRCh38
rs758507409 Chr.1: 15482283-15482283 on Build GRCh38
rs769568982 Chr.1: 15482236-15482236 on Build GRCh38
rs757421140 Chr.1: 15482343-15482343 on Build GRCh38
rs930727944 Chr.1: 15482143-15482143 on Build GRCh38
rs780413983 Chr.1: 15482499-15482499 on Build GRCh38
rs368356642 Chr.1: 15482294-15482294 on Build GRCh38

View Full Product Details