Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11239481-11240077 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPTL8
Entrez Gene ID
55908
Gene Name
angiopoietin like 8
Gene Aliases
C19orf80, PRO1185, PVPA599, RIFL, TD26
UniGene
Hs.534467
Gene Chromosome Location
Chr.19: 11239619-11241943 on Build GRCh38
Transcripts
NM_018687.6 NM_020812.3 XM_005260000.2 XM_005260001.2 XM_011528150.1 XM_011528151.1 XM_011528152.1

Gene Symbol
DOCK6
Entrez Gene ID
57572
Gene Name
dedicator of cytokinesis 6
Gene Aliases
AOS2, ZIR1
UniGene
Hs.591002
Gene Chromosome Location
Chr.19: 11199293-11262502 on Build GRCh38
Transcripts
NM_018687.6 NM_020812.3 XM_005260000.2 XM_005260001.2 XM_011528150.1 XM_011528151.1 XM_011528152.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
990845 c.30G>T p.W10C Chr.19: 11239667-11239667 on Build GRCh38
4529649 c.162G>A p.R54R Chr.19: 11239799-11239799 on Build GRCh38
1611534 c.40A>C p.M14L Chr.19: 11239677-11239677 on Build GRCh38
1303984 c.59C>T p.S20L Chr.19: 11239696-11239696 on Build GRCh38
4534859 c.212G>A p.G71D Chr.19: 11239849-11239849 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367883984 Chr.19: 11239505-11239505 on Build GRCh38
rs759780922 Chr.19: 11239595-11239595 on Build GRCh38
rs536232063 Chr.19: 11239890-11239890 on Build GRCh38
rs920613212 Chr.19: 11240029-11240029 on Build GRCh38
rs778187151 Chr.19: 11239686-11239686 on Build GRCh38
rs765895517 Chr.19: 11239867-11239867 on Build GRCh38
rs371530336 Chr.19: 11239805-11239805 on Build GRCh38
rs767657753 Chr.19: 11239605-11239605 on Build GRCh38
rs368237348 Chr.19: 11239961-11239961 on Build GRCh38
rs968623484 Chr.19: 11239696-11239696 on Build GRCh38
rs201199003 Chr.19: 11239713-11239713 on Build GRCh38
rs767264573 Chr.19: 11239971-11239971 on Build GRCh38
rs748522487 Chr.19: 11239679-11239679 on Build GRCh38
rs368144125 Chr.19: 11239925-11239925 on Build GRCh38
rs912202472 Chr.19: 11239649-11239649 on Build GRCh38
rs375214158 Chr.19: 11239952-11239952 on Build GRCh38
rs756500046 Chr.19: 11239684-11239684 on Build GRCh38
rs987310947 Chr.19: 11239714-11239714 on Build GRCh38
rs749820131 Chr.19: 11239687-11239687 on Build GRCh38
rs200010590 Chr.19: 11239670-11239670 on Build GRCh38
rs759226211 Chr.19: 11239871-11239871 on Build GRCh38
rs768721053 Chr.19: 11239810-11239810 on Build GRCh38
rs906665047 Chr.19: 11239558-11239558 on Build GRCh38
rs377021496 Chr.19: 11239853-11239853 on Build GRCh38
rs910621593 Chr.19: 11239506-11239506 on Build GRCh38
rs776430454 Chr.19: 11239579-11239579 on Build GRCh38
rs781259374 Chr.19: 11239943-11239943 on Build GRCh38
rs752084449 Chr.19: 11239852-11239852 on Build GRCh38
rs779796333 Chr.19: 11239672-11239672 on Build GRCh38
rs906765465 Chr.19: 11239855-11239855 on Build GRCh38
rs1028528511 Chr.19: 11239590-11239590 on Build GRCh38
rs778228300 Chr.19: 11239787-11239787 on Build GRCh38
rs576699306 Chr.19: 11239568-11239568 on Build GRCh38
rs761873450 Chr.19: 11239716-11239716 on Build GRCh38
rs772107121 Chr.19: 11239798-11239798 on Build GRCh38
rs1032037025 Chr.19: 11239934-11239934 on Build GRCh38
rs973431605 Chr.19: 11240013-11240013 on Build GRCh38
rs1541922 Chr.19: 11239757-11239757 on Build GRCh38
rs2278426 Chr.19: 11239812-11239812 on Build GRCh38
rs761644691 Chr.19: 11239847-11239847 on Build GRCh38
rs549677886 Chr.19: 11239763-11239763 on Build GRCh38
rs771383574 Chr.19: 11239691-11239691 on Build GRCh38
rs929574862 Chr.19: 11239841-11239841 on Build GRCh38
rs530487180 Chr.19: 11239975-11239975 on Build GRCh38
rs1541924 Chr.19: 11239513-11239513 on Build GRCh38
rs746415379 Chr.19: 11239644-11239654 on Build GRCh38
rs532252931 Chr.19: 11239804-11239804 on Build GRCh38
rs762507505 Chr.19: 11239786-11239786 on Build GRCh38
rs770398169 Chr.19: 11239702-11239704 on Build GRCh38
rs747694751 Chr.19: 11239889-11239889 on Build GRCh38
rs779376663 Chr.19: 11239789-11239789 on Build GRCh38
rs865986153 Chr.19: 11239933-11239933 on Build GRCh38
rs946067343 Chr.19: 11239998-11239998 on Build GRCh38
rs746995630 Chr.19: 11239935-11239935 on Build GRCh38
rs1010470904 Chr.19: 11239514-11239514 on Build GRCh38
rs543622267 Chr.19: 11239583-11239583 on Build GRCh38
rs899464934 Chr.19: 11239538-11239538 on Build GRCh38
rs376063419 Chr.19: 11239712-11239712 on Build GRCh38
rs1037661138 Chr.19: 11239533-11239533 on Build GRCh38
rs879709062 Chr.19: 11239546-11239546 on Build GRCh38
rs750989745 Chr.19: 11239869-11239869 on Build GRCh38
rs765200793 Chr.19: 11239731-11239731 on Build GRCh38
rs778603181 Chr.19: 11239861-11239861 on Build GRCh38
rs570216272 Chr.19: 11239968-11239968 on Build GRCh38
rs757612643 Chr.19: 11239886-11239886 on Build GRCh38
rs376405688 Chr.19: 11239891-11239891 on Build GRCh38
rs938750315 Chr.19: 11239723-11239723 on Build GRCh38
rs1057171644 Chr.19: 11239728-11239728 on Build GRCh38
rs372882086 Chr.19: 11239700-11239700 on Build GRCh38
rs904782865 Chr.19: 11240050-11240050 on Build GRCh38
rs541132639 Chr.19: 11239630-11239630 on Build GRCh38
rs756488531 Chr.19: 11239784-11239784 on Build GRCh38
rs368909299 Chr.19: 11239699-11239699 on Build GRCh38
rs372853879 Chr.19: 11239967-11239967 on Build GRCh38
rs773758808 Chr.19: 11239577-11239577 on Build GRCh38
rs774416873 Chr.19: 11239697-11239697 on Build GRCh38
rs768937257 Chr.19: 11239937-11239937 on Build GRCh38
rs764003138 Chr.19: 11239621-11239621 on Build GRCh38
rs1029130397 Chr.19: 11239839-11239839 on Build GRCh38
rs757566619 Chr.19: 11239576-11239576 on Build GRCh38
rs1021869217 Chr.19: 11239545-11239545 on Build GRCh38
rs147405465 Chr.19: 11239858-11239858 on Build GRCh38
rs913309382 Chr.19: 11239985-11239985 on Build GRCh38
rs904159174 Chr.19: 11239918-11239918 on Build GRCh38
rs752312893 Chr.19: 11239973-11239973 on Build GRCh38
rs554624483 Chr.19: 11239951-11239951 on Build GRCh38
rs372031853 Chr.19: 11239944-11239944 on Build GRCh38
rs200458610 Chr.19: 11239669-11239669 on Build GRCh38
rs751197941 Chr.19: 11239769-11239769 on Build GRCh38
rs74810158 Chr.19: 11239884-11239884 on Build GRCh38
rs142800818 Chr.19: 11239788-11239788 on Build GRCh38
rs577224723 Chr.19: 11239629-11239629 on Build GRCh38
rs866315955 Chr.19: 11239834-11239834 on Build GRCh38
rs964774097 Chr.19: 11239987-11239987 on Build GRCh38
rs775067650 Chr.19: 11239772-11239772 on Build GRCh38
rs770219255 Chr.19: 11239576-11239576 on Build GRCh38
rs754255586 Chr.19: 11239885-11239885 on Build GRCh38
rs1002643598 Chr.19: 11239564-11239564 on Build GRCh38
rs892066 Chr.19: 11239664-11239664 on Build GRCh38
rs746013489 Chr.19: 11239792-11239792 on Build GRCh38
rs760426491 Chr.19: 11239707-11239707 on Build GRCh38
rs555285747 Chr.19: 11239547-11239547 on Build GRCh38
rs758952234 Chr.19: 11239969-11239969 on Build GRCh38
rs537756153 Chr.19: 11240026-11240026 on Build GRCh38
rs1541923 Chr.19: 11239618-11239618 on Build GRCh38
rs781616294 Chr.19: 11239585-11239593 on Build GRCh38
rs879209915 Chr.19: 11239851-11239851 on Build GRCh38
rs59168178 Chr.19: 11239650-11239650 on Build GRCh38
rs773334866 Chr.19: 11239954-11239954 on Build GRCh38
rs372492452 Chr.19: 11239613-11239613 on Build GRCh38
rs749968127 Chr.19: 11239745-11239745 on Build GRCh38

View Full Product Details