Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 123008621-123009217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACADSB
Entrez Gene ID
36
Gene Name
acyl-CoA dehydrogenase, short/branched chain
Gene Aliases
2-MEBCAD, ACAD7, SBCAD
UniGene
Hs.81934
Gene Chromosome Location
Chr.10: 123008913-123058290 on Build GRCh38
Transcripts
NM_001609.3 XM_017016194.1 NM_001271840.1 NR_073475.1 NR_073476.1 NR_073477.1 NR_073478.1 NR_073479.1 XM_006717947.3 XM_017016550.1 XM_017016551.1 XM_017016552.1 XM_017016553.1 XM_017016554.1 XM_017016555.1

Gene Symbol
IKZF5
Entrez Gene ID
64376
Gene Name
IKAROS family zinc finger 5
Gene Aliases
PEGASUS, ZNFN1A5
UniGene
Hs.501289
Gene Chromosome Location
Chr.10: 122990806-123008850 on Build GRCh38
Transcripts
NM_001609.3 XM_017016194.1 NM_001271840.1 NR_073475.1 NR_073476.1 NR_073477.1 NR_073478.1 NR_073479.1 XM_006717947.3 XM_017016550.1 XM_017016551.1 XM_017016552.1 XM_017016553.1 XM_017016554.1 XM_017016555.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5635306 c.10C>T p.L4L Chr.10: 123009039-123009039 on Build GRCh38
6222106 c.14C>T p.A5V Chr.10: 123009043-123009043 on Build GRCh38
427201 c.38G>A p.R13K Chr.10: 123009067-123009067 on Build GRCh38
6374737 c.34A>G p.S12G Chr.10: 123009063-123009063 on Build GRCh38
7199954 c.9C>T p.G3G Chr.10: 123009038-123009038 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs947896053 Chr.10: 123008772-123008772 on Build GRCh38
rs1042184444 Chr.10: 123008741-123008742 on Build GRCh38
rs571674926 Chr.10: 123009137-123009137 on Build GRCh38
rs1056299687 Chr.10: 123009000-123009000 on Build GRCh38
rs547039642 Chr.10: 123008830-123008830 on Build GRCh38
rs1025901745 Chr.10: 123008651-123008655 on Build GRCh38
rs376265483 Chr.10: 123009043-123009043 on Build GRCh38
rs1018536342 Chr.10: 123009083-123009083 on Build GRCh38
rs542929323 Chr.10: 123009080-123009080 on Build GRCh38
rs2277250 Chr.10: 123008776-123008776 on Build GRCh38
rs960524222 Chr.10: 123009163-123009163 on Build GRCh38
rs202143221 Chr.10: 123009103-123009103 on Build GRCh38
rs761504522 Chr.10: 123009121-123009121 on Build GRCh38
rs532491098 Chr.10: 123009192-123009192 on Build GRCh38
rs190149746 Chr.10: 123008923-123008923 on Build GRCh38
rs751100267 Chr.10: 123008995-123008995 on Build GRCh38
rs958040966 Chr.10: 123008723-123008723 on Build GRCh38
rs546197770 Chr.10: 123009015-123009015 on Build GRCh38
rs879640273 Chr.10: 123008927-123008927 on Build GRCh38
rs201117089 Chr.10: 123008921-123008922 on Build GRCh38
rs1025788754 Chr.10: 123008862-123008862 on Build GRCh38
rs916399128 Chr.10: 123008751-123008751 on Build GRCh38
rs541906572 Chr.10: 123009028-123009028 on Build GRCh38
rs991912307 Chr.10: 123009171-123009171 on Build GRCh38
rs999172391 Chr.10: 123008925-123008925 on Build GRCh38
rs969206708 Chr.10: 123008968-123008968 on Build GRCh38
rs150850221 Chr.10: 123008943-123008943 on Build GRCh38
rs886046774 Chr.10: 123008928-123008929 on Build GRCh38
rs759070889 Chr.10: 123008997-123009004 on Build GRCh38
rs117921026 Chr.10: 123009098-123009098 on Build GRCh38
rs374517787 Chr.10: 123008780-123008780 on Build GRCh38
rs767081552 Chr.10: 123009122-123009122 on Build GRCh38
rs886046775 Chr.10: 123008928-123008928 on Build GRCh38
rs181857440 Chr.10: 123009118-123009118 on Build GRCh38
rs961935221 Chr.10: 123009185-123009185 on Build GRCh38
rs927909728 Chr.10: 123008810-123008810 on Build GRCh38
rs557282183 Chr.10: 123008879-123008879 on Build GRCh38
rs546681688 Chr.10: 123009119-123009119 on Build GRCh38
rs754696589 Chr.10: 123009085-123009085 on Build GRCh38
rs368228443 Chr.10: 123009089-123009089 on Build GRCh38
rs978164233 Chr.10: 123008681-123008681 on Build GRCh38
rs563472823 Chr.10: 123009008-123009008 on Build GRCh38
rs901402559 Chr.10: 123009068-123009068 on Build GRCh38
rs773839189 Chr.10: 123008651-123008674 on Build GRCh38
rs1025841198 Chr.10: 123009056-123009056 on Build GRCh38
rs1019859492 Chr.10: 123008676-123008676 on Build GRCh38
rs1010615110 Chr.10: 123009101-123009101 on Build GRCh38
rs915114465 Chr.10: 123008695-123008695 on Build GRCh38
rs141443308 Chr.10: 123008941-123008941 on Build GRCh38
rs965072894 Chr.10: 123008677-123008677 on Build GRCh38
rs940615681 Chr.10: 123008996-123008996 on Build GRCh38
rs772331528 Chr.10: 123009044-123009044 on Build GRCh38
rs148327034 Chr.10: 123008933-123008933 on Build GRCh38
rs995533084 Chr.10: 123008837-123008837 on Build GRCh38
rs577664948 Chr.10: 123008950-123008950 on Build GRCh38
rs371211749 Chr.10: 123008997-123008997 on Build GRCh38
rs12263012 Chr.10: 123009067-123009067 on Build GRCh38
rs749559827 Chr.10: 123009022-123009022 on Build GRCh38
rs1017430106 Chr.10: 123008815-123008815 on Build GRCh38
rs768384949 Chr.10: 123009102-123009102 on Build GRCh38
rs1031280322 Chr.10: 123009092-123009092 on Build GRCh38
rs766888009 Chr.10: 123009026-123009026 on Build GRCh38
rs957749339 Chr.10: 123008928-123008928 on Build GRCh38
rs571672612 Chr.10: 123008848-123008848 on Build GRCh38
rs927866589 Chr.10: 123008986-123008986 on Build GRCh38
rs898538271 Chr.10: 123008821-123008821 on Build GRCh38
rs1048492401 Chr.10: 123008842-123008842 on Build GRCh38
rs996253516 Chr.10: 123009086-123009086 on Build GRCh38
rs554868976 Chr.10: 123008929-123008929 on Build GRCh38
rs750986257 Chr.10: 123009011-123009011 on Build GRCh38
rs954237824 Chr.10: 123008651-123008651 on Build GRCh38
rs923807005 Chr.10: 123009143-123009143 on Build GRCh38
rs745863047 Chr.10: 123009071-123009071 on Build GRCh38
rs778965920 Chr.10: 123009037-123009037 on Build GRCh38
rs994239544 Chr.10: 123009040-123009040 on Build GRCh38
rs938655682 Chr.10: 123008811-123008811 on Build GRCh38
rs923680280 Chr.10: 123008709-123008709 on Build GRCh38
rs916490844 Chr.10: 123009172-123009172 on Build GRCh38
rs902449078 Chr.10: 123008742-123008747 on Build GRCh38
rs539177953 Chr.10: 123008872-123008872 on Build GRCh38
rs1012364384 Chr.10: 123008789-123008789 on Build GRCh38
rs1037446566 Chr.10: 123008827-123008827 on Build GRCh38
rs939245269 Chr.10: 123008761-123008761 on Build GRCh38
rs974884993 Chr.10: 123008688-123008688 on Build GRCh38
rs567645898 Chr.10: 123008662-123008662 on Build GRCh38
rs911536378 Chr.10: 123008672-123008672 on Build GRCh38
rs989290411 Chr.10: 123008739-123008739 on Build GRCh38
rs1007054497 Chr.10: 123008873-123008873 on Build GRCh38
rs886046776 Chr.10: 123008991-123008991 on Build GRCh38
rs895185970 Chr.10: 123009002-123009002 on Build GRCh38
rs918571828 Chr.10: 123009003-123009003 on Build GRCh38
rs752252664 Chr.10: 123009179-123009179 on Build GRCh38
rs981909827 Chr.10: 123009164-123009164 on Build GRCh38
rs981942353 Chr.10: 123008976-123008976 on Build GRCh38
rs72839755 Chr.10: 123008907-123008907 on Build GRCh38
rs939297382 Chr.10: 123008998-123008998 on Build GRCh38
rs955572170 Chr.10: 123008652-123008652 on Build GRCh38

View Full Product Details