Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 47368289-47368651 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMPK1
Entrez Gene ID
51727
Gene Name
cytidine/uridine monophosphate kinase 1
Gene Aliases
CK, CMK, CMPK, UMK, UMP-CMPK, UMPK
UniGene
Hs.731647
Gene Chromosome Location
Chr.1: 47333797-47378839 on Build GRCh38
Transcripts
NM_001136140.1 NM_016308.2 NR_046394.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
146504 c.240G>T p.Q80H Chr.1: 47368537-47368537 on Build GRCh38
7410771 c.242A>G p.Y81C Chr.1: 47368539-47368539 on Build GRCh38
910428 c.212G>C p.R71P Chr.1: 47368509-47368509 on Build GRCh38
4938027 c.316A>G p.R106G Chr.1: 47368613-47368613 on Build GRCh38
1201482 c.183C>A p.Y61* Chr.1: 47368480-47368480 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs926543038 Chr.1: 47368401-47368401 on Build GRCh38
rs555761559 Chr.1: 47368498-47368498 on Build GRCh38
rs369595846 Chr.1: 47368452-47368452 on Build GRCh38
rs764910329 Chr.1: 47368617-47368617 on Build GRCh38
rs141304390 Chr.1: 47368520-47368520 on Build GRCh38
rs745886225 Chr.1: 47368508-47368508 on Build GRCh38
rs950361502 Chr.1: 47368411-47368411 on Build GRCh38
rs80046781 Chr.1: 47368493-47368493 on Build GRCh38
rs12079592 Chr.1: 47368341-47368341 on Build GRCh38
rs768881199 Chr.1: 47368533-47368533 on Build GRCh38
rs939364249 Chr.1: 47368401-47368402 on Build GRCh38
rs759663153 Chr.1: 47368424-47368424 on Build GRCh38
rs201653602 Chr.1: 47368535-47368535 on Build GRCh38
rs200584751 Chr.1: 47368480-47368480 on Build GRCh38
rs72553945 Chr.1: 47368466-47368466 on Build GRCh38
rs374560186 Chr.1: 47368549-47368549 on Build GRCh38
rs143595256 Chr.1: 47368550-47368550 on Build GRCh38
rs981115877 Chr.1: 47368375-47368375 on Build GRCh38
rs762169930 Chr.1: 47368619-47368620 on Build GRCh38
rs35687416 Chr.1: 47368537-47368537 on Build GRCh38
rs925677563 Chr.1: 47368330-47368330 on Build GRCh38
rs201488707 Chr.1: 47368521-47368521 on Build GRCh38
rs75846696 Chr.1: 47368494-47368494 on Build GRCh38
rs540094424 Chr.1: 47368364-47368364 on Build GRCh38
rs558486406 Chr.1: 47368434-47368434 on Build GRCh38
rs764778220 Chr.1: 47368474-47368474 on Build GRCh38
rs777766014 Chr.1: 47368482-47368482 on Build GRCh38
rs377653530 Chr.1: 47368483-47368483 on Build GRCh38
rs887231423 Chr.1: 47368419-47368419 on Build GRCh38
rs765520132 Chr.1: 47368621-47368622 on Build GRCh38
rs190292865 Chr.1: 47368435-47368435 on Build GRCh38
rs370864008 Chr.1: 47368525-47368525 on Build GRCh38
rs780641351 Chr.1: 47368432-47368432 on Build GRCh38
rs758940039 Chr.1: 47368600-47368600 on Build GRCh38
rs752049181 Chr.1: 47368445-47368445 on Build GRCh38
rs1053174257 Chr.1: 47368344-47368344 on Build GRCh38
rs753247958 Chr.1: 47368459-47368459 on Build GRCh38
rs769965982 Chr.1: 47368509-47368509 on Build GRCh38
rs768074260 Chr.1: 47368456-47368456 on Build GRCh38
rs932292400 Chr.1: 47368317-47368317 on Build GRCh38
rs1008556485 Chr.1: 47368562-47368562 on Build GRCh38
rs144452656 Chr.1: 47368517-47368517 on Build GRCh38
rs752380854 Chr.1: 47368625-47368625 on Build GRCh38
rs767864189 Chr.1: 47368547-47368547 on Build GRCh38
rs200895100 Chr.1: 47368526-47368526 on Build GRCh38
rs757070749 Chr.1: 47368492-47368492 on Build GRCh38
rs987810047 Chr.1: 47368337-47368337 on Build GRCh38
rs978513138 Chr.1: 47368318-47368318 on Build GRCh38
rs956452878 Chr.1: 47368331-47368337 on Build GRCh38
rs764480752 Chr.1: 47368442-47368442 on Build GRCh38

View Full Product Details