Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43361855-43362437 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC20
Entrez Gene ID
991
Gene Name
cell division cycle 20
Gene Aliases
CDC20A, bA276H19.3, p55CDC
UniGene
Hs.524947
Gene Chromosome Location
Chr.1: 43358955-43363203 on Build GRCh38
Transcripts
NM_001255.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
909760 c.1276G>T p.V426F Chr.1: 43362267-43362267 on Build GRCh38
6624101 c.1256G>T p.G419V Chr.1: 43362247-43362247 on Build GRCh38
119918 c.1301C>A p.A434D Chr.1: 43362292-43362292 on Build GRCh38
6360448 c.1253A>T p.H418L Chr.1: 43362244-43362244 on Build GRCh38
4820650 c.1237G>C p.E413Q Chr.1: 43362228-43362228 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769816067 Chr.1: 43362033-43362033 on Build GRCh38
rs199735457 Chr.1: 43362191-43362191 on Build GRCh38
rs778954437 Chr.1: 43362341-43362341 on Build GRCh38
rs745411506 Chr.1: 43362343-43362343 on Build GRCh38
rs139019074 Chr.1: 43362254-43362254 on Build GRCh38
rs373228573 Chr.1: 43362334-43362334 on Build GRCh38
rs968331457 Chr.1: 43362051-43362051 on Build GRCh38
rs751582244 Chr.1: 43362172-43362172 on Build GRCh38
rs776328979 Chr.1: 43362182-43362186 on Build GRCh38
rs185755685 Chr.1: 43362176-43362176 on Build GRCh38
rs777632153 Chr.1: 43362192-43362192 on Build GRCh38
rs377717351 Chr.1: 43362340-43362340 on Build GRCh38
rs181063440 Chr.1: 43361952-43361952 on Build GRCh38
rs932025480 Chr.1: 43362003-43362003 on Build GRCh38
rs752953958 Chr.1: 43362317-43362317 on Build GRCh38
rs781204017 Chr.1: 43361926-43361926 on Build GRCh38
rs917017587 Chr.1: 43362129-43362129 on Build GRCh38
rs755023413 Chr.1: 43362313-43362313 on Build GRCh38
rs924209657 Chr.1: 43362086-43362086 on Build GRCh38
rs925238494 Chr.1: 43362052-43362052 on Build GRCh38
rs773384058 Chr.1: 43362255-43362255 on Build GRCh38
rs766822161 Chr.1: 43362285-43362285 on Build GRCh38
rs1044842113 Chr.1: 43362123-43362123 on Build GRCh38
rs1044034588 Chr.1: 43362025-43362025 on Build GRCh38
rs878991751 Chr.1: 43362094-43362094 on Build GRCh38
rs769025243 Chr.1: 43362371-43362371 on Build GRCh38
rs758476676 Chr.1: 43362155-43362155 on Build GRCh38
rs541721235 Chr.1: 43361916-43361916 on Build GRCh38
rs770703760 Chr.1: 43362218-43362218 on Build GRCh38
rs747924777 Chr.1: 43362356-43362356 on Build GRCh38
rs771685450 Chr.1: 43362350-43362350 on Build GRCh38
rs781357124 Chr.1: 43362179-43362179 on Build GRCh38
rs1048140729 Chr.1: 43362184-43362184 on Build GRCh38
rs545900189 Chr.1: 43362130-43362130 on Build GRCh38
rs201154230 Chr.1: 43362253-43362253 on Build GRCh38
rs746679971 Chr.1: 43362353-43362353 on Build GRCh38
rs747980505 Chr.1: 43362187-43362187 on Build GRCh38
rs45443196 Chr.1: 43362195-43362195 on Build GRCh38
rs570640580 Chr.1: 43362416-43362416 on Build GRCh38
rs983626579 Chr.1: 43361911-43361911 on Build GRCh38
rs776415600 Chr.1: 43362206-43362206 on Build GRCh38
rs2069387 Chr.1: 43362409-43362409 on Build GRCh38
rs765701911 Chr.1: 43362145-43362145 on Build GRCh38
rs532070224 Chr.1: 43362104-43362104 on Build GRCh38
rs1019067950 Chr.1: 43361978-43361978 on Build GRCh38
rs773648579 Chr.1: 43362361-43362361 on Build GRCh38
rs763187931 Chr.1: 43362363-43362363 on Build GRCh38
rs755926042 Chr.1: 43362331-43362331 on Build GRCh38
rs1027946607 Chr.1: 43361888-43361888 on Build GRCh38
rs995126810 Chr.1: 43361927-43361927 on Build GRCh38
rs749411658 Chr.1: 43362315-43362315 on Build GRCh38
rs745320429 Chr.1: 43362230-43362230 on Build GRCh38
rs1027873058 Chr.1: 43362148-43362148 on Build GRCh38
rs952181442 Chr.1: 43361900-43361900 on Build GRCh38
rs762132307 Chr.1: 43362250-43362250 on Build GRCh38
rs1013768011 Chr.1: 43362114-43362114 on Build GRCh38
rs11555250 Chr.1: 43362278-43362278 on Build GRCh38
rs775251958 Chr.1: 43362236-43362236 on Build GRCh38
rs1022276463 Chr.1: 43362034-43362034 on Build GRCh38
rs544075088 Chr.1: 43362083-43362083 on Build GRCh38
rs917729667 Chr.1: 43362369-43362369 on Build GRCh38
rs775340138 Chr.1: 43362352-43362352 on Build GRCh38
rs771468303 Chr.1: 43362231-43362231 on Build GRCh38
rs996198280 Chr.1: 43362012-43362012 on Build GRCh38
rs886309136 Chr.1: 43361938-43361938 on Build GRCh38
rs35668070 Chr.1: 43361995-43361996 on Build GRCh38
rs995217113 Chr.1: 43362135-43362135 on Build GRCh38
rs113743211 Chr.1: 43361908-43361909 on Build GRCh38
rs145786996 Chr.1: 43361996-43361996 on Build GRCh38
rs372058967 Chr.1: 43362162-43362162 on Build GRCh38
rs768138175 Chr.1: 43362357-43362357 on Build GRCh38
rs751670358 Chr.1: 43362309-43362309 on Build GRCh38
rs138353916 Chr.1: 43362093-43362093 on Build GRCh38
rs141880288 Chr.1: 43362335-43362335 on Build GRCh38
rs774006843 Chr.1: 43362228-43362228 on Build GRCh38
rs1047557487 Chr.1: 43361930-43361930 on Build GRCh38

View Full Product Details