Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 55870401-55870962 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFEMP1
Entrez Gene ID
2202
Gene Name
EGF containing fibulin like extracellular matrix protein 1
Gene Aliases
DHRD, DRAD, FBLN3, FBNL, FIBL-3, MLVT, MTLV, S1-5
UniGene
Hs.732348
Gene Chromosome Location
Chr.2: 55865962-55924163 on Build GRCh38
Transcripts
NM_001039348.2 NM_001039349.2 XM_005264205.4 XM_017003586.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1021905 c.1136G>A p.C379Y Chr.2: 55870904-55870904 on Build GRCh38
3582356 c.1203A>T p.R401R Chr.2: 55870837-55870837 on Build GRCh38
6232035 c.1289C>G p.S430C Chr.2: 55870751-55870751 on Build GRCh38
3582355 c.1205C>T p.S402F Chr.2: 55870835-55870835 on Build GRCh38
3582354 c.1265C>T p.T422I Chr.2: 55870775-55870775 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142194873 Chr.2: 55870788-55870788 on Build GRCh38
rs192404906 Chr.2: 55870929-55870929 on Build GRCh38
rs534909303 Chr.2: 55870744-55870744 on Build GRCh38
rs146446706 Chr.2: 55870880-55870880 on Build GRCh38
rs775080680 Chr.2: 55870838-55870838 on Build GRCh38
rs745343709 Chr.2: 55870933-55870933 on Build GRCh38
rs373990093 Chr.2: 55870721-55870721 on Build GRCh38
rs766866113 Chr.2: 55870882-55870882 on Build GRCh38
rs1017931305 Chr.2: 55870602-55870602 on Build GRCh38
rs11682872 Chr.2: 55870576-55870576 on Build GRCh38
rs780292007 Chr.2: 55870708-55870708 on Build GRCh38
rs984037164 Chr.2: 55870529-55870529 on Build GRCh38
rs780590305 Chr.2: 55870931-55870931 on Build GRCh38
rs1012296244 Chr.2: 55870848-55870848 on Build GRCh38
rs199564083 Chr.2: 55870696-55870696 on Build GRCh38
rs972954826 Chr.2: 55870490-55870490 on Build GRCh38
rs200624494 Chr.2: 55870692-55870692 on Build GRCh38
rs780733918 Chr.2: 55870643-55870644 on Build GRCh38
rs941338554 Chr.2: 55870458-55870458 on Build GRCh38
rs1045913768 Chr.2: 55870528-55870528 on Build GRCh38
rs903325840 Chr.2: 55870562-55870562 on Build GRCh38
rs569670545 Chr.2: 55870619-55870619 on Build GRCh38
rs777439665 Chr.2: 55870720-55870720 on Build GRCh38
rs768478335 Chr.2: 55870646-55870647 on Build GRCh38
rs1054678579 Chr.2: 55870580-55870580 on Build GRCh38
rs750963516 Chr.2: 55870700-55870700 on Build GRCh38
rs560854074 Chr.2: 55870506-55870506 on Build GRCh38
rs901903477 Chr.2: 55870644-55870652 on Build GRCh38
rs1010527106 Chr.2: 55870601-55870601 on Build GRCh38
rs372003038 Chr.2: 55870691-55870691 on Build GRCh38
rs962983556 Chr.2: 55870791-55870791 on Build GRCh38
rs1042977278 Chr.2: 55870644-55870658 on Build GRCh38
rs779233876 Chr.2: 55870913-55870913 on Build GRCh38
rs933403907 Chr.2: 55870644-55870655 on Build GRCh38
rs909803841 Chr.2: 55870438-55870438 on Build GRCh38
rs767503998 Chr.2: 55870850-55870850 on Build GRCh38
rs758074708 Chr.2: 55870716-55870716 on Build GRCh38
rs762126695 Chr.2: 55870841-55870841 on Build GRCh38
rs756581829 Chr.2: 55870706-55870706 on Build GRCh38
rs532405890 Chr.2: 55870668-55870668 on Build GRCh38
rs367901228 Chr.2: 55870890-55870890 on Build GRCh38
rs963754384 Chr.2: 55870614-55870614 on Build GRCh38
rs754178385 Chr.2: 55870709-55870709 on Build GRCh38
rs776362721 Chr.2: 55870760-55870760 on Build GRCh38
rs1025522294 Chr.2: 55870934-55870934 on Build GRCh38
rs547851604 Chr.2: 55870787-55870787 on Build GRCh38
rs1039133013 Chr.2: 55870624-55870624 on Build GRCh38
rs1001109920 Chr.2: 55870667-55870667 on Build GRCh38
rs1055841911 Chr.2: 55870481-55870481 on Build GRCh38
rs754310502 Chr.2: 55870885-55870885 on Build GRCh38
rs915936853 Chr.2: 55870862-55870862 on Build GRCh38
rs893380597 Chr.2: 55870585-55870585 on Build GRCh38
rs112260833 Chr.2: 55870604-55870604 on Build GRCh38
rs530792570 Chr.2: 55870450-55870450 on Build GRCh38
rs751279285 Chr.2: 55870920-55870920 on Build GRCh38
rs529831738 Chr.2: 55870521-55870521 on Build GRCh38
rs780748234 Chr.2: 55870765-55870765 on Build GRCh38
rs202135588 Chr.2: 55870644-55870646 on Build GRCh38
rs567386311 Chr.2: 55870830-55870830 on Build GRCh38
rs139142559 Chr.2: 55870829-55870829 on Build GRCh38
rs545015967 Chr.2: 55870514-55870514 on Build GRCh38
rs950047658 Chr.2: 55870876-55870876 on Build GRCh38
rs775180017 Chr.2: 55870819-55870819 on Build GRCh38
rs770484375 Chr.2: 55870745-55870745 on Build GRCh38
rs780161368 Chr.2: 55870766-55870766 on Build GRCh38
rs750455273 Chr.2: 55870875-55870875 on Build GRCh38
rs991475191 Chr.2: 55870834-55870834 on Build GRCh38
rs1025111338 Chr.2: 55870468-55870468 on Build GRCh38
rs763290570 Chr.2: 55870690-55870690 on Build GRCh38

View Full Product Details