Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30892818-30893398 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL7C
Entrez Gene ID
9274
Gene Name
BCL tumor suppressor 7C
Gene Aliases
-
UniGene
Hs.658547
Gene Chromosome Location
Chr.16: 30834041-30894550 on Build GRCh38
Transcripts
NM_001286526.1 NM_004765.3 XM_011545980.2 XM_017023885.1 XM_017023886.1 XM_017023887.1 XM_017023888.1 NR_110940.1 NR_110941.1 NR_110942.1

Gene Symbol
MIR762HG
Entrez Gene ID
101928736
Gene Name
MIR762 host gene
Gene Aliases
-
UniGene
Hs.303197
Gene Chromosome Location
Chr.16: 30875460-30895220 on Build GRCh38
Transcripts
NM_001286526.1 NM_004765.3 XM_011545980.2 XM_017023885.1 XM_017023886.1 XM_017023887.1 XM_017023888.1 NR_110940.1 NR_110941.1 NR_110942.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
970019 c.209G>A p.R70H Chr.16: 30892911-30892911 on Build GRCh38
4578958 c.135C>T p.I45I Chr.16: 30893248-30893248 on Build GRCh38
1588414 c.209G>A p.R70H Chr.16: 30892911-30892911 on Build GRCh38
5815624 c.160C>T p.P54S Chr.16: 30893223-30893223 on Build GRCh38
4060261 c.145G>A p.V49M Chr.16: 30893238-30893238 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1042765609 Chr.16: 30893209-30893209 on Build GRCh38
rs770467743 Chr.16: 30893338-30893339 on Build GRCh38
rs764985607 Chr.16: 30892886-30892886 on Build GRCh38
rs767515381 Chr.16: 30892865-30892865 on Build GRCh38
rs751147279 Chr.16: 30892998-30892998 on Build GRCh38
rs933137129 Chr.16: 30893137-30893137 on Build GRCh38
rs1006916595 Chr.16: 30893099-30893099 on Build GRCh38
rs760585991 Chr.16: 30893057-30893057 on Build GRCh38
rs151160739 Chr.16: 30892913-30892913 on Build GRCh38
rs375463873 Chr.16: 30892966-30892966 on Build GRCh38
rs747562410 Chr.16: 30892916-30892917 on Build GRCh38
rs1018248915 Chr.16: 30893100-30893100 on Build GRCh38
rs566157935 Chr.16: 30893365-30893365 on Build GRCh38
rs769264449 Chr.16: 30893303-30893303 on Build GRCh38
rs772485153 Chr.16: 30893253-30893253 on Build GRCh38
rs947105182 Chr.16: 30893200-30893200 on Build GRCh38
rs367674953 Chr.16: 30892874-30892874 on Build GRCh38
rs974344809 Chr.16: 30893343-30893343 on Build GRCh38
rs774738787 Chr.16: 30892927-30892927 on Build GRCh38
rs1051189798 Chr.16: 30893054-30893054 on Build GRCh38
rs775514072 Chr.16: 30892951-30892951 on Build GRCh38
rs747834008 Chr.16: 30893223-30893224 on Build GRCh38
rs188345023 Chr.16: 30892912-30892912 on Build GRCh38
rs772932570 Chr.16: 30893338-30893338 on Build GRCh38
rs781729572 Chr.16: 30893203-30893205 on Build GRCh38
rs984059250 Chr.16: 30893064-30893064 on Build GRCh38
rs11558080 Chr.16: 30893282-30893282 on Build GRCh38
rs930079973 Chr.16: 30893046-30893046 on Build GRCh38
rs747158565 Chr.16: 30893283-30893283 on Build GRCh38
rs138080236 Chr.16: 30893130-30893130 on Build GRCh38
rs765027584 Chr.16: 30893308-30893308 on Build GRCh38
rs1015536052 Chr.16: 30893071-30893072 on Build GRCh38
rs763812959 Chr.16: 30892870-30892870 on Build GRCh38
rs751012085 Chr.16: 30893340-30893340 on Build GRCh38
rs778394196 Chr.16: 30893187-30893187 on Build GRCh38
rs199729916 Chr.16: 30892896-30892896 on Build GRCh38
rs781611007 Chr.16: 30892899-30892899 on Build GRCh38
rs897318822 Chr.16: 30893042-30893042 on Build GRCh38
rs756701332 Chr.16: 30892905-30892905 on Build GRCh38
rs962293372 Chr.16: 30893084-30893084 on Build GRCh38
rs35860871 Chr.16: 30892885-30892886 on Build GRCh38
rs543690251 Chr.16: 30892897-30892897 on Build GRCh38
rs1031289499 Chr.16: 30893247-30893247 on Build GRCh38
rs564218486 Chr.16: 30892869-30892869 on Build GRCh38
rs529563945 Chr.16: 30892952-30892952 on Build GRCh38
rs140570412 Chr.16: 30892928-30892928 on Build GRCh38
rs761445872 Chr.16: 30893304-30893304 on Build GRCh38
rs753728645 Chr.16: 30892871-30892871 on Build GRCh38
rs138503357 Chr.16: 30893252-30893252 on Build GRCh38
rs1038967149 Chr.16: 30892999-30892999 on Build GRCh38
rs752715761 Chr.16: 30892868-30892868 on Build GRCh38
rs776439179 Chr.16: 30893300-30893300 on Build GRCh38
rs376639437 Chr.16: 30893145-30893145 on Build GRCh38
rs749703437 Chr.16: 30892914-30892914 on Build GRCh38
rs772286392 Chr.16: 30892939-30892939 on Build GRCh38
rs769123797 Chr.16: 30892937-30892938 on Build GRCh38
rs764127931 Chr.16: 30892954-30892954 on Build GRCh38
rs562072564 Chr.16: 30892915-30892915 on Build GRCh38
rs1022964772 Chr.16: 30893047-30893047 on Build GRCh38
rs765861182 Chr.16: 30892994-30892994 on Build GRCh38
rs1000768790 Chr.16: 30893148-30893148 on Build GRCh38
rs779212353 Chr.16: 30893249-30893249 on Build GRCh38
rs776264213 Chr.16: 30892955-30892955 on Build GRCh38
rs749568126 Chr.16: 30893245-30893245 on Build GRCh38
rs760516010 Chr.16: 30893124-30893124 on Build GRCh38
rs927554649 Chr.16: 30892986-30892986 on Build GRCh38
rs762322590 Chr.16: 30893337-30893338 on Build GRCh38
rs145774897 Chr.16: 30893248-30893248 on Build GRCh38
rs372038719 Chr.16: 30892976-30892976 on Build GRCh38
rs930132381 Chr.16: 30893367-30893367 on Build GRCh38
rs774247463 Chr.16: 30893339-30893339 on Build GRCh38
rs773014933 Chr.16: 30892945-30892945 on Build GRCh38
rs917615271 Chr.16: 30892911-30892911 on Build GRCh38
rs1013425960 Chr.16: 30893001-30893001 on Build GRCh38
rs903800433 Chr.16: 30893110-30893110 on Build GRCh38
rs748667097 Chr.16: 30892900-30892900 on Build GRCh38
rs910280858 Chr.16: 30893189-30893189 on Build GRCh38
rs369885968 Chr.16: 30893301-30893301 on Build GRCh38
rs1024681730 Chr.16: 30893264-30893264 on Build GRCh38
rs908860435 Chr.16: 30893328-30893328 on Build GRCh38
rs371323761 Chr.16: 30893183-30893183 on Build GRCh38
rs761688237 Chr.16: 30892963-30892963 on Build GRCh38

View Full Product Details