Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 42155343-42155912 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGKH
Entrez Gene ID
160851
Gene Name
diacylglycerol kinase eta
Gene Aliases
DGKeta
UniGene
Hs.630380
Gene Chromosome Location
Chr.13: 42040036-42256584 on Build GRCh38
Transcripts
NM_001204504.2 NM_001204505.2 NM_001204506.2 NM_001297429.1 NM_152910.5 NM_178009.4 NR_123714.1 NR_123715.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4047661 c.147A>G p.R49R Chr.13: 42155732-42155732 on Build GRCh38
7312995 c.65C>T p.T22I Chr.13: 42155379-42155379 on Build GRCh38
4496777 c.74C>T p.P25L Chr.13: 42155388-42155388 on Build GRCh38
7263906 c.145C>T p.R49* Chr.13: 42155730-42155730 on Build GRCh38
6463222 c.470A>G p.Q157R Chr.13: 42155376-42155376 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765248663 Chr.13: 42155390-42155390 on Build GRCh38
rs377267510 Chr.13: 42155767-42155767 on Build GRCh38
rs908901587 Chr.13: 42155825-42155825 on Build GRCh38
rs759132220 Chr.13: 42155617-42155617 on Build GRCh38
rs745720508 Chr.13: 42155839-42155839 on Build GRCh38
rs368631270 Chr.13: 42155675-42155675 on Build GRCh38
rs777819084 Chr.13: 42155782-42155782 on Build GRCh38
rs572562790 Chr.13: 42155529-42155529 on Build GRCh38
rs200423019 Chr.13: 42155638-42155638 on Build GRCh38
rs758028199 Chr.13: 42155780-42155780 on Build GRCh38
rs994711443 Chr.13: 42155872-42155872 on Build GRCh38
rs533713720 Chr.13: 42155795-42155795 on Build GRCh38
rs1022813653 Chr.13: 42155454-42155454 on Build GRCh38
rs898034284 Chr.13: 42155467-42155467 on Build GRCh38
rs887413604 Chr.13: 42155425-42155425 on Build GRCh38
rs915422340 Chr.13: 42155615-42155615 on Build GRCh38
rs748801311 Chr.13: 42155639-42155639 on Build GRCh38
rs149168750 Chr.13: 42155380-42155380 on Build GRCh38
rs555895059 Chr.13: 42155472-42155472 on Build GRCh38
rs368434176 Chr.13: 42155849-42155849 on Build GRCh38
rs756562795 Chr.13: 42155637-42155637 on Build GRCh38
rs756431628 Chr.13: 42155737-42155737 on Build GRCh38
rs773279659 Chr.13: 42155387-42155387 on Build GRCh38
rs748030391 Chr.13: 42155653-42155653 on Build GRCh38
rs760236266 Chr.13: 42155599-42155599 on Build GRCh38
rs563703003 Chr.13: 42155879-42155879 on Build GRCh38
rs759188749 Chr.13: 42155406-42155406 on Build GRCh38
rs778615031 Chr.13: 42155651-42155651 on Build GRCh38
rs771457901 Chr.13: 42155458-42155458 on Build GRCh38
rs774412881 Chr.13: 42155715-42155715 on Build GRCh38
rs1023186160 Chr.13: 42155814-42155814 on Build GRCh38
rs753372667 Chr.13: 42155748-42155748 on Build GRCh38
rs753332605 Chr.13: 42155604-42155604 on Build GRCh38
rs750939540 Chr.13: 42155429-42155429 on Build GRCh38
rs778479763 Chr.13: 42155663-42155663 on Build GRCh38
rs919512127 Chr.13: 42155868-42155868 on Build GRCh38
rs370908868 Chr.13: 42155423-42155423 on Build GRCh38
rs763254069 Chr.13: 42155710-42155710 on Build GRCh38
rs912102056 Chr.13: 42155557-42155557 on Build GRCh38
rs543825290 Chr.13: 42155861-42155861 on Build GRCh38
rs61756563 Chr.13: 42155696-42155696 on Build GRCh38
rs956573742 Chr.13: 42155668-42155668 on Build GRCh38
rs754636578 Chr.13: 42155649-42155649 on Build GRCh38
rs78999869 Chr.13: 42155459-42155459 on Build GRCh38
rs776493614 Chr.13: 42155831-42155831 on Build GRCh38
rs541664420 Chr.13: 42155545-42155545 on Build GRCh38
rs750970180 Chr.13: 42155729-42155729 on Build GRCh38
rs1012926380 Chr.13: 42155539-42155539 on Build GRCh38
rs772129158 Chr.13: 42155673-42155673 on Build GRCh38
rs747682024 Chr.13: 42155443-42155443 on Build GRCh38
rs766788357 Chr.13: 42155747-42155747 on Build GRCh38
rs548428629 Chr.13: 42155418-42155418 on Build GRCh38
rs767991654 Chr.13: 42155727-42155727 on Build GRCh38
rs760392856 Chr.13: 42155515-42155515 on Build GRCh38
rs752061769 Chr.13: 42155792-42155792 on Build GRCh38
rs752392320 Chr.13: 42155804-42155804 on Build GRCh38
rs193136089 Chr.13: 42155726-42155726 on Build GRCh38
rs765687085 Chr.13: 42155600-42155600 on Build GRCh38
rs754475056 Chr.13: 42155386-42155387 on Build GRCh38
rs772954930 Chr.13: 42155485-42155485 on Build GRCh38
rs775530076 Chr.13: 42155699-42155699 on Build GRCh38
rs757610730 Chr.13: 42155422-42155422 on Build GRCh38
rs760625960 Chr.13: 42155388-42155388 on Build GRCh38
rs112210188 Chr.13: 42155410-42155410 on Build GRCh38
rs746775394 Chr.13: 42155687-42155687 on Build GRCh38
rs768588983 Chr.13: 42155392-42155392 on Build GRCh38
rs774235073 Chr.13: 42155409-42155409 on Build GRCh38
rs754662217 Chr.13: 42155759-42155759 on Build GRCh38
rs199776370 Chr.13: 42155714-42155714 on Build GRCh38
rs941691268 Chr.13: 42155843-42155843 on Build GRCh38
rs200846322 Chr.13: 42155374-42155374 on Build GRCh38
rs770578656 Chr.13: 42155817-42155817 on Build GRCh38
rs542334578 Chr.13: 42155468-42155468 on Build GRCh38

View Full Product Details