Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 98807764-98808326 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCBLD2
Entrez Gene ID
131566
Gene Name
discoidin, CUB and LCCL domain containing 2
Gene Aliases
CLCP1, ESDN
UniGene
Hs.203691
Gene Chromosome Location
Chr.3: 98795970-98901689 on Build GRCh38
Transcripts
NM_080927.3 XM_011512419.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5524815 c.1661G>A p.W554* Chr.3: 98808090-98808090 on Build GRCh38
5524814 c.1661G>A p.W554* Chr.3: 98808090-98808090 on Build GRCh38
4584677 c.1611G>A p.L537L Chr.3: 98808140-98808140 on Build GRCh38
4469538 c.1603C>T p.P535S Chr.3: 98808148-98808148 on Build GRCh38
6862327 c.1616T>C p.M539T Chr.3: 98808135-98808135 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202236000 Chr.3: 98808140-98808140 on Build GRCh38
rs984222040 Chr.3: 98807987-98807988 on Build GRCh38
rs771675780 Chr.3: 98808072-98808072 on Build GRCh38
rs778608317 Chr.3: 98808151-98808151 on Build GRCh38
rs775730018 Chr.3: 98808202-98808202 on Build GRCh38
rs1030002526 Chr.3: 98808183-98808183 on Build GRCh38
rs909941069 Chr.3: 98808282-98808282 on Build GRCh38
rs1021790043 Chr.3: 98807806-98807806 on Build GRCh38
rs376491392 Chr.3: 98808112-98808112 on Build GRCh38
rs778030660 Chr.3: 98808033-98808033 on Build GRCh38
rs769152177 Chr.3: 98808060-98808060 on Build GRCh38
rs1018625135 Chr.3: 98808147-98808147 on Build GRCh38
rs747016845 Chr.3: 98808047-98808047 on Build GRCh38
rs200611163 Chr.3: 98808087-98808087 on Build GRCh38
rs997148616 Chr.3: 98808180-98808180 on Build GRCh38
rs1009980555 Chr.3: 98808296-98808296 on Build GRCh38
rs200123782 Chr.3: 98808134-98808134 on Build GRCh38
rs758552243 Chr.3: 98808117-98808117 on Build GRCh38
rs900432235 Chr.3: 98808176-98808176 on Build GRCh38
rs185960453 Chr.3: 98807907-98807907 on Build GRCh38
rs765541850 Chr.3: 98808105-98808105 on Build GRCh38
rs770268811 Chr.3: 98808199-98808199 on Build GRCh38
rs529257250 Chr.3: 98807928-98807928 on Build GRCh38
rs780970624 Chr.3: 98808283-98808283 on Build GRCh38
rs984641118 Chr.3: 98808272-98808272 on Build GRCh38
rs34472032 Chr.3: 98808167-98808167 on Build GRCh38
rs764352446 Chr.3: 98808121-98808121 on Build GRCh38
rs35912538 Chr.3: 98808106-98808106 on Build GRCh38
rs753064561 Chr.3: 98808113-98808113 on Build GRCh38
rs372584623 Chr.3: 98808182-98808182 on Build GRCh38
rs777072457 Chr.3: 98808100-98808100 on Build GRCh38
rs28417568 Chr.3: 98808020-98808020 on Build GRCh38
rs751815009 Chr.3: 98808122-98808122 on Build GRCh38
rs760122121 Chr.3: 98808076-98808076 on Build GRCh38
rs976545131 Chr.3: 98807813-98807813 on Build GRCh38
rs918683773 Chr.3: 98807838-98807838 on Build GRCh38
rs140457302 Chr.3: 98808196-98808197 on Build GRCh38
rs992802194 Chr.3: 98808062-98808062 on Build GRCh38
rs747912159 Chr.3: 98808165-98808165 on Build GRCh38
rs909898626 Chr.3: 98807989-98807989 on Build GRCh38
rs578068977 Chr.3: 98807945-98807945 on Build GRCh38
rs375936942 Chr.3: 98808196-98808200 on Build GRCh38
rs951479913 Chr.3: 98807917-98807917 on Build GRCh38
rs756131651 Chr.3: 98808053-98808053 on Build GRCh38
rs759991123 Chr.3: 98808186-98808186 on Build GRCh38
rs951176799 Chr.3: 98808232-98808232 on Build GRCh38
rs201629117 Chr.3: 98808138-98808138 on Build GRCh38
rs369024442 Chr.3: 98808168-98808168 on Build GRCh38
rs768371584 Chr.3: 98808056-98808056 on Build GRCh38
rs763465355 Chr.3: 98808204-98808204 on Build GRCh38
rs1021821498 Chr.3: 98808300-98808300 on Build GRCh38
rs377166133 Chr.3: 98807811-98807812 on Build GRCh38
rs760057507 Chr.3: 98808102-98808102 on Build GRCh38
rs772717161 Chr.3: 98808073-98808073 on Build GRCh38
rs866355994 Chr.3: 98807984-98807984 on Build GRCh38
rs774176936 Chr.3: 98808059-98808059 on Build GRCh38
rs780195068 Chr.3: 98808055-98808055 on Build GRCh38

View Full Product Details