Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 181243517-181243955 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RACK1
Entrez Gene ID
10399
Gene Name
receptor for activated C kinase 1
Gene Aliases
GNB2L1, Gnb2-rs1, H12.3, HLC-7, PIG21
UniGene
Hs.5662
Gene Chromosome Location
Chr.5: 181236928-181243906 on Build GRCh38
Transcripts
NM_006098.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6573172 c.106C>G p.R36G Chr.5: 181243695-181243695 on Build GRCh38
4745968 c.22C>T p.R8C Chr.5: 181243779-181243779 on Build GRCh38
6573173 c.106C>G p.R36G Chr.5: 181243695-181243695 on Build GRCh38
1253364 c.107G>C p.R36P Chr.5: 181243694-181243694 on Build GRCh38
7002800 c.82C>T p.P28S Chr.5: 181243719-181243719 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs531179975 Chr.5: 181243555-181243555 on Build GRCh38
rs11540191 Chr.5: 181243785-181243785 on Build GRCh38
rs535673815 Chr.5: 181243546-181243546 on Build GRCh38
rs752963603 Chr.5: 181243708-181243708 on Build GRCh38
rs750045936 Chr.5: 181243902-181243902 on Build GRCh38
rs933427267 Chr.5: 181243864-181243864 on Build GRCh38
rs11540179 Chr.5: 181243874-181243874 on Build GRCh38
rs2241369 Chr.5: 181243858-181243858 on Build GRCh38
rs535433416 Chr.5: 181243900-181243901 on Build GRCh38
rs762326344 Chr.5: 181243927-181243927 on Build GRCh38
rs11540182 Chr.5: 181243859-181243859 on Build GRCh38
rs746632997 Chr.5: 181243904-181243905 on Build GRCh38
rs900598779 Chr.5: 181243582-181243582 on Build GRCh38
rs776038456 Chr.5: 181243816-181243816 on Build GRCh38
rs985316572 Chr.5: 181243569-181243569 on Build GRCh38
rs764512172 Chr.5: 181243821-181243821 on Build GRCh38
rs748101749 Chr.5: 181243556-181243556 on Build GRCh38
rs975278547 Chr.5: 181243930-181243930 on Build GRCh38
rs11540173 Chr.5: 181243732-181243732 on Build GRCh38
rs11540174 Chr.5: 181243831-181243831 on Build GRCh38
rs540731944 Chr.5: 181243891-181243891 on Build GRCh38
rs751944608 Chr.5: 181243683-181243684 on Build GRCh38
rs11249779 Chr.5: 181243801-181243801 on Build GRCh38
rs113011333 Chr.5: 181243691-181243691 on Build GRCh38
rs754057723 Chr.5: 181243729-181243729 on Build GRCh38
rs533647123 Chr.5: 181243895-181243895 on Build GRCh38
rs745953408 Chr.5: 181243748-181243748 on Build GRCh38
rs1059315 Chr.5: 181243851-181243851 on Build GRCh38
rs535004360 Chr.5: 181243652-181243652 on Build GRCh38
rs762479736 Chr.5: 181243810-181243810 on Build GRCh38
rs951197299 Chr.5: 181243910-181243910 on Build GRCh38
rs768468889 Chr.5: 181243783-181243783 on Build GRCh38
rs566222261 Chr.5: 181243622-181243622 on Build GRCh38
rs61735487 Chr.5: 181243904-181243904 on Build GRCh38
rs11540164 Chr.5: 181243756-181243756 on Build GRCh38
rs909504579 Chr.5: 181243921-181243921 on Build GRCh38
rs1018348667 Chr.5: 181243577-181243577 on Build GRCh38
rs983808036 Chr.5: 181243911-181243911 on Build GRCh38
rs959783483 Chr.5: 181243806-181243806 on Build GRCh38
rs928913281 Chr.5: 181243680-181243680 on Build GRCh38
rs757617206 Chr.5: 181243734-181243734 on Build GRCh38
rs746639954 Chr.5: 181243901-181243904 on Build GRCh38
rs1048283863 Chr.5: 181243554-181243554 on Build GRCh38
rs997522727 Chr.5: 181243587-181243587 on Build GRCh38
rs768144690 Chr.5: 181243665-181243665 on Build GRCh38
rs759976987 Chr.5: 181243695-181243695 on Build GRCh38
rs746969023 Chr.5: 181243768-181243768 on Build GRCh38
rs758531415 Chr.5: 181243841-181243841 on Build GRCh38
rs1030302881 Chr.5: 181243644-181243644 on Build GRCh38
rs1279737 Chr.5: 181243885-181243885 on Build GRCh38
rs769447572 Chr.5: 181243804-181243804 on Build GRCh38
rs746964357 Chr.5: 181243845-181243845 on Build GRCh38
rs775914317 Chr.5: 181243541-181243541 on Build GRCh38
rs569717293 Chr.5: 181243542-181243542 on Build GRCh38
rs772641866 Chr.5: 181243807-181243807 on Build GRCh38
rs763566484 Chr.5: 181243639-181243639 on Build GRCh38
rs891864288 Chr.5: 181243872-181243872 on Build GRCh38
rs776357433 Chr.5: 181243789-181243789 on Build GRCh38
rs532780109 Chr.5: 181243908-181243908 on Build GRCh38
rs909840517 Chr.5: 181243839-181243839 on Build GRCh38
rs1029569422 Chr.5: 181243827-181243827 on Build GRCh38
rs558141707 Chr.5: 181243676-181243676 on Build GRCh38
rs1039510075 Chr.5: 181243563-181243563 on Build GRCh38
rs537314343 Chr.5: 181243792-181243792 on Build GRCh38
rs891027251 Chr.5: 181243922-181243922 on Build GRCh38
rs779081202 Chr.5: 181243737-181243737 on Build GRCh38
rs574132667 Chr.5: 181243849-181243849 on Build GRCh38
rs765591595 Chr.5: 181243830-181243830 on Build GRCh38
rs749991697 Chr.5: 181243688-181243688 on Build GRCh38
rs1025764756 Chr.5: 181243867-181243867 on Build GRCh38
rs754170527 Chr.5: 181243824-181243824 on Build GRCh38
rs929895759 Chr.5: 181243553-181243553 on Build GRCh38
rs536177657 Chr.5: 181243906-181243906 on Build GRCh38
rs904773818 Chr.5: 181243881-181243881 on Build GRCh38
rs369560198 Chr.5: 181243818-181243818 on Build GRCh38
rs73814547 Chr.5: 181243834-181243834 on Build GRCh38
rs766166534 Chr.5: 181243907-181243907 on Build GRCh38
rs776216120 Chr.5: 181243809-181243809 on Build GRCh38
rs918589722 Chr.5: 181243632-181243632 on Build GRCh38
rs1006817008 Chr.5: 181243925-181243925 on Build GRCh38
rs764723382 Chr.5: 181243686-181243686 on Build GRCh38
rs866495306 Chr.5: 181243775-181243775 on Build GRCh38
rs11540165 Chr.5: 181243720-181243720 on Build GRCh38
rs775362711 Chr.5: 181243651-181243651 on Build GRCh38
rs774778243 Chr.5: 181243916-181243916 on Build GRCh38
rs756520547 Chr.5: 181243717-181243717 on Build GRCh38
rs1051821861 Chr.5: 181243865-181243865 on Build GRCh38
rs757490189 Chr.5: 181243825-181243825 on Build GRCh38
rs11540176 Chr.5: 181243876-181243876 on Build GRCh38
rs746024453 Chr.5: 181243907-181243911 on Build GRCh38
rs930025507 Chr.5: 181243633-181243633 on Build GRCh38
rs376324276 Chr.5: 181243682-181243682 on Build GRCh38
rs761337978 Chr.5: 181243837-181243837 on Build GRCh38
rs41285573 Chr.5: 181243898-181243898 on Build GRCh38
rs578087778 Chr.5: 181243705-181243705 on Build GRCh38
rs955190780 Chr.5: 181243565-181243565 on Build GRCh38
rs918256211 Chr.5: 181243613-181243633 on Build GRCh38
rs767959123 Chr.5: 181243814-181243814 on Build GRCh38
rs777449136 Chr.5: 181243846-181243846 on Build GRCh38
rs918264910 Chr.5: 181243826-181243826 on Build GRCh38
rs778459490 Chr.5: 181243753-181243753 on Build GRCh38

View Full Product Details