Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 14696014-14696610 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf60
Entrez Gene ID
144608
Gene Name
chromosome 12 open reading frame 60
Gene Aliases
-
UniGene
Hs.659951
Gene Chromosome Location
Chr.12: 14567991-14823858 on Build GRCh38
Transcripts
XM_017018871.1 XM_017018872.1 NM_004963.3 XM_011520631.2

Gene Symbol
GUCY2C
Entrez Gene ID
2984
Gene Name
guanylate cyclase 2C
Gene Aliases
DIAR6, GUC2C, MECIL, MUCIL, STAR
UniGene
Hs.524278
Gene Chromosome Location
Chr.12: 14612632-14696625 on Build GRCh38
Transcripts
XM_017018871.1 XM_017018872.1 NM_004963.3 XM_011520631.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3458608 c.202C>G p.R68G Chr.12: 14696247-14696247 on Build GRCh38
71007 c.67A>G p.S23G Chr.12: 14696382-14696382 on Build GRCh38
937542 c.51C>A p.P17P Chr.12: 14696398-14696398 on Build GRCh38
3458610 c.138C>T p.A46A Chr.12: 14696311-14696311 on Build GRCh38
4040575 c.98G>T p.G33V Chr.12: 14696351-14696351 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1048000280 Chr.12: 14696304-14696304 on Build GRCh38
rs867791055 Chr.12: 14696343-14696343 on Build GRCh38
rs983358248 Chr.12: 14696456-14696456 on Build GRCh38
rs557403064 Chr.12: 14696046-14696046 on Build GRCh38
rs776591086 Chr.12: 14696194-14696194 on Build GRCh38
rs955308418 Chr.12: 14696111-14696111 on Build GRCh38
rs1011246395 Chr.12: 14696570-14696570 on Build GRCh38
rs777382774 Chr.12: 14696375-14696375 on Build GRCh38
rs770700132 Chr.12: 14696061-14696061 on Build GRCh38
rs372188312 Chr.12: 14696389-14696389 on Build GRCh38
rs148552421 Chr.12: 14696460-14696460 on Build GRCh38
rs1030500856 Chr.12: 14696091-14696091 on Build GRCh38
rs367797255 Chr.12: 14696335-14696335 on Build GRCh38
rs757449116 Chr.12: 14696234-14696234 on Build GRCh38
rs770392197 Chr.12: 14696396-14696396 on Build GRCh38
rs764466536 Chr.12: 14696222-14696222 on Build GRCh38
rs915647221 Chr.12: 14696252-14696255 on Build GRCh38
rs201089497 Chr.12: 14696348-14696348 on Build GRCh38
rs368411824 Chr.12: 14696148-14696150 on Build GRCh38
rs1005992342 Chr.12: 14696068-14696068 on Build GRCh38
rs771612876 Chr.12: 14696279-14696279 on Build GRCh38
rs542694223 Chr.12: 14696090-14696090 on Build GRCh38
rs929235200 Chr.12: 14696285-14696285 on Build GRCh38
rs756459521 Chr.12: 14696258-14696258 on Build GRCh38
rs56142849 Chr.12: 14696361-14696361 on Build GRCh38
rs750874628 Chr.12: 14696358-14696358 on Build GRCh38
rs1022308769 Chr.12: 14696368-14696368 on Build GRCh38
rs779803278 Chr.12: 14696491-14696491 on Build GRCh38
rs1051477404 Chr.12: 14696573-14696573 on Build GRCh38
rs550836055 Chr.12: 14696246-14696246 on Build GRCh38
rs780575858 Chr.12: 14696397-14696397 on Build GRCh38
rs149332679 Chr.12: 14696440-14696440 on Build GRCh38
rs1010347173 Chr.12: 14696109-14696109 on Build GRCh38
rs55930625 Chr.12: 14696398-14696398 on Build GRCh38
rs141230079 Chr.12: 14696281-14696281 on Build GRCh38
rs569085139 Chr.12: 14696292-14696292 on Build GRCh38
rs992870090 Chr.12: 14696231-14696231 on Build GRCh38
rs750915945 Chr.12: 14696247-14696247 on Build GRCh38
rs538893070 Chr.12: 14696461-14696461 on Build GRCh38
rs747780878 Chr.12: 14696278-14696278 on Build GRCh38
rs193019947 Chr.12: 14696124-14696124 on Build GRCh38
rs933094927 Chr.12: 14696566-14696566 on Build GRCh38
rs764376792 Chr.12: 14696334-14696334 on Build GRCh38
rs981603322 Chr.12: 14696587-14696587 on Build GRCh38
rs766765359 Chr.12: 14696451-14696451 on Build GRCh38
rs557633657 Chr.12: 14696537-14696537 on Build GRCh38
rs751681985 Chr.12: 14696226-14696226 on Build GRCh38
rs999185188 Chr.12: 14696170-14696170 on Build GRCh38
rs565108190 Chr.12: 14696199-14696199 on Build GRCh38
rs754417503 Chr.12: 14696452-14696452 on Build GRCh38
rs780283983 Chr.12: 14696276-14696276 on Build GRCh38
rs752927373 Chr.12: 14696218-14696218 on Build GRCh38
rs760876233 Chr.12: 14696441-14696441 on Build GRCh38
rs190691284 Chr.12: 14696103-14696103 on Build GRCh38
rs980184030 Chr.12: 14696200-14696200 on Build GRCh38
rs760029303 Chr.12: 14696434-14696435 on Build GRCh38
rs774667505 Chr.12: 14696397-14696398 on Build GRCh38
rs534813812 Chr.12: 14696438-14696438 on Build GRCh38
rs746492221 Chr.12: 14696294-14696294 on Build GRCh38
rs755695884 Chr.12: 14696489-14696489 on Build GRCh38
rs202042180 Chr.12: 14696393-14696393 on Build GRCh38
rs1039318578 Chr.12: 14696495-14696495 on Build GRCh38
rs766300237 Chr.12: 14696192-14696192 on Build GRCh38
rs900136512 Chr.12: 14696076-14696076 on Build GRCh38
rs892030117 Chr.12: 14696341-14696341 on Build GRCh38
rs767736094 Chr.12: 14696356-14696356 on Build GRCh38
rs113101805 Chr.12: 14696064-14696064 on Build GRCh38
rs772536502 Chr.12: 14696499-14696500 on Build GRCh38
rs1021819287 Chr.12: 14696128-14696128 on Build GRCh38
rs76042810 Chr.12: 14696048-14696048 on Build GRCh38
rs371202760 Chr.12: 14696188-14696188 on Build GRCh38
rs138766659 Chr.12: 14696400-14696400 on Build GRCh38
rs774673667 Chr.12: 14696431-14696431 on Build GRCh38
rs373943895 Chr.12: 14696311-14696311 on Build GRCh38
rs900263375 Chr.12: 14696326-14696326 on Build GRCh38
rs1039212057 Chr.12: 14696074-14696074 on Build GRCh38
rs770482109 Chr.12: 14696299-14696299 on Build GRCh38
rs142982970 Chr.12: 14696551-14696551 on Build GRCh38
rs780962783 Chr.12: 14696471-14696471 on Build GRCh38
rs767646045 Chr.12: 14696439-14696439 on Build GRCh38
rs3217156 Chr.12: 14696198-14696199 on Build GRCh38
rs762628946 Chr.12: 14696401-14696401 on Build GRCh38
rs745335107 Chr.12: 14696475-14696475 on Build GRCh38
rs369567498 Chr.12: 14696362-14696362 on Build GRCh38
rs776134465 Chr.12: 14696501-14696513 on Build GRCh38
rs969764040 Chr.12: 14696575-14696576 on Build GRCh38
rs754150242 Chr.12: 14696371-14696371 on Build GRCh38
rs759356904 Chr.12: 14696209-14696209 on Build GRCh38
rs144941598 Chr.12: 14696306-14696306 on Build GRCh38
rs771542070 Chr.12: 14696186-14696186 on Build GRCh38
rs1051406865 Chr.12: 14696336-14696336 on Build GRCh38
rs756919611 Chr.12: 14696466-14696466 on Build GRCh38
rs771309755 Chr.12: 14696323-14696325 on Build GRCh38
rs79112402 Chr.12: 14696212-14696212 on Build GRCh38
rs148527471 Chr.12: 14696362-14696363 on Build GRCh38
rs768594460 Chr.12: 14696414-14696414 on Build GRCh38
rs760236901 Chr.12: 14696191-14696191 on Build GRCh38
rs755377049 Chr.12: 14696374-14696374 on Build GRCh38
rs774857024 Chr.12: 14696266-14696266 on Build GRCh38
rs776506885 Chr.12: 14696301-14696301 on Build GRCh38
rs370587605 Chr.12: 14696220-14696220 on Build GRCh38
rs540380156 Chr.12: 14696157-14696157 on Build GRCh38
rs890972657 Chr.12: 14696470-14696470 on Build GRCh38

View Full Product Details