Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155709592-155710083 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAP3
Entrez Gene ID
7818
Gene Name
death associated protein 3
Gene Aliases
DAP-3, MRP-S29, MRPS29, bMRP-10
UniGene
Hs.516746
Gene Chromosome Location
Chr.1: 155688104-155739010 on Build GRCh38
Transcripts
NM_001199849.1 NM_001199850.1 NM_001199851.1 NM_004632.3 NM_033657.2 XM_005245480.2 XM_005245481.2 XM_005245482.4 XM_011509972.2 XM_017002289.1 XM_017002290.1 XM_017002291.1 XM_017002292.1 XM_017002293.1 XM_017002294.1 XM_017002295.1 XM_017002296.1 XM_017002297.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
243954 c.13G>A p.G5R Chr.1: 155709792-155709792 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs369497354 Chr.1: 155709827-155709827 on Build GRCh38
rs921903734 Chr.1: 155710055-155710055 on Build GRCh38
rs757952600 Chr.1: 155709917-155709917 on Build GRCh38
rs983082739 Chr.1: 155709672-155709672 on Build GRCh38
rs577450784 Chr.1: 155709930-155709930 on Build GRCh38
rs374129610 Chr.1: 155709745-155709745 on Build GRCh38
rs768798268 Chr.1: 155709752-155709752 on Build GRCh38
rs891253244 Chr.1: 155709777-155709777 on Build GRCh38
rs912018958 Chr.1: 155709918-155709918 on Build GRCh38
rs80349525 Chr.1: 155709764-155709764 on Build GRCh38
rs569493537 Chr.1: 155709990-155709993 on Build GRCh38
rs974781984 Chr.1: 155709952-155709952 on Build GRCh38
rs41264961 Chr.1: 155709967-155709967 on Build GRCh38
rs191412913 Chr.1: 155709768-155709768 on Build GRCh38
rs768368044 Chr.1: 155709770-155709770 on Build GRCh38
rs201816092 Chr.1: 155709766-155709766 on Build GRCh38
rs1055411821 Chr.1: 155709890-155709890 on Build GRCh38
rs372695062 Chr.1: 155709753-155709753 on Build GRCh38
rs908842661 Chr.1: 155709684-155709684 on Build GRCh38
rs775220162 Chr.1: 155709848-155709849 on Build GRCh38
rs764932759 Chr.1: 155709695-155709695 on Build GRCh38
rs758030216 Chr.1: 155709759-155709759 on Build GRCh38
rs776958230 Chr.1: 155709753-155709754 on Build GRCh38
rs933167319 Chr.1: 155709762-155709762 on Build GRCh38
rs528088972 Chr.1: 155709873-155709873 on Build GRCh38
rs752960570 Chr.1: 155709737-155709737 on Build GRCh38
rs759250668 Chr.1: 155709753-155709755 on Build GRCh38
rs1013747794 Chr.1: 155709935-155709935 on Build GRCh38
rs560182486 Chr.1: 155709990-155709990 on Build GRCh38
rs186517382 Chr.1: 155709622-155709622 on Build GRCh38
rs764152940 Chr.1: 155709820-155709820 on Build GRCh38
rs574730874 Chr.1: 155709978-155709978 on Build GRCh38
rs767566212 Chr.1: 155709732-155709732 on Build GRCh38
rs527803451 Chr.1: 155710002-155710002 on Build GRCh38
rs760898731 Chr.1: 155709814-155709814 on Build GRCh38
rs746997478 Chr.1: 155709744-155709744 on Build GRCh38
rs765613275 Chr.1: 155709840-155709840 on Build GRCh38
rs775495982 Chr.1: 155709807-155709807 on Build GRCh38
rs199978100 Chr.1: 155709834-155709834 on Build GRCh38
rs1001258091 Chr.1: 155709854-155709854 on Build GRCh38
rs535872383 Chr.1: 155709747-155709747 on Build GRCh38
rs764536960 Chr.1: 155709742-155709742 on Build GRCh38
rs758926936 Chr.1: 155709862-155709862 on Build GRCh38
rs974799431 Chr.1: 155710045-155710045 on Build GRCh38
rs554664018 Chr.1: 155709754-155709754 on Build GRCh38
rs113934070 Chr.1: 155709766-155709767 on Build GRCh38
rs950469351 Chr.1: 155709634-155709634 on Build GRCh38
rs756284884 Chr.1: 155709741-155709741 on Build GRCh38
rs747962666 Chr.1: 155709779-155709779 on Build GRCh38
rs950994884 Chr.1: 155709829-155709829 on Build GRCh38
rs372811843 Chr.1: 155709785-155709785 on Build GRCh38
rs540017090 Chr.1: 155710030-155710030 on Build GRCh38
rs541870883 Chr.1: 155709979-155709979 on Build GRCh38
rs878885778 Chr.1: 155709689-155709689 on Build GRCh38
rs762850317 Chr.1: 155709794-155709794 on Build GRCh38
rs544928178 Chr.1: 155709959-155709959 on Build GRCh38
rs751007324 Chr.1: 155709843-155709843 on Build GRCh38
rs781046714 Chr.1: 155709778-155709778 on Build GRCh38
rs61755343 Chr.1: 155709793-155709793 on Build GRCh38
rs770252868 Chr.1: 155709752-155709753 on Build GRCh38
rs1033447582 Chr.1: 155709701-155709701 on Build GRCh38
rs772166563 Chr.1: 155709799-155709799 on Build GRCh38

View Full Product Details