Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 24030475-24031074 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDCP
Entrez Gene ID
80304
Gene Name
WD repeat and coiled coil containing
Gene Aliases
C2orf44, PP384
UniGene
Hs.24624
Gene Chromosome Location
Chr.2: 24029336-24047426 on Build GRCh38
Transcripts
NM_001142319.1 NM_025203.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4093113 c.2165G>C p.*722S Chr.2: 24030934-24030934 on Build GRCh38
3580118 c.2097C>A p.V699V Chr.2: 24031002-24031002 on Build GRCh38
4583386 c.2129C>A p.T710N Chr.2: 24030970-24030970 on Build GRCh38
418783 c.2057C>G p.S686C Chr.2: 24031042-24031042 on Build GRCh38
2910752 c.2078C>T p.A693V Chr.2: 24031021-24031021 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748960024 Chr.2: 24030768-24030768 on Build GRCh38
rs1045860467 Chr.2: 24030832-24030832 on Build GRCh38
rs572531714 Chr.2: 24030843-24030843 on Build GRCh38
rs116230547 Chr.2: 24030856-24030856 on Build GRCh38
rs949762017 Chr.2: 24030824-24030824 on Build GRCh38
rs771463002 Chr.2: 24030726-24030726 on Build GRCh38
rs926141760 Chr.2: 24030767-24030767 on Build GRCh38
rs746298644 Chr.2: 24030959-24030959 on Build GRCh38
rs543973180 Chr.2: 24030730-24030730 on Build GRCh38
rs754592659 Chr.2: 24030770-24030770 on Build GRCh38
rs747960017 Chr.2: 24030978-24030978 on Build GRCh38
rs773676280 Chr.2: 24030865-24030865 on Build GRCh38
rs1007407822 Chr.2: 24030882-24030882 on Build GRCh38
rs922716903 Chr.2: 24030722-24030722 on Build GRCh38
rs116735256 Chr.2: 24030897-24030897 on Build GRCh38
rs768269427 Chr.2: 24030970-24030970 on Build GRCh38
rs761406175 Chr.2: 24030875-24030875 on Build GRCh38
rs769510936 Chr.2: 24030985-24030985 on Build GRCh38
rs1014480203 Chr.2: 24030584-24030584 on Build GRCh38
rs779839076 Chr.2: 24030731-24030731 on Build GRCh38
rs779599314 Chr.2: 24030952-24030952 on Build GRCh38
rs977613758 Chr.2: 24030757-24030757 on Build GRCh38
rs1018218154 Chr.2: 24030644-24030644 on Build GRCh38
rs754228362 Chr.2: 24030938-24030938 on Build GRCh38
rs764591367 Chr.2: 24030926-24030926 on Build GRCh38
rs1028375501 Chr.2: 24031041-24031041 on Build GRCh38
rs12622321 Chr.2: 24030661-24030661 on Build GRCh38
rs761003836 Chr.2: 24031025-24031025 on Build GRCh38
rs866386521 Chr.2: 24031022-24031022 on Build GRCh38
rs369394458 Chr.2: 24031031-24031031 on Build GRCh38
rs538497130 Chr.2: 24030846-24030846 on Build GRCh38
rs199629924 Chr.2: 24030944-24030944 on Build GRCh38
rs71439110 Chr.2: 24030792-24030792 on Build GRCh38
rs908490881 Chr.2: 24030775-24030775 on Build GRCh38
rs181820409 Chr.2: 24030593-24030593 on Build GRCh38
rs759665546 Chr.2: 24030892-24030892 on Build GRCh38
rs921282196 Chr.2: 24030721-24030721 on Build GRCh38
rs897625844 Chr.2: 24030576-24030576 on Build GRCh38
rs576606015 Chr.2: 24030709-24030709 on Build GRCh38
rs375795379 Chr.2: 24030883-24030883 on Build GRCh38
rs937773209 Chr.2: 24030829-24030829 on Build GRCh38
rs753019586 Chr.2: 24030900-24030900 on Build GRCh38
rs755458330 Chr.2: 24031037-24031038 on Build GRCh38
rs746745069 Chr.2: 24030964-24030964 on Build GRCh38
rs896128423 Chr.2: 24030548-24030548 on Build GRCh38
rs776459122 Chr.2: 24030972-24030972 on Build GRCh38
rs559961538 Chr.2: 24030858-24030858 on Build GRCh38
rs746344688 Chr.2: 24030915-24030915 on Build GRCh38
rs554829509 Chr.2: 24030703-24030703 on Build GRCh38
rs137952507 Chr.2: 24030792-24030793 on Build GRCh38
rs1045505598 Chr.2: 24030533-24030533 on Build GRCh38
rs530359373 Chr.2: 24030868-24030868 on Build GRCh38
rs915558946 Chr.2: 24030817-24030817 on Build GRCh38
rs1007465247 Chr.2: 24030634-24030634 on Build GRCh38
rs1056079452 Chr.2: 24030811-24030811 on Build GRCh38
rs964031205 Chr.2: 24030708-24030708 on Build GRCh38
rs372743789 Chr.2: 24030905-24030905 on Build GRCh38
rs557313845 Chr.2: 24030615-24030615 on Build GRCh38
rs201298184 Chr.2: 24031004-24031004 on Build GRCh38
rs771358286 Chr.2: 24030792-24030794 on Build GRCh38
rs767701889 Chr.2: 24031024-24031024 on Build GRCh38
rs376272507 Chr.2: 24030963-24030963 on Build GRCh38
rs757941554 Chr.2: 24030960-24030960 on Build GRCh38
rs773147439 Chr.2: 24030986-24030986 on Build GRCh38
rs1033795280 Chr.2: 24030542-24030542 on Build GRCh38
rs936222363 Chr.2: 24030795-24030795 on Build GRCh38
rs556436514 Chr.2: 24030723-24030723 on Build GRCh38
rs1033162627 Chr.2: 24030669-24030669 on Build GRCh38
rs373936545 Chr.2: 24031033-24031033 on Build GRCh38
rs986721672 Chr.2: 24030774-24030774 on Build GRCh38
rs1001944955 Chr.2: 24030515-24030515 on Build GRCh38
rs758774827 Chr.2: 24030508-24030508 on Build GRCh38
rs887641291 Chr.2: 24030880-24030880 on Build GRCh38
rs1027490944 Chr.2: 24030609-24030609 on Build GRCh38
rs957714616 Chr.2: 24030689-24030689 on Build GRCh38
rs1033158850 Chr.2: 24031019-24031019 on Build GRCh38
rs762784196 Chr.2: 24031007-24031007 on Build GRCh38

View Full Product Details