Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 57911596-57912025 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNGB1
Entrez Gene ID
1258
Gene Name
cyclic nucleotide gated channel beta 1
Gene Aliases
CNCG2, CNCG3L, CNCG4, CNG4, CNGB1B, GAR1, GARP, GARP2, RCNC2, RCNCb, RCNCbeta, RP45
UniGene
Hs.147062
Gene Chromosome Location
Chr.16: 57882340-57971116 on Build GRCh38
Transcripts
NM_001286130.1 NM_001297.4 XM_011522870.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3510576 c.2486G>A p.G829E Chr.16: 57911759-57911759 on Build GRCh38
4767872 c.2422C>A p.L808I Chr.16: 57911823-57911823 on Build GRCh38
3421053 c.2438C>T p.S813L Chr.16: 57911807-57911807 on Build GRCh38
3888698 c.2422C>T p.L808F Chr.16: 57911823-57911823 on Build GRCh38
971911 c.2454C>T p.L818L Chr.16: 57911791-57911791 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757437448 Chr.16: 57911902-57911902 on Build GRCh38
rs770908387 Chr.16: 57911893-57911893 on Build GRCh38
rs528283367 Chr.16: 57911791-57911791 on Build GRCh38
rs765152738 Chr.16: 57911889-57911889 on Build GRCh38
rs777741759 Chr.16: 57911898-57911898 on Build GRCh38
rs140865914 Chr.16: 57911692-57911692 on Build GRCh38
rs780585094 Chr.16: 57911910-57911910 on Build GRCh38
rs377762874 Chr.16: 57911872-57911872 on Build GRCh38
rs200323140 Chr.16: 57911793-57911793 on Build GRCh38
rs747258045 Chr.16: 57911807-57911807 on Build GRCh38
rs763344849 Chr.16: 57911866-57911866 on Build GRCh38
rs758363643 Chr.16: 57911894-57911894 on Build GRCh38
rs184343851 Chr.16: 57911734-57911734 on Build GRCh38
rs1024140088 Chr.16: 57911856-57911856 on Build GRCh38
rs776062884 Chr.16: 57911859-57911859 on Build GRCh38
rs778604026 Chr.16: 57911730-57911730 on Build GRCh38
rs758261344 Chr.16: 57911798-57911798 on Build GRCh38
rs535718543 Chr.16: 57911938-57911938 on Build GRCh38
rs750044654 Chr.16: 57911767-57911767 on Build GRCh38
rs775734464 Chr.16: 57911753-57911753 on Build GRCh38
rs774568659 Chr.16: 57911816-57911816 on Build GRCh38
rs200934249 Chr.16: 57911770-57911770 on Build GRCh38
rs752064224 Chr.16: 57911643-57911643 on Build GRCh38
rs189232287 Chr.16: 57911973-57911973 on Build GRCh38
rs151016462 Chr.16: 57911656-57911656 on Build GRCh38
rs201075437 Chr.16: 57911874-57911874 on Build GRCh38
rs754891107 Chr.16: 57911706-57911706 on Build GRCh38
rs760347343 Chr.16: 57911873-57911873 on Build GRCh38
rs376076728 Chr.16: 57911756-57911756 on Build GRCh38
rs372268628 Chr.16: 57911809-57911809 on Build GRCh38
rs774698647 Chr.16: 57911922-57911922 on Build GRCh38
rs763293997 Chr.16: 57911763-57911763 on Build GRCh38
rs762906693 Chr.16: 57911762-57911763 on Build GRCh38
rs754732440 Chr.16: 57911893-57911894 on Build GRCh38
rs570057689 Chr.16: 57911944-57911944 on Build GRCh38
rs559097172 Chr.16: 57911985-57911985 on Build GRCh38
rs548328739 Chr.16: 57911942-57911942 on Build GRCh38
rs563271089 Chr.16: 57911749-57911749 on Build GRCh38
rs77399988 Chr.16: 57911764-57911764 on Build GRCh38
rs181361909 Chr.16: 57911688-57911688 on Build GRCh38
rs374373659 Chr.16: 57911884-57911884 on Build GRCh38
rs1034133341 Chr.16: 57911769-57911769 on Build GRCh38
rs546742803 Chr.16: 57911909-57911909 on Build GRCh38
rs747127148 Chr.16: 57911740-57911740 on Build GRCh38
rs149722515 Chr.16: 57911915-57911915 on Build GRCh38
rs1049379058 Chr.16: 57911957-57911957 on Build GRCh38
rs746004352 Chr.16: 57911907-57911907 on Build GRCh38
rs189773427 Chr.16: 57911634-57911634 on Build GRCh38
rs778192245 Chr.16: 57911806-57911806 on Build GRCh38
rs374417317 Chr.16: 57911833-57911833 on Build GRCh38
rs747921487 Chr.16: 57911775-57911775 on Build GRCh38
rs751419074 Chr.16: 57911892-57911893 on Build GRCh38
rs766108478 Chr.16: 57911763-57911764 on Build GRCh38
rs753301003 Chr.16: 57911882-57911882 on Build GRCh38
rs1032798176 Chr.16: 57911890-57911890 on Build GRCh38
rs753193042 Chr.16: 57911654-57911654 on Build GRCh38
rs778126589 Chr.16: 57911735-57911735 on Build GRCh38
rs929535352 Chr.16: 57911930-57911930 on Build GRCh38
rs771113499 Chr.16: 57911745-57911745 on Build GRCh38
rs770265883 Chr.16: 57911853-57911853 on Build GRCh38
rs781467261 Chr.16: 57911906-57911906 on Build GRCh38
rs530382203 Chr.16: 57911693-57911693 on Build GRCh38
rs530551814 Chr.16: 57911752-57911752 on Build GRCh38
rs1041003408 Chr.16: 57911625-57911625 on Build GRCh38
rs781140236 Chr.16: 57911830-57911830 on Build GRCh38
rs751334014 Chr.16: 57911900-57911900 on Build GRCh38

View Full Product Details