Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 13608164-13608642 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGFL6
Entrez Gene ID
25975
Gene Name
EGF like domain multiple 6
Gene Aliases
MAEG, W80
UniGene
Hs.12844
Gene Chromosome Location
Chr.X: 13569575-13633575 on Build GRCh38
Transcripts
NM_001167890.1 NM_015507.3

SNP Detail

SNP ID Chromosome Location
rs984598582 Chr.X: 13608554-13608554 on Build GRCh38
rs966111722 Chr.X: 13608407-13608407 on Build GRCh38
rs749606751 Chr.X: 13608371-13608371 on Build GRCh38
rs753517471 Chr.X: 13608302-13608302 on Build GRCh38
rs776553348 Chr.X: 13608448-13608448 on Build GRCh38
rs199623111 Chr.X: 13608354-13608354 on Build GRCh38
rs773329040 Chr.X: 13608573-13608574 on Build GRCh38
rs773301882 Chr.X: 13608486-13608486 on Build GRCh38
rs140902386 Chr.X: 13608387-13608387 on Build GRCh38
rs16979025 Chr.X: 13608480-13608480 on Build GRCh38
rs781114200 Chr.X: 13608497-13608497 on Build GRCh38
rs763418340 Chr.X: 13608290-13608290 on Build GRCh38
rs771600453 Chr.X: 13608433-13608433 on Build GRCh38
rs761866445 Chr.X: 13608326-13608326 on Build GRCh38
rs760279727 Chr.X: 13608294-13608294 on Build GRCh38
rs771536175 Chr.X: 13608385-13608385 on Build GRCh38
rs1005921750 Chr.X: 13608570-13608570 on Build GRCh38
rs761478873 Chr.X: 13608456-13608456 on Build GRCh38
rs765821641 Chr.X: 13608254-13608254 on Build GRCh38
rs746475253 Chr.X: 13608445-13608445 on Build GRCh38
rs758105635 Chr.X: 13608370-13608370 on Build GRCh38
rs1034139872 Chr.X: 13608256-13608256 on Build GRCh38
rs751727031 Chr.X: 13608341-13608341 on Build GRCh38
rs755083555 Chr.X: 13608342-13608342 on Build GRCh38
rs112681548 Chr.X: 13608548-13608548 on Build GRCh38
rs1003015802 Chr.X: 13608245-13608245 on Build GRCh38
rs755305083 Chr.X: 13608196-13608196 on Build GRCh38
rs752317655 Chr.X: 13608349-13608349 on Build GRCh38
rs909057761 Chr.X: 13608555-13608555 on Build GRCh38
rs777612732 Chr.X: 13608426-13608426 on Build GRCh38
rs1010038522 Chr.X: 13608361-13608361 on Build GRCh38
rs936057618 Chr.X: 13608345-13608345 on Build GRCh38
rs961533468 Chr.X: 13608606-13608606 on Build GRCh38
rs765078838 Chr.X: 13608334-13608334 on Build GRCh38
rs143287924 Chr.X: 13608501-13608501 on Build GRCh38
rs2370236 Chr.X: 13608284-13608284 on Build GRCh38
rs774913289 Chr.X: 13608436-13608436 on Build GRCh38
rs907090559 Chr.X: 13608429-13608429 on Build GRCh38
rs1050169505 Chr.X: 13608280-13608280 on Build GRCh38
rs1034415304 Chr.X: 13608504-13608504 on Build GRCh38
rs760751484 Chr.X: 13608609-13608609 on Build GRCh38
rs753173608 Chr.X: 13608355-13608355 on Build GRCh38
rs769779163 Chr.X: 13608547-13608547 on Build GRCh38
rs760386505 Chr.X: 13608441-13608441 on Build GRCh38
rs940574545 Chr.X: 13608579-13608579 on Build GRCh38
rs376308744 Chr.X: 13608300-13608300 on Build GRCh38
rs762456228 Chr.X: 13608208-13608208 on Build GRCh38

View Full Product Details