Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 80653874-80654464 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHFR
Entrez Gene ID
1719
Gene Name
dihydrofolate reductase
Gene Aliases
DHFRP1, DYR
UniGene
Hs.592364
Gene Chromosome Location
Chr.5: 80626226-80654981 on Build GRCh38
Transcripts
NM_000791.3 NM_001290354.1 NM_001290357.1 NR_110936.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5817660 c.45G>A p.M15I Chr.5: 80654445-80654445 on Build GRCh38
3947752 c.98G>C p.R33T Chr.5: 80654046-80654046 on Build GRCh38
5996285 c.76C>A p.P26T Chr.5: 80654414-80654414 on Build GRCh38
6326465 c.136+9G>A p.? Chr.5: 80653999-80653999 on Build GRCh38
6402013 c.75G>T p.W25C Chr.5: 80654415-80654415 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs572081813 Chr.5: 80654068-80654068 on Build GRCh38
rs778172287 Chr.5: 80654077-80654077 on Build GRCh38
rs368073490 Chr.5: 80654367-80654367 on Build GRCh38
rs781302806 Chr.5: 80654361-80654361 on Build GRCh38
rs878952102 Chr.5: 80654299-80654299 on Build GRCh38
rs559411604 Chr.5: 80654226-80654226 on Build GRCh38
rs556862773 Chr.5: 80653905-80653905 on Build GRCh38
rs773136060 Chr.5: 80654045-80654045 on Build GRCh38
rs188843682 Chr.5: 80653926-80653926 on Build GRCh38
rs953635799 Chr.5: 80653907-80653907 on Build GRCh38
rs767425256 Chr.5: 80654038-80654038 on Build GRCh38
rs371588943 Chr.5: 80654423-80654423 on Build GRCh38
rs769655762 Chr.5: 80654410-80654412 on Build GRCh38
rs752194110 Chr.5: 80654424-80654424 on Build GRCh38
rs935641131 Chr.5: 80654276-80654276 on Build GRCh38
rs926061488 Chr.5: 80654295-80654295 on Build GRCh38
rs762459617 Chr.5: 80653998-80653998 on Build GRCh38
rs771048956 Chr.5: 80653968-80653968 on Build GRCh38
rs773189076 Chr.5: 80654390-80654390 on Build GRCh38
rs761687618 Chr.5: 80653981-80653982 on Build GRCh38
rs773005169 Chr.5: 80654444-80654445 on Build GRCh38
rs769778564 Chr.5: 80654368-80654368 on Build GRCh38
rs758957010 Chr.5: 80654425-80654425 on Build GRCh38
rs924288999 Chr.5: 80654275-80654275 on Build GRCh38
rs965218942 Chr.5: 80654123-80654126 on Build GRCh38
rs1054100832 Chr.5: 80654085-80654085 on Build GRCh38
rs993049221 Chr.5: 80654189-80654189 on Build GRCh38
rs766155926 Chr.5: 80654395-80654395 on Build GRCh38
rs752068745 Chr.5: 80654442-80654442 on Build GRCh38
rs1019222419 Chr.5: 80654261-80654261 on Build GRCh38
rs775644985 Chr.5: 80654371-80654371 on Build GRCh38
rs753426178 Chr.5: 80654060-80654060 on Build GRCh38
rs747303120 Chr.5: 80653964-80653964 on Build GRCh38
rs533184784 Chr.5: 80654134-80654134 on Build GRCh38
rs986684826 Chr.5: 80654217-80654217 on Build GRCh38
rs1011918306 Chr.5: 80654281-80654281 on Build GRCh38
rs747316557 Chr.5: 80654356-80654356 on Build GRCh38
rs776823823 Chr.5: 80653981-80653981 on Build GRCh38
rs191745317 Chr.5: 80654146-80654146 on Build GRCh38
rs562836282 Chr.5: 80654129-80654129 on Build GRCh38
rs555346066 Chr.5: 80654061-80654061 on Build GRCh38
rs1044887788 Chr.5: 80653945-80653945 on Build GRCh38
rs764674335 Chr.5: 80654062-80654062 on Build GRCh38
rs70991108 Chr.5: 80654344-80654345 on Build GRCh38
rs61736208 Chr.5: 80654438-80654438 on Build GRCh38
rs868450990 Chr.5: 80654343-80654344 on Build GRCh38
rs1009457092 Chr.5: 80654353-80654353 on Build GRCh38
rs184211767 Chr.5: 80654151-80654151 on Build GRCh38
rs201745474 Chr.5: 80654054-80654054 on Build GRCh38
rs1027710637 Chr.5: 80654172-80654172 on Build GRCh38
rs977604508 Chr.5: 80654271-80654271 on Build GRCh38
rs944411304 Chr.5: 80653935-80653935 on Build GRCh38
rs200822257 Chr.5: 80654360-80654360 on Build GRCh38
rs908795833 Chr.5: 80654158-80654158 on Build GRCh38
rs769880102 Chr.5: 80653999-80653999 on Build GRCh38
rs936010608 Chr.5: 80654309-80654310 on Build GRCh38
rs1032250222 Chr.5: 80654011-80654011 on Build GRCh38
rs966687152 Chr.5: 80654270-80654270 on Build GRCh38
rs549250404 Chr.5: 80654214-80654214 on Build GRCh38
rs1053151573 Chr.5: 80654333-80654333 on Build GRCh38
rs994929672 Chr.5: 80654154-80654154 on Build GRCh38
rs948539157 Chr.5: 80654108-80654108 on Build GRCh38
rs776167521 Chr.5: 80654396-80654396 on Build GRCh38
rs891808441 Chr.5: 80654350-80654350 on Build GRCh38
rs1019423802 Chr.5: 80654078-80654078 on Build GRCh38
rs961568251 Chr.5: 80654188-80654188 on Build GRCh38
rs894323747 Chr.5: 80654096-80654096 on Build GRCh38
rs144847410 Chr.5: 80653988-80653988 on Build GRCh38
rs904499589 Chr.5: 80653967-80653967 on Build GRCh38
rs953590305 Chr.5: 80654181-80654181 on Build GRCh38
rs939918888 Chr.5: 80654400-80654400 on Build GRCh38
rs761785469 Chr.5: 80654025-80654025 on Build GRCh38
rs765924078 Chr.5: 80654055-80654055 on Build GRCh38
rs531484931 Chr.5: 80654401-80654401 on Build GRCh38
rs1057270142 Chr.5: 80653904-80653906 on Build GRCh38
rs753212015 Chr.5: 80654064-80654064 on Build GRCh38
rs895991162 Chr.5: 80653919-80653919 on Build GRCh38
rs774452953 Chr.5: 80654021-80654021 on Build GRCh38
rs6151593 Chr.5: 80653984-80653984 on Build GRCh38
rs868777143 Chr.5: 80654318-80654318 on Build GRCh38
rs778049706 Chr.5: 80653958-80653958 on Build GRCh38
rs989878569 Chr.5: 80654303-80654303 on Build GRCh38
rs539629126 Chr.5: 80653901-80653901 on Build GRCh38
rs541144933 Chr.5: 80654117-80654117 on Build GRCh38
rs912105347 Chr.5: 80653933-80653933 on Build GRCh38
rs6151594 Chr.5: 80654147-80654147 on Build GRCh38
rs760354608 Chr.5: 80654394-80654394 on Build GRCh38
rs748163318 Chr.5: 80654379-80654379 on Build GRCh38

View Full Product Details