Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 154002262-154002849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM114A2
Entrez Gene ID
10827
Gene Name
family with sequence similarity 114 member A2
Gene Aliases
133K02, C5orf3
UniGene
Hs.166551
Gene Chromosome Location
Chr.5: 153990131-154038938 on Build GRCh38
Transcripts
NM_001317993.1 NM_001317994.1 NM_001317995.1 NM_018691.3 XM_005268360.4 XM_005268361.4 XM_011537540.2 XM_011537541.2 XM_011537542.2 XM_017008967.1 XM_017008968.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4383384 c.1115A>C p.E372A Chr.5: 154002848-154002848 on Build GRCh38
160883 c.1179C>A p.F393L Chr.5: 154002328-154002328 on Build GRCh38
7144756 c.1155G>C p.L385L Chr.5: 154002352-154002352 on Build GRCh38
7144755 c.1239G>C p.R413S Chr.5: 154002268-154002268 on Build GRCh38
5697182 c.1155G>A p.L385L Chr.5: 154002352-154002352 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs918599750 Chr.5: 154002440-154002440 on Build GRCh38
rs374460608 Chr.5: 154002376-154002376 on Build GRCh38
rs766099452 Chr.5: 154002379-154002379 on Build GRCh38
rs35121544 Chr.5: 154002521-154002523 on Build GRCh38
rs775588851 Chr.5: 154002365-154002365 on Build GRCh38
rs778482580 Chr.5: 154002431-154002431 on Build GRCh38
rs761767529 Chr.5: 154002368-154002368 on Build GRCh38
rs371964497 Chr.5: 154002822-154002822 on Build GRCh38
rs373865452 Chr.5: 154002796-154002796 on Build GRCh38
rs1016436748 Chr.5: 154002322-154002322 on Build GRCh38
rs914488437 Chr.5: 154002494-154002494 on Build GRCh38
rs955812890 Chr.5: 154002685-154002685 on Build GRCh38
rs973708154 Chr.5: 154002389-154002389 on Build GRCh38
rs968553522 Chr.5: 154002739-154002739 on Build GRCh38
rs761041894 Chr.5: 154002361-154002361 on Build GRCh38
rs370862982 Chr.5: 154002370-154002370 on Build GRCh38
rs1004999860 Chr.5: 154002636-154002636 on Build GRCh38
rs561088405 Chr.5: 154002501-154002501 on Build GRCh38
rs749806938 Chr.5: 154002432-154002432 on Build GRCh38
rs780697366 Chr.5: 154002301-154002301 on Build GRCh38
rs777577975 Chr.5: 154002575-154002575 on Build GRCh38
rs1017094618 Chr.5: 154002404-154002405 on Build GRCh38
rs11748836 Chr.5: 154002288-154002288 on Build GRCh38
rs571269502 Chr.5: 154002606-154002606 on Build GRCh38
rs74196375 Chr.5: 154002438-154002438 on Build GRCh38
rs939303592 Chr.5: 154002457-154002457 on Build GRCh38
rs1025859723 Chr.5: 154002773-154002773 on Build GRCh38
rs542838981 Chr.5: 154002459-154002459 on Build GRCh38
rs760462959 Chr.5: 154002356-154002371 on Build GRCh38
rs766830399 Chr.5: 154002395-154002395 on Build GRCh38
rs766970633 Chr.5: 154002396-154002396 on Build GRCh38
rs183682516 Chr.5: 154002425-154002425 on Build GRCh38
rs766334340 Chr.5: 154002376-154002376 on Build GRCh38
rs774174048 Chr.5: 154002543-154002543 on Build GRCh38
rs953856114 Chr.5: 154002424-154002424 on Build GRCh38
rs775272637 Chr.5: 154002380-154002380 on Build GRCh38
rs988675395 Chr.5: 154002463-154002463 on Build GRCh38
rs778342546 Chr.5: 154002500-154002500 on Build GRCh38
rs1006582191 Chr.5: 154002718-154002718 on Build GRCh38
rs111455860 Chr.5: 154002699-154002699 on Build GRCh38
rs905836929 Chr.5: 154002553-154002553 on Build GRCh38
rs535298331 Chr.5: 154002704-154002704 on Build GRCh38
rs532292043 Chr.5: 154002639-154002639 on Build GRCh38
rs927205186 Chr.5: 154002753-154002753 on Build GRCh38
rs754382121 Chr.5: 154002405-154002405 on Build GRCh38
rs754454894 Chr.5: 154002292-154002292 on Build GRCh38
rs202232335 Chr.5: 154002804-154002804 on Build GRCh38
rs755574736 Chr.5: 154002726-154002726 on Build GRCh38
rs2560060 Chr.5: 154002564-154002564 on Build GRCh38
rs1057450743 Chr.5: 154002821-154002821 on Build GRCh38
rs1008315666 Chr.5: 154002675-154002675 on Build GRCh38
rs774503196 Chr.5: 154002345-154002345 on Build GRCh38
rs980020501 Chr.5: 154002752-154002752 on Build GRCh38
rs953696800 Chr.5: 154002670-154002670 on Build GRCh38
rs759662984 Chr.5: 154002820-154002821 on Build GRCh38
rs765196839 Chr.5: 154002369-154002369 on Build GRCh38
rs138263333 Chr.5: 154002313-154002313 on Build GRCh38
rs753922199 Chr.5: 154002438-154002441 on Build GRCh38
rs746756847 Chr.5: 154002585-154002585 on Build GRCh38
rs189120262 Chr.5: 154002786-154002786 on Build GRCh38
rs771409700 Chr.5: 154002339-154002339 on Build GRCh38
rs35082504 Chr.5: 154002513-154002515 on Build GRCh38
rs545649031 Chr.5: 154002387-154002387 on Build GRCh38
rs960701369 Chr.5: 154002778-154002778 on Build GRCh38
rs778396391 Chr.5: 154002323-154002323 on Build GRCh38
rs1057178024 Chr.5: 154002574-154002574 on Build GRCh38
rs373853052 Chr.5: 154002624-154002625 on Build GRCh38
rs1044779628 Chr.5: 154002529-154002529 on Build GRCh38
rs145160134 Chr.5: 154002659-154002659 on Build GRCh38
rs947309302 Chr.5: 154002527-154002527 on Build GRCh38
rs114020061 Chr.5: 154002660-154002660 on Build GRCh38
rs935940758 Chr.5: 154002582-154002583 on Build GRCh38
rs773106930 Chr.5: 154002372-154002372 on Build GRCh38
rs74775091 Chr.5: 154002658-154002658 on Build GRCh38
rs749719044 Chr.5: 154002324-154002324 on Build GRCh38
rs149647426 Chr.5: 154002650-154002650 on Build GRCh38
rs772056516 Chr.5: 154002360-154002360 on Build GRCh38
rs528516213 Chr.5: 154002547-154002547 on Build GRCh38
rs1053019556 Chr.5: 154002620-154002620 on Build GRCh38
rs746035433 Chr.5: 154002355-154002355 on Build GRCh38
rs765593629 Chr.5: 154002821-154002821 on Build GRCh38
rs12110144 Chr.5: 154002738-154002738 on Build GRCh38
rs1019352639 Chr.5: 154002681-154002681 on Build GRCh38

View Full Product Details