Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 207089075-207089674 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4BPB
Entrez Gene ID
725
Gene Name
complement component 4 binding protein beta
Gene Aliases
C4BP
UniGene
Hs.99886
Gene Chromosome Location
Chr.1: 207088826-207099993 on Build GRCh38
Transcripts
NM_000716.3 NM_001017364.1 NM_001017365.2 NM_001017366.2 NM_001017367.1 XM_005273254.4 XM_005273255.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3864315 c.41G>A p.R14Q Chr.1: 207089572-207089572 on Build GRCh38
6500660 c.9G>C p.A3A Chr.1: 207089567-207089567 on Build GRCh38
6295615 c.18G>T p.A6A Chr.1: 207089549-207089549 on Build GRCh38
1601594 c.3_4insT p.W4fs*17 Chr.1: 207089534-207089535 on Build GRCh38
903088 c.40C>T p.R14* Chr.1: 207089571-207089571 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779025858 Chr.1: 207089579-207089579 on Build GRCh38
rs539172734 Chr.1: 207089390-207089390 on Build GRCh38
rs894840074 Chr.1: 207089534-207089534 on Build GRCh38
rs779133566 Chr.1: 207089635-207089635 on Build GRCh38
rs769234807 Chr.1: 207089357-207089357 on Build GRCh38
rs990678935 Chr.1: 207089418-207089418 on Build GRCh38
rs1004689013 Chr.1: 207089572-207089572 on Build GRCh38
rs921744442 Chr.1: 207089354-207089354 on Build GRCh38
rs1013853792 Chr.1: 207089461-207089461 on Build GRCh38
rs909280216 Chr.1: 207089242-207089242 on Build GRCh38
rs146846149 Chr.1: 207089566-207089566 on Build GRCh38
rs1052511 Chr.1: 207089305-207089305 on Build GRCh38
rs763015262 Chr.1: 207089417-207089417 on Build GRCh38
rs575077693 Chr.1: 207089604-207089604 on Build GRCh38
rs774906117 Chr.1: 207089535-207089535 on Build GRCh38
rs188656684 Chr.1: 207089567-207089567 on Build GRCh38
rs762929016 Chr.1: 207089542-207089542 on Build GRCh38
rs369369078 Chr.1: 207089401-207089401 on Build GRCh38
rs959593595 Chr.1: 207089127-207089127 on Build GRCh38
rs367552925 Chr.1: 207089619-207089619 on Build GRCh38
rs773791430 Chr.1: 207089494-207089494 on Build GRCh38
rs1025608224 Chr.1: 207089471-207089471 on Build GRCh38
rs12046484 Chr.1: 207089101-207089101 on Build GRCh38
rs558739747 Chr.1: 207089180-207089180 on Build GRCh38
rs745487237 Chr.1: 207089626-207089626 on Build GRCh38
rs541195698 Chr.1: 207089362-207089362 on Build GRCh38
rs201791990 Chr.1: 207089554-207089554 on Build GRCh38
rs1055175640 Chr.1: 207089208-207089208 on Build GRCh38
rs778923199 Chr.1: 207089484-207089484 on Build GRCh38
rs371525248 Chr.1: 207089549-207089549 on Build GRCh38
rs79737422 Chr.1: 207089446-207089446 on Build GRCh38
rs776365834 Chr.1: 207089525-207089525 on Build GRCh38
rs17020462 Chr.1: 207089266-207089266 on Build GRCh38
rs1003224261 Chr.1: 207089116-207089116 on Build GRCh38
rs755035584 Chr.1: 207089482-207089482 on Build GRCh38
rs774255769 Chr.1: 207089593-207089593 on Build GRCh38
rs751515179 Chr.1: 207089560-207089560 on Build GRCh38
rs77632202 Chr.1: 207089466-207089466 on Build GRCh38
rs370014026 Chr.1: 207089423-207089423 on Build GRCh38
rs568269279 Chr.1: 207089134-207089134 on Build GRCh38
rs994512193 Chr.1: 207089552-207089552 on Build GRCh38
rs55754417 Chr.1: 207089459-207089459 on Build GRCh38
rs776225122 Chr.1: 207089295-207089295 on Build GRCh38
rs771428087 Chr.1: 207089511-207089511 on Build GRCh38
rs748127905 Chr.1: 207089590-207089590 on Build GRCh38
rs758536456 Chr.1: 207089594-207089594 on Build GRCh38
rs775234519 Chr.1: 207089640-207089640 on Build GRCh38
rs754051271 Chr.1: 207089571-207089571 on Build GRCh38
rs537251347 Chr.1: 207089262-207089262 on Build GRCh38
rs371751934 Chr.1: 207089456-207089456 on Build GRCh38
rs1018829528 Chr.1: 207089320-207089320 on Build GRCh38
rs771409897 Chr.1: 207089605-207089605 on Build GRCh38
rs772503090 Chr.1: 207089493-207089493 on Build GRCh38
rs771089497 Chr.1: 207089594-207089595 on Build GRCh38
rs992387850 Chr.1: 207089137-207089137 on Build GRCh38
rs532435531 Chr.1: 207089114-207089114 on Build GRCh38
rs950742623 Chr.1: 207089157-207089157 on Build GRCh38
rs937537028 Chr.1: 207089473-207089473 on Build GRCh38
rs12711513 Chr.1: 207089635-207089635 on Build GRCh38
rs965909471 Chr.1: 207089361-207089361 on Build GRCh38
rs1027400421 Chr.1: 207089284-207089284 on Build GRCh38
rs904259568 Chr.1: 207089436-207089436 on Build GRCh38
rs140157538 Chr.1: 207089533-207089533 on Build GRCh38
rs532362224 Chr.1: 207089541-207089541 on Build GRCh38
rs917938102 Chr.1: 207089138-207089138 on Build GRCh38
rs759321706 Chr.1: 207089526-207089526 on Build GRCh38
rs748656391 Chr.1: 207089490-207089490 on Build GRCh38
rs778229281 Chr.1: 207089623-207089623 on Build GRCh38
rs762334688 Chr.1: 207089641-207089641 on Build GRCh38

View Full Product Details