Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 33709629-33710207 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD59
Entrez Gene ID
966
Gene Name
CD59 molecule
Gene Aliases
16.3A5, 1F5, EJ16, EJ30, EL32, G344, HRF-20, HRF20, MAC-IP, MACIF, MEM43, MIC11, MIN1, MIN2, MIN3, MIRL, MSK21, p18-20
UniGene
Hs.278573
Gene Chromosome Location
Chr.11: 33703010-33736479 on Build GRCh38
Transcripts
NM_000611.5 NM_001127223.1 NM_001127225.1 NM_001127226.1 NM_001127227.1 NM_203329.2 NM_203330.2 NM_203331.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6346243 c.337C>A p.L113I Chr.11: 33710176-33710176 on Build GRCh38
3986132 c.307G>A p.G103S Chr.11: 33710206-33710206 on Build GRCh38
6231875 c.309T>C p.G103G Chr.11: 33710204-33710204 on Build GRCh38
6623837 c.321A>G p.L107L Chr.11: 33710192-33710192 on Build GRCh38
926550 c.374G>A p.S125N Chr.11: 33710139-33710139 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs12136 Chr.11: 33710078-33710078 on Build GRCh38
rs953314397 Chr.11: 33709923-33709923 on Build GRCh38
rs41275158 Chr.11: 33709786-33709786 on Build GRCh38
rs891173064 Chr.11: 33709746-33709746 on Build GRCh38
rs1020407889 Chr.11: 33709884-33709884 on Build GRCh38
rs560195222 Chr.11: 33710114-33710114 on Build GRCh38
rs979186125 Chr.11: 33709667-33709667 on Build GRCh38
rs563206374 Chr.11: 33710171-33710171 on Build GRCh38
rs1029418330 Chr.11: 33709922-33709922 on Build GRCh38
rs921850877 Chr.11: 33709708-33709708 on Build GRCh38
rs763058880 Chr.11: 33710150-33710150 on Build GRCh38
rs369200735 Chr.11: 33710131-33710131 on Build GRCh38
rs764428686 Chr.11: 33710172-33710174 on Build GRCh38
rs897115390 Chr.11: 33710046-33710046 on Build GRCh38
rs370698576 Chr.11: 33710084-33710084 on Build GRCh38
rs911761540 Chr.11: 33709724-33709724 on Build GRCh38
rs772773185 Chr.11: 33709873-33709873 on Build GRCh38
rs969228502 Chr.11: 33709894-33709894 on Build GRCh38
rs987388631 Chr.11: 33709941-33709941 on Build GRCh38
rs541468361 Chr.11: 33709970-33709970 on Build GRCh38
rs1006979869 Chr.11: 33709758-33709758 on Build GRCh38
rs572909545 Chr.11: 33709863-33709863 on Build GRCh38
rs765997205 Chr.11: 33710013-33710013 on Build GRCh38
rs181012235 Chr.11: 33710077-33710077 on Build GRCh38
rs774419867 Chr.11: 33709790-33709790 on Build GRCh38
rs987233168 Chr.11: 33709723-33709723 on Build GRCh38
rs11552626 Chr.11: 33710144-33710144 on Build GRCh38
rs745640190 Chr.11: 33710088-33710088 on Build GRCh38
rs758222204 Chr.11: 33710158-33710158 on Build GRCh38
rs113409956 Chr.11: 33709900-33709900 on Build GRCh38
rs1001793974 Chr.11: 33709845-33709845 on Build GRCh38
rs77171493 Chr.11: 33709853-33709853 on Build GRCh38
rs842 Chr.11: 33709860-33709860 on Build GRCh38
rs978748036 Chr.11: 33709914-33709914 on Build GRCh38
rs561306801 Chr.11: 33710112-33710112 on Build GRCh38
rs371621250 Chr.11: 33710110-33710110 on Build GRCh38
rs956907618 Chr.11: 33710163-33710163 on Build GRCh38
rs778770889 Chr.11: 33710085-33710085 on Build GRCh38
rs778646046 Chr.11: 33710148-33710148 on Build GRCh38
rs750135648 Chr.11: 33710157-33710157 on Build GRCh38
rs929968944 Chr.11: 33710062-33710062 on Build GRCh38
rs949799297 Chr.11: 33709937-33709937 on Build GRCh38
rs781050268 Chr.11: 33710183-33710183 on Build GRCh38
rs930297136 Chr.11: 33709813-33709813 on Build GRCh38
rs1018836756 Chr.11: 33709771-33709771 on Build GRCh38
rs774842026 Chr.11: 33710045-33710045 on Build GRCh38
rs555679428 Chr.11: 33710021-33710021 on Build GRCh38
rs1045147691 Chr.11: 33709833-33709833 on Build GRCh38
rs775255324 Chr.11: 33710092-33710092 on Build GRCh38
rs1031283167 Chr.11: 33709834-33709834 on Build GRCh38
rs768526855 Chr.11: 33710104-33710104 on Build GRCh38
rs372984787 Chr.11: 33710176-33710176 on Build GRCh38
rs746821921 Chr.11: 33710175-33710175 on Build GRCh38
rs549099946 Chr.11: 33710129-33710129 on Build GRCh38
rs375331661 Chr.11: 33710089-33710089 on Build GRCh38
rs41275160 Chr.11: 33709855-33709855 on Build GRCh38
rs368593192 Chr.11: 33710093-33710093 on Build GRCh38
rs2231461 Chr.11: 33709999-33709999 on Build GRCh38
rs753381105 Chr.11: 33710138-33710138 on Build GRCh38
rs1010305212 Chr.11: 33709882-33709882 on Build GRCh38
rs1029741879 Chr.11: 33709683-33709683 on Build GRCh38
rs191085190 Chr.11: 33709661-33709661 on Build GRCh38
rs979893418 Chr.11: 33709952-33709952 on Build GRCh38
rs11552628 Chr.11: 33710056-33710056 on Build GRCh38
rs866243991 Chr.11: 33710161-33710161 on Build GRCh38
rs764547290 Chr.11: 33710133-33710133 on Build GRCh38
rs968259373 Chr.11: 33709785-33709785 on Build GRCh38
rs759828865 Chr.11: 33710118-33710118 on Build GRCh38
rs192111748 Chr.11: 33710121-33710121 on Build GRCh38

View Full Product Details