Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 55967870-55968410 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK2
Entrez Gene ID
1017
Gene Name
cyclin dependent kinase 2
Gene Aliases
CDKN2, p33(CDK2)
UniGene
Hs.19192
Gene Chromosome Location
Chr.12: 55966769-55972789 on Build GRCh38
Transcripts
NM_001290230.1 NM_001798.4 NM_052827.3 XM_011537732.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1362966 c.250C>T p.H84Y Chr.12: 55968104-55968104 on Build GRCh38
6306063 c.249G>T p.L83L Chr.12: 55968103-55968103 on Build GRCh38
6227644 c.194+10C>T p.? Chr.12: 55967944-55967944 on Build GRCh38
3792818 c.202G>A p.D68N Chr.12: 55968056-55968056 on Build GRCh38
1200555 c.266A>C p.K89T Chr.12: 55968120-55968120 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761831055 Chr.12: 55968205-55968205 on Build GRCh38
rs1048010267 Chr.12: 55968201-55968201 on Build GRCh38
rs746270101 Chr.12: 55967977-55967977 on Build GRCh38
rs774473582 Chr.12: 55968012-55968012 on Build GRCh38
rs1000506207 Chr.12: 55967895-55967895 on Build GRCh38
rs984124329 Chr.12: 55968294-55968294 on Build GRCh38
rs749001657 Chr.12: 55967944-55967944 on Build GRCh38
rs757201758 Chr.12: 55967909-55967909 on Build GRCh38
rs757834416 Chr.12: 55968364-55968364 on Build GRCh38
rs1011991015 Chr.12: 55968224-55968224 on Build GRCh38
rs977207520 Chr.12: 55968329-55968332 on Build GRCh38
rs922686627 Chr.12: 55968339-55968339 on Build GRCh38
rs767614315 Chr.12: 55968036-55968037 on Build GRCh38
rs772956348 Chr.12: 55968217-55968217 on Build GRCh38
rs750222849 Chr.12: 55967914-55967914 on Build GRCh38
rs747502900 Chr.12: 55967962-55967962 on Build GRCh38
rs373277065 Chr.12: 55968203-55968203 on Build GRCh38
rs763241768 Chr.12: 55967999-55967999 on Build GRCh38
rs780953386 Chr.12: 55967911-55967911 on Build GRCh38
rs943979756 Chr.12: 55967972-55967972 on Build GRCh38
rs371108331 Chr.12: 55967950-55967950 on Build GRCh38
rs757819756 Chr.12: 55968138-55968138 on Build GRCh38
rs774390450 Chr.12: 55968204-55968204 on Build GRCh38
rs147314596 Chr.12: 55968103-55968103 on Build GRCh38
rs11554375 Chr.12: 55968060-55968060 on Build GRCh38
rs369071236 Chr.12: 55968013-55968013 on Build GRCh38
rs754439865 Chr.12: 55968316-55968316 on Build GRCh38
rs777107767 Chr.12: 55967973-55967973 on Build GRCh38
rs200294932 Chr.12: 55968207-55968207 on Build GRCh38
rs544826452 Chr.12: 55968143-55968143 on Build GRCh38
rs779522936 Chr.12: 55967934-55967934 on Build GRCh38
rs767940603 Chr.12: 55968383-55968383 on Build GRCh38
rs372713896 Chr.12: 55967918-55967918 on Build GRCh38
rs766197388 Chr.12: 55968063-55968063 on Build GRCh38
rs376663678 Chr.12: 55967945-55967945 on Build GRCh38
rs551123688 Chr.12: 55968139-55968139 on Build GRCh38
rs75012426 Chr.12: 55968376-55968376 on Build GRCh38
rs755767646 Chr.12: 55967926-55967926 on Build GRCh38
rs539865729 Chr.12: 55968327-55968327 on Build GRCh38
rs778560038 Chr.12: 55968106-55968106 on Build GRCh38
rs754843910 Chr.12: 55968018-55968019 on Build GRCh38
rs769987474 Chr.12: 55968173-55968173 on Build GRCh38
rs775538312 Chr.12: 55967983-55967983 on Build GRCh38
rs747703071 Chr.12: 55968107-55968107 on Build GRCh38
rs377117205 Chr.12: 55968031-55968031 on Build GRCh38
rs186450393 Chr.12: 55968182-55968182 on Build GRCh38
rs867970674 Chr.12: 55968288-55968288 on Build GRCh38
rs770219786 Chr.12: 55967980-55967980 on Build GRCh38
rs1036895432 Chr.12: 55968016-55968016 on Build GRCh38
rs749459750 Chr.12: 55968184-55968184 on Build GRCh38
rs552071819 Chr.12: 55968330-55968330 on Build GRCh38
rs372922692 Chr.12: 55968017-55968017 on Build GRCh38
rs113713715 Chr.12: 55968042-55968042 on Build GRCh38
rs764103884 Chr.12: 55968010-55968010 on Build GRCh38
rs755825258 Chr.12: 55968033-55968033 on Build GRCh38
rs369617221 Chr.12: 55968090-55968090 on Build GRCh38
rs771630413 Chr.12: 55967967-55967967 on Build GRCh38
rs908577230 Chr.12: 55968297-55968297 on Build GRCh38
rs906684364 Chr.12: 55968347-55968347 on Build GRCh38
rs1002329912 Chr.12: 55968053-55968053 on Build GRCh38
rs746422426 Chr.12: 55968140-55968140 on Build GRCh38

View Full Product Details