Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 165657658-165658207 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSRNP3
Entrez Gene ID
80034
Gene Name
cysteine and serine rich nuclear protein 3
Gene Aliases
FAM130A2, PPP1R73, TAIP-2, TAIP2
UniGene
Hs.470479
Gene Chromosome Location
Chr.2: 165469647-165689407 on Build GRCh38
Transcripts
NM_001172173.1 NM_024969.3 XM_005246865.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1305791 c.155C>G p.S52C Chr.2: 165657767-165657767 on Build GRCh38
6335528 c.345G>T p.R115S Chr.2: 165657957-165657957 on Build GRCh38
4593628 c.271A>C p.T91P Chr.2: 165657883-165657883 on Build GRCh38
3895165 c.266G>A p.G89E Chr.2: 165657878-165657878 on Build GRCh38
4594371 c.275T>G p.L92R Chr.2: 165657887-165657887 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371176147 Chr.2: 165658162-165658162 on Build GRCh38
rs550668645 Chr.2: 165658120-165658120 on Build GRCh38
rs768975428 Chr.2: 165657740-165657740 on Build GRCh38
rs1034365522 Chr.2: 165658116-165658116 on Build GRCh38
rs763933992 Chr.2: 165657836-165657836 on Build GRCh38
rs182598681 Chr.2: 165658129-165658129 on Build GRCh38
rs753342620 Chr.2: 165657745-165657748 on Build GRCh38
rs763122842 Chr.2: 165657938-165657938 on Build GRCh38
rs374214581 Chr.2: 165657917-165657917 on Build GRCh38
rs552887047 Chr.2: 165658029-165658029 on Build GRCh38
rs748873004 Chr.2: 165657900-165657900 on Build GRCh38
rs548230415 Chr.2: 165657729-165657729 on Build GRCh38
rs373080590 Chr.2: 165658103-165658103 on Build GRCh38
rs756887499 Chr.2: 165657720-165657720 on Build GRCh38
rs749713008 Chr.2: 165658005-165658005 on Build GRCh38
rs769304262 Chr.2: 165657769-165657769 on Build GRCh38
rs748780351 Chr.2: 165657695-165657695 on Build GRCh38
rs747421770 Chr.2: 165657728-165657728 on Build GRCh38
rs189894482 Chr.2: 165657940-165657940 on Build GRCh38
rs778125869 Chr.2: 165657899-165657899 on Build GRCh38
rs752699378 Chr.2: 165657825-165657825 on Build GRCh38
rs199902919 Chr.2: 165657822-165657822 on Build GRCh38
rs764142469 Chr.2: 165657927-165657927 on Build GRCh38
rs142720587 Chr.2: 165657913-165657913 on Build GRCh38
rs201590164 Chr.2: 165658064-165658064 on Build GRCh38
rs374031933 Chr.2: 165658052-165658052 on Build GRCh38
rs770491603 Chr.2: 165658028-165658028 on Build GRCh38
rs370657022 Chr.2: 165657844-165657844 on Build GRCh38
rs777271740 Chr.2: 165657692-165657692 on Build GRCh38
rs760560179 Chr.2: 165658054-165658054 on Build GRCh38
rs10803800 Chr.2: 165658084-165658084 on Build GRCh38
rs150706497 Chr.2: 165657704-165657704 on Build GRCh38
rs572430682 Chr.2: 165658145-165658146 on Build GRCh38
rs903342988 Chr.2: 165658023-165658023 on Build GRCh38
rs756410608 Chr.2: 165657979-165657979 on Build GRCh38
rs537049441 Chr.2: 165658113-165658113 on Build GRCh38
rs144570009 Chr.2: 165657876-165657876 on Build GRCh38
rs776651542 Chr.2: 165658065-165658065 on Build GRCh38
rs766111079 Chr.2: 165657788-165657788 on Build GRCh38
rs200315219 Chr.2: 165657912-165657912 on Build GRCh38
rs146855337 Chr.2: 165657899-165657900 on Build GRCh38
rs557062715 Chr.2: 165658158-165658158 on Build GRCh38
rs756203845 Chr.2: 165658083-165658083 on Build GRCh38
rs755121781 Chr.2: 165657954-165657954 on Build GRCh38
rs78315589 Chr.2: 165657805-165657805 on Build GRCh38
rs745366796 Chr.2: 165658043-165658043 on Build GRCh38
rs777153783 Chr.2: 165657742-165657742 on Build GRCh38
rs947608891 Chr.2: 165657990-165657990 on Build GRCh38
rs773848096 Chr.2: 165657789-165657789 on Build GRCh38
rs759438129 Chr.2: 165657813-165657813 on Build GRCh38
rs778377776 Chr.2: 165657980-165657980 on Build GRCh38
rs368420776 Chr.2: 165657942-165657942 on Build GRCh38
rs761846158 Chr.2: 165658159-165658159 on Build GRCh38
rs753115769 Chr.2: 165657965-165657965 on Build GRCh38
rs748604034 Chr.2: 165657745-165657745 on Build GRCh38
rs770397223 Chr.2: 165657895-165657895 on Build GRCh38
rs560723927 Chr.2: 165657787-165657787 on Build GRCh38
rs755358554 Chr.2: 165657883-165657883 on Build GRCh38
rs76022415 Chr.2: 165658104-165658104 on Build GRCh38
rs780127357 Chr.2: 165657726-165657726 on Build GRCh38
rs950696447 Chr.2: 165658107-165658107 on Build GRCh38
rs11689899 Chr.2: 165657989-165657989 on Build GRCh38
rs563868095 Chr.2: 165657916-165657916 on Build GRCh38
rs753686562 Chr.2: 165657936-165657936 on Build GRCh38
rs772708499 Chr.2: 165657779-165657779 on Build GRCh38
rs960090346 Chr.2: 165658125-165658125 on Build GRCh38
rs868555198 Chr.2: 165658166-165658166 on Build GRCh38
rs781473977 Chr.2: 165657885-165657885 on Build GRCh38
rs748654810 Chr.2: 165657887-165657887 on Build GRCh38
rs1001988321 Chr.2: 165658115-165658115 on Build GRCh38
rs111310428 Chr.2: 165657959-165657959 on Build GRCh38
rs1050098480 Chr.2: 165658025-165658025 on Build GRCh38
rs770309060 Chr.2: 165657901-165657901 on Build GRCh38
rs774078840 Chr.2: 165657902-165657902 on Build GRCh38
rs1043700504 Chr.2: 165658163-165658163 on Build GRCh38
rs999071289 Chr.2: 165658030-165658030 on Build GRCh38
rs373310534 Chr.2: 165657758-165657758 on Build GRCh38
rs1010344715 Chr.2: 165657732-165657732 on Build GRCh38
rs758387079 Chr.2: 165657873-165657873 on Build GRCh38
rs750576720 Chr.2: 165657857-165657857 on Build GRCh38
rs892277448 Chr.2: 165657723-165657723 on Build GRCh38
rs757707970 Chr.2: 165658049-165658050 on Build GRCh38
rs761597012 Chr.2: 165658067-165658067 on Build GRCh38
rs781384120 Chr.2: 165657964-165657964 on Build GRCh38
rs113015888 Chr.2: 165658170-165658170 on Build GRCh38

View Full Product Details