Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 39136348-39136947 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GEMIN2
Entrez Gene ID
8487
Gene Name
gem nuclear organelle associated protein 2
Gene Aliases
SIP1, SIP1-delta
UniGene
Hs.652307
Gene Chromosome Location
Chr.14: 39114261-39136973 on Build GRCh38
Transcripts
NM_001009182.1 NM_001009183.1 NM_003616.2 XM_017021709.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4889523 c.839C>A p.S280Y Chr.14: 39136475-39136475 on Build GRCh38
6289553 c.836C>T p.P279L Chr.14: 39136472-39136472 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs752213455 Chr.14: 39136479-39136479 on Build GRCh38
rs538117439 Chr.14: 39136693-39136693 on Build GRCh38
rs375268766 Chr.14: 39136424-39136425 on Build GRCh38
rs578052393 Chr.14: 39136853-39136853 on Build GRCh38
rs759863090 Chr.14: 39136827-39136827 on Build GRCh38
rs759581171 Chr.14: 39136416-39136416 on Build GRCh38
rs143390356 Chr.14: 39136468-39136468 on Build GRCh38
rs912682437 Chr.14: 39136652-39136652 on Build GRCh38
rs868349490 Chr.14: 39136585-39136585 on Build GRCh38
rs762722515 Chr.14: 39136468-39136469 on Build GRCh38
rs1006838894 Chr.14: 39136915-39136915 on Build GRCh38
rs374547314 Chr.14: 39136862-39136865 on Build GRCh38
rs758525626 Chr.14: 39136401-39136401 on Build GRCh38
rs982671447 Chr.14: 39136824-39136824 on Build GRCh38
rs146999348 Chr.14: 39136418-39136419 on Build GRCh38
rs761845092 Chr.14: 39136425-39136425 on Build GRCh38
rs755640581 Chr.14: 39136483-39136483 on Build GRCh38
rs766021119 Chr.14: 39136513-39136514 on Build GRCh38
rs542525433 Chr.14: 39136776-39136778 on Build GRCh38
rs536378262 Chr.14: 39136609-39136609 on Build GRCh38
rs567034887 Chr.14: 39136481-39136481 on Build GRCh38
rs757580828 Chr.14: 39136623-39136623 on Build GRCh38
rs975293673 Chr.14: 39136775-39136775 on Build GRCh38
rs967001437 Chr.14: 39136634-39136634 on Build GRCh38
rs941484993 Chr.14: 39136410-39136410 on Build GRCh38
rs991380959 Chr.14: 39136731-39136731 on Build GRCh38
rs770775640 Chr.14: 39136404-39136404 on Build GRCh38
rs748979281 Chr.14: 39136500-39136500 on Build GRCh38
rs939606236 Chr.14: 39136685-39136685 on Build GRCh38
rs142465191 Chr.14: 39136453-39136453 on Build GRCh38
rs968359008 Chr.14: 39136537-39136537 on Build GRCh38
rs1020455872 Chr.14: 39136586-39136586 on Build GRCh38
rs778874791 Chr.14: 39136517-39136517 on Build GRCh38
rs764882305 Chr.14: 39136425-39136429 on Build GRCh38
rs772086435 Chr.14: 39136421-39136421 on Build GRCh38
rs554244152 Chr.14: 39136912-39136912 on Build GRCh38
rs957336484 Chr.14: 39136464-39136464 on Build GRCh38
rs765445794 Chr.14: 39136439-39136439 on Build GRCh38
rs759668755 Chr.14: 39136418-39136418 on Build GRCh38
rs897141508 Chr.14: 39136417-39136417 on Build GRCh38
rs192707772 Chr.14: 39136425-39136425 on Build GRCh38
rs543243298 Chr.14: 39136879-39136879 on Build GRCh38
rs751872461 Chr.14: 39136454-39136454 on Build GRCh38
rs777092867 Chr.14: 39136495-39136495 on Build GRCh38
rs1029498798 Chr.14: 39136434-39136434 on Build GRCh38
rs76588905 Chr.14: 39136642-39136642 on Build GRCh38
rs1018132220 Chr.14: 39136460-39136460 on Build GRCh38
rs938293787 Chr.14: 39136847-39136847 on Build GRCh38
rs941541068 Chr.14: 39136805-39136805 on Build GRCh38
rs755523946 Chr.14: 39136465-39136465 on Build GRCh38
rs986856478 Chr.14: 39136649-39136649 on Build GRCh38
rs750670217 Chr.14: 39136441-39136441 on Build GRCh38
rs750217852 Chr.14: 39136450-39136452 on Build GRCh38
rs1046754903 Chr.14: 39136423-39136424 on Build GRCh38
rs147141713 Chr.14: 39136621-39136621 on Build GRCh38
rs928198438 Chr.14: 39136664-39136664 on Build GRCh38
rs1001497375 Chr.14: 39136544-39136544 on Build GRCh38
rs756823342 Chr.14: 39136509-39136509 on Build GRCh38
rs1038418906 Chr.14: 39136819-39136819 on Build GRCh38
rs1024205202 Chr.14: 39136536-39136536 on Build GRCh38
rs745636046 Chr.14: 39136525-39136525 on Build GRCh38
rs376097663 Chr.14: 39136447-39136447 on Build GRCh38
rs776735117 Chr.14: 39136661-39136661 on Build GRCh38
rs964475995 Chr.14: 39136771-39136771 on Build GRCh38
rs752102448 Chr.14: 39136592-39136592 on Build GRCh38
rs576320425 Chr.14: 39136480-39136480 on Build GRCh38
rs774225790 Chr.14: 39136412-39136412 on Build GRCh38
rs749098074 Chr.14: 39136399-39136399 on Build GRCh38
rs1002641444 Chr.14: 39136446-39136446 on Build GRCh38
rs1030980618 Chr.14: 39136638-39136638 on Build GRCh38
rs558432253 Chr.14: 39136754-39136754 on Build GRCh38
rs918680863 Chr.14: 39136821-39136821 on Build GRCh38
rs78082091 Chr.14: 39136381-39136381 on Build GRCh38
rs761923594 Chr.14: 39136435-39136435 on Build GRCh38
rs184121106 Chr.14: 39136890-39136890 on Build GRCh38
rs564743517 Chr.14: 39136858-39136861 on Build GRCh38

View Full Product Details