Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7439105-7439653 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF11
Entrez Gene ID
2256
Gene Name
fibroblast growth factor 11
Gene Aliases
FGF-11, FHF-3, FHF3
UniGene
Hs.655193
Gene Chromosome Location
Chr.17: 7438273-7444932 on Build GRCh38
Transcripts
NM_004112.3 NR_130156.1

SNP Detail

SNP ID Chromosome Location
rs1011969510 Chr.17: 7439561-7439561 on Build GRCh38
rs11869879 Chr.17: 7439487-7439487 on Build GRCh38
rs960650525 Chr.17: 7439477-7439477 on Build GRCh38
rs979340691 Chr.17: 7439250-7439250 on Build GRCh38
rs940308366 Chr.17: 7439519-7439519 on Build GRCh38
rs35567432 Chr.17: 7439474-7439475 on Build GRCh38
rs754716185 Chr.17: 7439617-7439617 on Build GRCh38
rs1028259893 Chr.17: 7439129-7439129 on Build GRCh38
rs766061155 Chr.17: 7439601-7439601 on Build GRCh38
rs371867789 Chr.17: 7439473-7439473 on Build GRCh38
rs931536156 Chr.17: 7439399-7439399 on Build GRCh38
rs925077920 Chr.17: 7439538-7439538 on Build GRCh38
rs764171007 Chr.17: 7439321-7439321 on Build GRCh38
rs4151123 Chr.17: 7439384-7439384 on Build GRCh38
rs948713887 Chr.17: 7439370-7439370 on Build GRCh38
rs945067512 Chr.17: 7439224-7439224 on Build GRCh38
rs953675222 Chr.17: 7439135-7439135 on Build GRCh38
rs1023437760 Chr.17: 7439208-7439208 on Build GRCh38
rs775735686 Chr.17: 7439181-7439181 on Build GRCh38
rs369618854 Chr.17: 7439629-7439629 on Build GRCh38
rs573356333 Chr.17: 7439314-7439314 on Build GRCh38
rs561289676 Chr.17: 7439583-7439583 on Build GRCh38
rs957756842 Chr.17: 7439566-7439566 on Build GRCh38
rs533536833 Chr.17: 7439245-7439245 on Build GRCh38
rs752546716 Chr.17: 7439624-7439624 on Build GRCh38
rs1000730306 Chr.17: 7439600-7439600 on Build GRCh38
rs923834440 Chr.17: 7439465-7439465 on Build GRCh38
rs942772073 Chr.17: 7439331-7439331 on Build GRCh38
rs1023405227 Chr.17: 7439580-7439580 on Build GRCh38
rs781108074 Chr.17: 7439619-7439619 on Build GRCh38
rs923952730 Chr.17: 7439267-7439267 on Build GRCh38
rs1056139366 Chr.17: 7439522-7439522 on Build GRCh38
rs756128069 Chr.17: 7439625-7439625 on Build GRCh38
rs995376027 Chr.17: 7439472-7439472 on Build GRCh38
rs572456213 Chr.17: 7439176-7439176 on Build GRCh38
rs549356495 Chr.17: 7439518-7439518 on Build GRCh38
rs554954557 Chr.17: 7439291-7439291 on Build GRCh38
rs767806727 Chr.17: 7439149-7439149 on Build GRCh38
rs545215677 Chr.17: 7439439-7439439 on Build GRCh38
rs1045746532 Chr.17: 7439379-7439379 on Build GRCh38
rs531798531 Chr.17: 7439610-7439610 on Build GRCh38
rs987172949 Chr.17: 7439616-7439616 on Build GRCh38
rs920110688 Chr.17: 7439392-7439392 on Build GRCh38
rs9892818 Chr.17: 7439403-7439403 on Build GRCh38
rs4151122 Chr.17: 7439363-7439363 on Build GRCh38
rs1055096772 Chr.17: 7439358-7439358 on Build GRCh38
rs1019256924 Chr.17: 7439502-7439502 on Build GRCh38
rs967914070 Chr.17: 7439227-7439227 on Build GRCh38
rs200324264 Chr.17: 7439476-7439476 on Build GRCh38
rs896210432 Chr.17: 7439553-7439553 on Build GRCh38
rs543993003 Chr.17: 7439316-7439316 on Build GRCh38
rs796718632 Chr.17: 7439466-7439466 on Build GRCh38
rs898300782 Chr.17: 7439448-7439448 on Build GRCh38
rs912308300 Chr.17: 7439213-7439213 on Build GRCh38
rs1004673167 Chr.17: 7439394-7439394 on Build GRCh38
rs986901212 Chr.17: 7439309-7439309 on Build GRCh38
rs560464724 Chr.17: 7439467-7439467 on Build GRCh38
rs865906808 Chr.17: 7439466-7439466 on Build GRCh38

View Full Product Details